Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 94469474-94469723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERPINA9
Entrez Gene ID
327657
Gene Name
serpin family A member 9
Gene Aliases
GCET1, SERPINA11, SERPINA11b
UniGene
Hs.317970
Gene Chromosome Location
Chr.14: 94462721-94476333 on Build GRCh38
Transcripts
NM_001042518.1 NM_001284275.1 NM_001284276.1 NM_175739.3 XM_011536714.2 XM_011536715.2 XM_011536716.2 XM_011536717.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1749046 c.252G>A p.Q84Q Chr.14: 94469643-94469643 on Build GRCh38
6457052 c.288C>T p.S96S Chr.14: 94469607-94469607 on Build GRCh38
7237287 c.287C>T p.S96F Chr.14: 94469608-94469608 on Build GRCh38
5382953 c.302C>T p.S101F Chr.14: 94469593-94469593 on Build GRCh38
6457054 c.180C>T p.S60S Chr.14: 94469607-94469607 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765359461 Chr.14: 94469589-94469589 on Build GRCh38
rs759598527 Chr.14: 94469582-94469582 on Build GRCh38
rs745486048 Chr.14: 94469549-94469549 on Build GRCh38
rs200602156 Chr.14: 94469697-94469697 on Build GRCh38
rs773688275 Chr.14: 94469637-94469637 on Build GRCh38
rs12879019 Chr.14: 94469564-94469564 on Build GRCh38
rs774654639 Chr.14: 94469565-94469565 on Build GRCh38
rs371331582 Chr.14: 94469515-94469515 on Build GRCh38
rs760492022 Chr.14: 94469576-94469576 on Build GRCh38
rs200371954 Chr.14: 94469649-94469649 on Build GRCh38
rs370684428 Chr.14: 94469506-94469506 on Build GRCh38
rs747519683 Chr.14: 94469695-94469695 on Build GRCh38
rs917893516 Chr.14: 94469587-94469587 on Build GRCh38
rs760418567 Chr.14: 94469590-94469591 on Build GRCh38
rs760946865 Chr.14: 94469578-94469578 on Build GRCh38
rs771270945 Chr.14: 94469647-94469647 on Build GRCh38
rs541943122 Chr.14: 94469630-94469632 on Build GRCh38
rs769479968 Chr.14: 94469552-94469552 on Build GRCh38
rs755752926 Chr.14: 94469612-94469612 on Build GRCh38
rs767993261 Chr.14: 94469625-94469625 on Build GRCh38
rs748563339 Chr.14: 94469635-94469635 on Build GRCh38
rs142619133 Chr.14: 94469579-94469579 on Build GRCh38
rs372208985 Chr.14: 94469577-94469577 on Build GRCh38
rs756426418 Chr.14: 94469496-94469496 on Build GRCh38
rs758602903 Chr.14: 94469686-94469686 on Build GRCh38
rs774962208 Chr.14: 94469563-94469563 on Build GRCh38
rs59773088 Chr.14: 94469648-94469648 on Build GRCh38
rs761176457 Chr.14: 94469639-94469639 on Build GRCh38
rs758331597 Chr.14: 94469507-94469507 on Build GRCh38
rs374230953 Chr.14: 94469551-94469551 on Build GRCh38
rs371625465 Chr.14: 94469687-94469687 on Build GRCh38
rs761355901 Chr.14: 94469703-94469704 on Build GRCh38
rs767504507 Chr.14: 94469670-94469670 on Build GRCh38
rs766525059 Chr.14: 94469499-94469499 on Build GRCh38
rs1047330150 Chr.14: 94469653-94469653 on Build GRCh38
rs572837692 Chr.14: 94469650-94469650 on Build GRCh38
rs761718690 Chr.14: 94469654-94469654 on Build GRCh38
rs750270201 Chr.14: 94469607-94469607 on Build GRCh38
rs575042815 Chr.14: 94469636-94469636 on Build GRCh38
rs780991821 Chr.14: 94469533-94469533 on Build GRCh38
rs377551906 Chr.14: 94469512-94469512 on Build GRCh38
rs544061591 Chr.14: 94469693-94469693 on Build GRCh38
rs375058481 Chr.14: 94469508-94469508 on Build GRCh38
rs190620890 Chr.14: 94469680-94469680 on Build GRCh38
rs990378863 Chr.14: 94469602-94469602 on Build GRCh38
rs375039631 Chr.14: 94469694-94469694 on Build GRCh38
rs556808192 Chr.14: 94469634-94469634 on Build GRCh38
rs751390983 Chr.14: 94469591-94469591 on Build GRCh38
rs201925577 Chr.14: 94469568-94469568 on Build GRCh38
rs199573923 Chr.14: 94469671-94469671 on Build GRCh38
rs139400221 Chr.14: 94469509-94469509 on Build GRCh38
rs757220526 Chr.14: 94469603-94469603 on Build GRCh38
rs1034288024 Chr.14: 94469572-94469572 on Build GRCh38
rs748977210 Chr.14: 94469618-94469618 on Build GRCh38
rs780938756 Chr.14: 94469606-94469606 on Build GRCh38
rs753937955 Chr.14: 94469581-94469581 on Build GRCh38
rs755841892 Chr.14: 94469672-94469672 on Build GRCh38
rs763737866 Chr.14: 94469597-94469598 on Build GRCh38
rs201522333 Chr.14: 94469681-94469681 on Build GRCh38
rs915932148 Chr.14: 94469617-94469617 on Build GRCh38

View Full Product Details