Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 68310728-68310983 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRMT7
Entrez Gene ID
54496
Gene Name
protein arginine methyltransferase 7
Gene Aliases
-
UniGene
Hs.640229
Gene Chromosome Location
Chr.16: 68310974-68358832 on Build GRCh38
Transcripts
NM_001184824.1 NM_019023.2 XM_011523112.2 XM_011523113.2 XM_011523116.2 XM_017023290.1 XM_017023293.1 XM_017023295.1 XM_017023296.1 XM_017023297.1 XM_017023298.1 XM_017023299.1 XM_017023300.1 XM_017023301.1 XM_017023304.1 XM_017023306.1 XM_017023312.1 NM_032178.2 XM_011523372.2

Gene Symbol
SLC7A6OS
Entrez Gene ID
84138
Gene Name
solute carrier family 7 member 6 opposite strand
Gene Aliases
-
UniGene
Hs.729520
Gene Chromosome Location
Chr.16: 68300615-68310965 on Build GRCh38
Transcripts
NM_001184824.1 NM_019023.2 XM_011523112.2 XM_011523113.2 XM_011523116.2 XM_017023290.1 XM_017023293.1 XM_017023295.1 XM_017023296.1 XM_017023297.1 XM_017023298.1 XM_017023299.1 XM_017023300.1 XM_017023301.1 XM_017023304.1 XM_017023306.1 XM_017023312.1 NM_032178.2 XM_011523372.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
301396 c.97G>A p.D33N Chr.16: 68310830-68310830 on Build GRCh38
7313251 c.6G>T p.E2D Chr.16: 68310921-68310921 on Build GRCh38
7279803 c.118C>A p.Q40K Chr.16: 68310809-68310809 on Build GRCh38
435576 c.134G>A p.G45D Chr.16: 68310793-68310793 on Build GRCh38
5851301 c.143G>A p.R48K Chr.16: 68310784-68310784 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763381823 Chr.16: 68310778-68310778 on Build GRCh38
rs771157870 Chr.16: 68310918-68310918 on Build GRCh38
rs751371070 Chr.16: 68310817-68310817 on Build GRCh38
rs374455092 Chr.16: 68310868-68310868 on Build GRCh38
rs767273041 Chr.16: 68310943-68310943 on Build GRCh38
rs779496516 Chr.16: 68310907-68310907 on Build GRCh38
rs910802650 Chr.16: 68310761-68310761 on Build GRCh38
rs866112460 Chr.16: 68310916-68310916 on Build GRCh38
rs775821973 Chr.16: 68310756-68310756 on Build GRCh38
rs749802854 Chr.16: 68310752-68310752 on Build GRCh38
rs140742717 Chr.16: 68310782-68310782 on Build GRCh38
rs535783486 Chr.16: 68310836-68310836 on Build GRCh38
rs751424036 Chr.16: 68310881-68310881 on Build GRCh38
rs755777695 Chr.16: 68310897-68310897 on Build GRCh38
rs906511103 Chr.16: 68310841-68310841 on Build GRCh38
rs765247208 Chr.16: 68310812-68310812 on Build GRCh38
rs1016132635 Chr.16: 68310910-68310910 on Build GRCh38
rs745415170 Chr.16: 68310838-68310838 on Build GRCh38
rs757020969 Chr.16: 68310822-68310822 on Build GRCh38
rs147968885 Chr.16: 68310815-68310815 on Build GRCh38
rs755527642 Chr.16: 68310911-68310911 on Build GRCh38
rs764104816 Chr.16: 68310875-68310875 on Build GRCh38
rs760618475 Chr.16: 68310801-68310801 on Build GRCh38
rs750562529 Chr.16: 68310795-68310795 on Build GRCh38
rs201990110 Chr.16: 68310874-68310874 on Build GRCh38
rs886586833 Chr.16: 68310925-68310925 on Build GRCh38
rs769119499 Chr.16: 68310849-68310849 on Build GRCh38
rs138348680 Chr.16: 68310914-68310914 on Build GRCh38
rs753952848 Chr.16: 68310871-68310871 on Build GRCh38
rs900173764 Chr.16: 68310906-68310906 on Build GRCh38
rs147089863 Chr.16: 68310861-68310861 on Build GRCh38
rs1039077727 Chr.16: 68310900-68310900 on Build GRCh38
rs35800405 Chr.16: 68310857-68310857 on Build GRCh38
rs769145053 Chr.16: 68310855-68310855 on Build GRCh38
rs747263697 Chr.16: 68310915-68310915 on Build GRCh38
rs200733664 Chr.16: 68310784-68310784 on Build GRCh38
rs773262305 Chr.16: 68310863-68310863 on Build GRCh38
rs762087304 Chr.16: 68310792-68310792 on Build GRCh38
rs987712370 Chr.16: 68310751-68310751 on Build GRCh38
rs755965231 Chr.16: 68310760-68310761 on Build GRCh38
rs774366753 Chr.16: 68310929-68310929 on Build GRCh38
rs537044509 Chr.16: 68310944-68310944 on Build GRCh38
rs764158334 Chr.16: 68310922-68310922 on Build GRCh38
rs749987906 Chr.16: 68310896-68310896 on Build GRCh38
rs576156259 Chr.16: 68310920-68310920 on Build GRCh38
rs754441241 Chr.16: 68310961-68310961 on Build GRCh38
rs372855610 Chr.16: 68310858-68310858 on Build GRCh38
rs867759910 Chr.16: 68310794-68310794 on Build GRCh38
rs779341752 Chr.16: 68310912-68310912 on Build GRCh38
rs757079912 Chr.16: 68310883-68310883 on Build GRCh38
rs1026496197 Chr.16: 68310951-68310951 on Build GRCh38
rs780623572 Chr.16: 68310850-68310850 on Build GRCh38
rs767203433 Chr.16: 68310885-68310885 on Build GRCh38
rs3803650 Chr.16: 68310793-68310793 on Build GRCh38
rs770358492 Chr.16: 68310750-68310750 on Build GRCh38
rs58760922 Chr.16: 68310923-68310923 on Build GRCh38
rs777780271 Chr.16: 68310814-68310814 on Build GRCh38
rs796322429 Chr.16: 68310877-68310877 on Build GRCh38
rs199568667 Chr.16: 68310813-68310813 on Build GRCh38
rs748763387 Chr.16: 68310909-68310909 on Build GRCh38
rs753365698 Chr.16: 68310956-68310956 on Build GRCh38
rs772301434 Chr.16: 68310862-68310862 on Build GRCh38
rs766031584 Chr.16: 68310953-68310953 on Build GRCh38
rs761715941 Chr.16: 68310939-68310939 on Build GRCh38
rs748540635 Chr.16: 68310913-68310913 on Build GRCh38
rs947172815 Chr.16: 68310879-68310879 on Build GRCh38

View Full Product Details