Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 75789044-75789295 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RABGGTB
Entrez Gene ID
5876
Gene Name
Rab geranylgeranyltransferase beta subunit
Gene Aliases
GGTB
UniGene
Hs.78948
Gene Chromosome Location
Chr.1: 75786194-75795090 on Build GRCh38
Transcripts
NM_004582.3 NR_073562.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5799967 c.208G>T p.E70* Chr.1: 75789255-75789255 on Build GRCh38
6761725 c.172G>A p.D58N Chr.1: 75789219-75789219 on Build GRCh38
5499762 c.193C>T p.R65C Chr.1: 75789240-75789240 on Build GRCh38
1731412 c.171G>T p.M57I Chr.1: 75789218-75789218 on Build GRCh38
5783651 c.152A>G p.Y51C Chr.1: 75789199-75789199 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761041672 Chr.1: 75789195-75789195 on Build GRCh38
rs999306337 Chr.1: 75789133-75789133 on Build GRCh38
rs541607406 Chr.1: 75789104-75789104 on Build GRCh38
rs774986714 Chr.1: 75789126-75789126 on Build GRCh38
rs1044185678 Chr.1: 75789096-75789096 on Build GRCh38
rs371350101 Chr.1: 75789240-75789240 on Build GRCh38
rs776717177 Chr.1: 75789144-75789145 on Build GRCh38
rs905687303 Chr.1: 75789106-75789106 on Build GRCh38
rs751553941 Chr.1: 75789208-75789208 on Build GRCh38
rs772672802 Chr.1: 75789103-75789104 on Build GRCh38
rs753976465 Chr.1: 75789200-75789200 on Build GRCh38
rs369023722 Chr.1: 75789206-75789206 on Build GRCh38
rs773290435 Chr.1: 75789194-75789194 on Build GRCh38
rs767976466 Chr.1: 75789188-75789188 on Build GRCh38
rs760573112 Chr.1: 75789140-75789140 on Build GRCh38
rs768198292 Chr.1: 75789266-75789266 on Build GRCh38
rs772058727 Chr.1: 75789149-75789149 on Build GRCh38
rs549019085 Chr.1: 75789143-75789144 on Build GRCh38
rs755777439 Chr.1: 75789247-75789247 on Build GRCh38
rs994540881 Chr.1: 75789176-75789176 on Build GRCh38
rs960438279 Chr.1: 75789145-75789145 on Build GRCh38
rs947151342 Chr.1: 75789082-75789082 on Build GRCh38
rs199755301 Chr.1: 75789135-75789135 on Build GRCh38
rs762056681 Chr.1: 75789164-75789164 on Build GRCh38
rs879885436 Chr.1: 75789226-75789226 on Build GRCh38
rs769177762 Chr.1: 75789142-75789142 on Build GRCh38
rs533455235 Chr.1: 75789199-75789199 on Build GRCh38
rs756978562 Chr.1: 75789217-75789217 on Build GRCh38
rs1023177203 Chr.1: 75789209-75789209 on Build GRCh38
rs375918526 Chr.1: 75789248-75789248 on Build GRCh38
rs764717220 Chr.1: 75789148-75789150 on Build GRCh38
rs748832608 Chr.1: 75789250-75789250 on Build GRCh38
rs769290302 Chr.1: 75789120-75789120 on Build GRCh38
rs773276807 Chr.1: 75789204-75789204 on Build GRCh38
rs745594464 Chr.1: 75789111-75789111 on Build GRCh38
rs750012267 Chr.1: 75789241-75789241 on Build GRCh38

View Full Product Details