Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 94448082-94448353 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERPINA11
Entrez Gene ID
256394
Gene Name
serpin family A member 11
Gene Aliases
-
UniGene
Hs.145890
Gene Chromosome Location
Chr.14: 94442438-94452857 on Build GRCh38
Transcripts
NM_001080451.1 XM_006720105.3

SNP Detail

SNP ID Chromosome Location
rs376367509 Chr.14: 94448207-94448207 on Build GRCh38
rs748263368 Chr.14: 94448310-94448310 on Build GRCh38
rs200680940 Chr.14: 94448281-94448281 on Build GRCh38
rs140179275 Chr.14: 94448171-94448171 on Build GRCh38
rs139522311 Chr.14: 94448262-94448262 on Build GRCh38
rs375293485 Chr.14: 94448271-94448271 on Build GRCh38
rs141809191 Chr.14: 94448327-94448327 on Build GRCh38
rs143070250 Chr.14: 94448152-94448152 on Build GRCh38
rs757766017 Chr.14: 94448172-94448172 on Build GRCh38
rs755817954 Chr.14: 94448136-94448138 on Build GRCh38
rs146841701 Chr.14: 94448208-94448208 on Build GRCh38
rs756265934 Chr.14: 94448186-94448186 on Build GRCh38
rs879179014 Chr.14: 94448226-94448226 on Build GRCh38
rs764790365 Chr.14: 94448205-94448205 on Build GRCh38
rs759074538 Chr.14: 94448159-94448159 on Build GRCh38
rs527979379 Chr.14: 94448326-94448326 on Build GRCh38
rs61738925 Chr.14: 94448330-94448330 on Build GRCh38
rs143460128 Chr.14: 94448211-94448211 on Build GRCh38
rs188994371 Chr.14: 94448167-94448167 on Build GRCh38
rs776243828 Chr.14: 94448199-94448199 on Build GRCh38
rs745372962 Chr.14: 94448117-94448117 on Build GRCh38
rs746881580 Chr.14: 94448194-94448194 on Build GRCh38
rs781496248 Chr.14: 94448180-94448180 on Build GRCh38
rs773413735 Chr.14: 94448225-94448225 on Build GRCh38
rs760760047 Chr.14: 94448280-94448280 on Build GRCh38
rs114327150 Chr.14: 94448162-94448162 on Build GRCh38
rs751099762 Chr.14: 94448218-94448218 on Build GRCh38
rs780127162 Chr.14: 94448290-94448290 on Build GRCh38
rs768915706 Chr.14: 94448193-94448193 on Build GRCh38
rs76289690 Chr.14: 94448297-94448297 on Build GRCh38
rs775380903 Chr.14: 94448116-94448116 on Build GRCh38
rs752106331 Chr.14: 94448185-94448185 on Build GRCh38
rs993393778 Chr.14: 94448124-94448124 on Build GRCh38
rs116380145 Chr.14: 94448189-94448189 on Build GRCh38
rs760533993 Chr.14: 94448149-94448149 on Build GRCh38
rs1017352721 Chr.14: 94448187-94448187 on Build GRCh38
rs372977604 Chr.14: 94448300-94448300 on Build GRCh38
rs148495961 Chr.14: 94448131-94448131 on Build GRCh38
rs768060913 Chr.14: 94448239-94448239 on Build GRCh38
rs143826161 Chr.14: 94448184-94448184 on Build GRCh38
rs75949282 Chr.14: 94448292-94448292 on Build GRCh38
rs757990411 Chr.14: 94448166-94448166 on Build GRCh38
rs749325818 Chr.14: 94448192-94448192 on Build GRCh38
rs770084681 Chr.14: 94448115-94448115 on Build GRCh38
rs967373475 Chr.14: 94448291-94448291 on Build GRCh38
rs116079830 Chr.14: 94448195-94448195 on Build GRCh38
rs184631809 Chr.14: 94448153-94448153 on Build GRCh38
rs10129672 Chr.14: 94448118-94448118 on Build GRCh38
rs146165398 Chr.14: 94448170-94448170 on Build GRCh38
rs554934752 Chr.14: 94448135-94448135 on Build GRCh38
rs757710820 Chr.14: 94448238-94448238 on Build GRCh38
rs746010310 Chr.14: 94448106-94448106 on Build GRCh38
rs897603517 Chr.14: 94448304-94448304 on Build GRCh38
rs759412188 Chr.14: 94448200-94448200 on Build GRCh38
rs201110965 Chr.14: 94448198-94448198 on Build GRCh38

View Full Product Details