Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 13295401-13295642 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHYH
Entrez Gene ID
5264
Gene Name
phytanoyl-CoA 2-hydroxylase
Gene Aliases
LN1, LNAP1, PAHX, PHYH1, RD
UniGene
Hs.498732
Gene Chromosome Location
Chr.10: 13277796-13300130 on Build GRCh38
Transcripts
NM_001037537.1 NM_001323080.1 NM_001323082.1 NM_001323083.1 NM_001323084.1 NM_006214.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3435751 c.227C>T p.A76V Chr.10: 13295514-13295514 on Build GRCh38
2023081 c.175A>C p.K59Q Chr.10: 13295566-13295566 on Build GRCh38
3435752 c.207C>T p.I69I Chr.10: 13295534-13295534 on Build GRCh38
2023082 c.166G>T p.E56* Chr.10: 13295575-13295575 on Build GRCh38
6432340 c.236A>G p.Q79R Chr.10: 13295505-13295505 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771973307 Chr.10: 13295527-13295527 on Build GRCh38
rs369671316 Chr.10: 13295606-13295606 on Build GRCh38
rs767030794 Chr.10: 13295593-13295593 on Build GRCh38
rs145404396 Chr.10: 13295497-13295497 on Build GRCh38
rs143820757 Chr.10: 13295535-13295535 on Build GRCh38
rs886046830 Chr.10: 13295496-13295496 on Build GRCh38
rs730882058 Chr.10: 13295577-13295577 on Build GRCh38
rs760606440 Chr.10: 13295454-13295454 on Build GRCh38
rs758923121 Chr.10: 13295467-13295467 on Build GRCh38
rs1005302176 Chr.10: 13295424-13295424 on Build GRCh38
rs1030341153 Chr.10: 13295519-13295519 on Build GRCh38
rs754281074 Chr.10: 13295601-13295601 on Build GRCh38
rs976571180 Chr.10: 13295496-13295496 on Build GRCh38
rs749408911 Chr.10: 13295510-13295510 on Build GRCh38
rs756504018 Chr.10: 13295590-13295590 on Build GRCh38
rs766238923 Chr.10: 13295456-13295456 on Build GRCh38
rs978740649 Chr.10: 13295578-13295578 on Build GRCh38
rs775279867 Chr.10: 13295580-13295580 on Build GRCh38
rs200937622 Chr.10: 13295465-13295465 on Build GRCh38
rs779247277 Chr.10: 13295513-13295513 on Build GRCh38
rs764973812 Chr.10: 13295471-13295471 on Build GRCh38
rs757696181 Chr.10: 13295492-13295492 on Build GRCh38
rs773196580 Chr.10: 13295446-13295446 on Build GRCh38
rs759133541 Chr.10: 13295562-13295562 on Build GRCh38
rs763777635 Chr.10: 13295582-13295582 on Build GRCh38
rs141855372 Chr.10: 13295514-13295514 on Build GRCh38
rs752268595 Chr.10: 13295473-13295473 on Build GRCh38
rs764637574 Chr.10: 13295568-13295568 on Build GRCh38
rs369205198 Chr.10: 13295503-13295503 on Build GRCh38
rs141952441 Chr.10: 13295604-13295604 on Build GRCh38
rs888134966 Chr.10: 13295423-13295423 on Build GRCh38
rs201578674 Chr.10: 13295608-13295608 on Build GRCh38
rs1057272016 Chr.10: 13295607-13295607 on Build GRCh38
rs756698271 Chr.10: 13295502-13295502 on Build GRCh38
rs776512475 Chr.10: 13295558-13295558 on Build GRCh38
rs180913758 Chr.10: 13295512-13295512 on Build GRCh38
rs145000664 Chr.10: 13295549-13295549 on Build GRCh38
rs1747682 Chr.10: 13295588-13295588 on Build GRCh38
rs886046831 Chr.10: 13295565-13295565 on Build GRCh38
rs746748064 Chr.10: 13295538-13295538 on Build GRCh38
rs947426619 Chr.10: 13295565-13295565 on Build GRCh38
rs964140486 Chr.10: 13295466-13295466 on Build GRCh38

View Full Product Details