Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 24066043-24066274 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC39C
Entrez Gene ID
125488
Gene Name
tetratricopeptide repeat domain 39C
Gene Aliases
C18orf17, HsT2697
UniGene
Hs.128576
Gene Chromosome Location
Chr.18: 23992773-24135952 on Build GRCh38
Transcripts
NM_001135993.1 NM_153211.3 XM_005258198.4 XM_006722392.3 XM_011525814.2 XM_017025550.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6841010 c.162+1G>A p.? Chr.18: 24066141-24066141 on Build GRCh38
1268856 c.141C>G p.I47M Chr.18: 24066119-24066119 on Build GRCh38
6003313 c.112A>G p.S38G Chr.18: 24066090-24066090 on Build GRCh38
473693 c.136A>G p.T46A Chr.18: 24066114-24066114 on Build GRCh38
3371019 c.121G>A p.A41T Chr.18: 24066099-24066099 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755209601 Chr.18: 24066151-24066151 on Build GRCh38
rs865897706 Chr.18: 24066083-24066083 on Build GRCh38
rs199669752 Chr.18: 24066211-24066211 on Build GRCh38
rs752792788 Chr.18: 24066159-24066159 on Build GRCh38
rs774002518 Chr.18: 24066098-24066098 on Build GRCh38
rs781150730 Chr.18: 24066170-24066170 on Build GRCh38
rs929915905 Chr.18: 24066174-24066174 on Build GRCh38
rs769514098 Chr.18: 24066084-24066084 on Build GRCh38
rs769615780 Chr.18: 24066188-24066188 on Build GRCh38
rs151171824 Chr.18: 24066074-24066074 on Build GRCh38
rs8096969 Chr.18: 24066219-24066219 on Build GRCh38
rs370006797 Chr.18: 24066144-24066144 on Build GRCh38
rs770928753 Chr.18: 24066066-24066066 on Build GRCh38
rs138298971 Chr.18: 24066095-24066095 on Build GRCh38
rs372849552 Chr.18: 24066168-24066168 on Build GRCh38
rs200341803 Chr.18: 24066178-24066178 on Build GRCh38
rs768092769 Chr.18: 24066134-24066134 on Build GRCh38
rs756471825 Chr.18: 24066141-24066141 on Build GRCh38
rs925032923 Chr.18: 24066209-24066209 on Build GRCh38
rs775201911 Chr.18: 24066085-24066085 on Build GRCh38
rs762751675 Chr.18: 24066088-24066088 on Build GRCh38
rs61734884 Chr.18: 24066140-24066140 on Build GRCh38
rs778901933 Chr.18: 24066152-24066152 on Build GRCh38
rs776235873 Chr.18: 24066072-24066072 on Build GRCh38
rs1018520881 Chr.18: 24066166-24066166 on Build GRCh38
rs780006835 Chr.18: 24066189-24066189 on Build GRCh38
rs8083357 Chr.18: 24066125-24066125 on Build GRCh38
rs549868577 Chr.18: 24066147-24066147 on Build GRCh38

View Full Product Details