Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 19355367-19355612 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDHA1
Entrez Gene ID
5160
Gene Name
pyruvate dehydrogenase (lipoamide) alpha 1
Gene Aliases
PDHA, PDHAD, PDHCE1A, PHE1A
UniGene
Hs.530331
Gene Chromosome Location
Chr.X: 19343893-19361707 on Build GRCh38
Transcripts
NM_000284.3 NM_001173454.1 NM_001173455.1 NM_001173456.1 XM_017029574.1

SNP Detail

SNP ID Chromosome Location
rs776832103 Chr.X: 19355552-19355552 on Build GRCh38
rs745992562 Chr.X: 19355529-19355530 on Build GRCh38
rs763679737 Chr.X: 19355538-19355538 on Build GRCh38
rs375488072 Chr.X: 19355514-19355514 on Build GRCh38
rs769735500 Chr.X: 19355530-19355541 on Build GRCh38
rs137853255 Chr.X: 19355472-19355472 on Build GRCh38
rs878905548 Chr.X: 19355573-19355573 on Build GRCh38
rs752319005 Chr.X: 19355534-19355534 on Build GRCh38
rs778268006 Chr.X: 19355521-19355522 on Build GRCh38
rs929262350 Chr.X: 19355408-19355408 on Build GRCh38
rs121917898 Chr.X: 19355393-19355393 on Build GRCh38
rs767940407 Chr.X: 19355533-19355533 on Build GRCh38
rs764871850 Chr.X: 19355441-19355441 on Build GRCh38
rs998769543 Chr.X: 19355529-19355529 on Build GRCh38
rs371673691 Chr.X: 19355542-19355542 on Build GRCh38
rs759881225 Chr.X: 19355528-19355528 on Build GRCh38
rs768600287 Chr.X: 19355501-19355501 on Build GRCh38
rs1029792526 Chr.X: 19355563-19355563 on Build GRCh38
rs755815904 Chr.X: 19355535-19355535 on Build GRCh38
rs762911767 Chr.X: 19355535-19355536 on Build GRCh38
rs200051912 Chr.X: 19355527-19355527 on Build GRCh38
rs11278403 Chr.X: 19355530-19355535 on Build GRCh38
rs745607005 Chr.X: 19355405-19355405 on Build GRCh38
rs863224145 Chr.X: 19355452-19355452 on Build GRCh38
rs748317932 Chr.X: 19355557-19355557 on Build GRCh38
rs771923478 Chr.X: 19355547-19355550 on Build GRCh38
rs138237215 Chr.X: 19355438-19355438 on Build GRCh38
rs753346155 Chr.X: 19355545-19355545 on Build GRCh38
rs750307419 Chr.X: 19355453-19355453 on Build GRCh38
rs201759248 Chr.X: 19355544-19355547 on Build GRCh38
rs999399233 Chr.X: 19355574-19355574 on Build GRCh38
rs774400918 Chr.X: 19355531-19355532 on Build GRCh38
rs756722741 Chr.X: 19355551-19355551 on Build GRCh38
rs372505888 Chr.X: 19355442-19355442 on Build GRCh38
rs369604349 Chr.X: 19355522-19355522 on Build GRCh38
rs769963488 Chr.X: 19355462-19355462 on Build GRCh38
rs778744930 Chr.X: 19355402-19355402 on Build GRCh38
rs762975321 Chr.X: 19355510-19355510 on Build GRCh38
rs748024050 Chr.X: 19355544-19355545 on Build GRCh38
rs747312889 Chr.X: 19355522-19355527 on Build GRCh38
rs372707521 Chr.X: 19355544-19355549 on Build GRCh38
rs1054646844 Chr.X: 19355575-19355575 on Build GRCh38
rs2643456 Chr.X: 19355513-19355513 on Build GRCh38
rs757568490 Chr.X: 19355528-19355528 on Build GRCh38

View Full Product Details