Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 124887690-124887961 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ROBO4
Entrez Gene ID
54538
Gene Name
roundabout guidance receptor 4
Gene Aliases
ECSM4, MRB
UniGene
Hs.524121
Gene Chromosome Location
Chr.11: 124884218-124897935 on Build GRCh38
Transcripts
NM_001301088.1 NM_019055.5 XM_006718861.1 XM_011542875.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
415282 c.1981C>T p.R661W Chr.11: 124887808-124887808 on Build GRCh38
924450 c.1970C>T p.S657F Chr.11: 124887819-124887819 on Build GRCh38
3444899 c.1995C>T p.S665S Chr.11: 124887794-124887794 on Build GRCh38
3444898 c.2021G>A p.G674E Chr.11: 124887768-124887768 on Build GRCh38
3444897 c.2051G>A p.S684N Chr.11: 124887738-124887738 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761883031 Chr.11: 124887750-124887750 on Build GRCh38
rs145067866 Chr.11: 124887804-124887804 on Build GRCh38
rs200076421 Chr.11: 124887858-124887858 on Build GRCh38
rs144446942 Chr.11: 124887816-124887816 on Build GRCh38
rs201758034 Chr.11: 124887808-124887808 on Build GRCh38
rs199554824 Chr.11: 124887784-124887784 on Build GRCh38
rs765353464 Chr.11: 124887781-124887804 on Build GRCh38
rs766691280 Chr.11: 124887854-124887854 on Build GRCh38
rs746282574 Chr.11: 124887833-124887833 on Build GRCh38
rs747711430 Chr.11: 124887803-124887803 on Build GRCh38
rs57713193 Chr.11: 124887862-124887862 on Build GRCh38
rs968958062 Chr.11: 124887902-124887902 on Build GRCh38
rs148980564 Chr.11: 124887748-124887748 on Build GRCh38
rs1021855645 Chr.11: 124887898-124887898 on Build GRCh38
rs200777531 Chr.11: 124887831-124887831 on Build GRCh38
rs980207872 Chr.11: 124887905-124887905 on Build GRCh38
rs59771534 Chr.11: 124887919-124887919 on Build GRCh38
rs774201793 Chr.11: 124887723-124887723 on Build GRCh38
rs764038221 Chr.11: 124887732-124887732 on Build GRCh38
rs750782331 Chr.11: 124887779-124887779 on Build GRCh38
rs778513919 Chr.11: 124887891-124887891 on Build GRCh38
rs1057321711 Chr.11: 124887728-124887728 on Build GRCh38
rs117896640 Chr.11: 124887866-124887866 on Build GRCh38
rs1025494270 Chr.11: 124887868-124887868 on Build GRCh38
rs984060525 Chr.11: 124887896-124887896 on Build GRCh38
rs4408324 Chr.11: 124887783-124887783 on Build GRCh38
rs748907249 Chr.11: 124887878-124887878 on Build GRCh38
rs574752013 Chr.11: 124887937-124887937 on Build GRCh38
rs367686562 Chr.11: 124887716-124887716 on Build GRCh38
rs769242452 Chr.11: 124887925-124887925 on Build GRCh38
rs148415408 Chr.11: 124887922-124887922 on Build GRCh38
rs745733396 Chr.11: 124887892-124887892 on Build GRCh38
rs570766237 Chr.11: 124887870-124887870 on Build GRCh38
rs772059095 Chr.11: 124887825-124887825 on Build GRCh38
rs763186591 Chr.11: 124887853-124887853 on Build GRCh38
rs761947550 Chr.11: 124887838-124887838 on Build GRCh38
rs780504528 Chr.11: 124887797-124887797 on Build GRCh38
rs756972175 Chr.11: 124887880-124887880 on Build GRCh38
rs974745841 Chr.11: 124887936-124887936 on Build GRCh38
rs770956089 Chr.11: 124887722-124887722 on Build GRCh38
rs140966335 Chr.11: 124887791-124887791 on Build GRCh38
rs935610422 Chr.11: 124887819-124887819 on Build GRCh38
rs927461466 Chr.11: 124887912-124887912 on Build GRCh38
rs750694224 Chr.11: 124887843-124887843 on Build GRCh38
rs765413851 Chr.11: 124887839-124887839 on Build GRCh38
rs758803778 Chr.11: 124887781-124887781 on Build GRCh38
rs749066541 Chr.11: 124887807-124887807 on Build GRCh38
rs534013556 Chr.11: 124887864-124887864 on Build GRCh38
rs532729044 Chr.11: 124887724-124887724 on Build GRCh38
rs759538505 Chr.11: 124887727-124887727 on Build GRCh38
rs765501810 Chr.11: 124887764-124887764 on Build GRCh38

View Full Product Details