Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 47286236-47286489 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNFRSF21
Entrez Gene ID
27242
Gene Name
TNF receptor superfamily member 21
Gene Aliases
BM-018, CD358, DR6
UniGene
Hs.443577
Gene Chromosome Location
Chr.6: 47231527-47309976 on Build GRCh38
Transcripts
NM_014452.4 XM_017010744.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6805066 c.256C>T p.R86C Chr.6: 47286436-47286436 on Build GRCh38
5463565 c.386G>A p.R129Q Chr.6: 47286306-47286306 on Build GRCh38
5919172 c.415C>T p.Q139* Chr.6: 47286277-47286277 on Build GRCh38
5569226 c.443C>T p.T148M Chr.6: 47286249-47286249 on Build GRCh38
3777721 c.310G>C p.E104Q Chr.6: 47286382-47286382 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757811556 Chr.6: 47286396-47286396 on Build GRCh38
rs549601816 Chr.6: 47286268-47286268 on Build GRCh38
rs150803938 Chr.6: 47286354-47286354 on Build GRCh38
rs768069063 Chr.6: 47286258-47286258 on Build GRCh38
rs374641516 Chr.6: 47286439-47286439 on Build GRCh38
rs944598243 Chr.6: 47286407-47286407 on Build GRCh38
rs139361891 Chr.6: 47286379-47286379 on Build GRCh38
rs769517193 Chr.6: 47286275-47286275 on Build GRCh38
rs756419943 Chr.6: 47286373-47286373 on Build GRCh38
rs915957947 Chr.6: 47286322-47286322 on Build GRCh38
rs767524762 Chr.6: 47286356-47286356 on Build GRCh38
rs766343276 Chr.6: 47286343-47286343 on Build GRCh38
rs746388779 Chr.6: 47286400-47286400 on Build GRCh38
rs754324198 Chr.6: 47286385-47286385 on Build GRCh38
rs148190245 Chr.6: 47286289-47286289 on Build GRCh38
rs752898980 Chr.6: 47286364-47286364 on Build GRCh38
rs371420584 Chr.6: 47286437-47286437 on Build GRCh38
rs762871435 Chr.6: 47286320-47286320 on Build GRCh38
rs141132373 Chr.6: 47286353-47286353 on Build GRCh38
rs947469538 Chr.6: 47286342-47286342 on Build GRCh38
rs142572323 Chr.6: 47286428-47286428 on Build GRCh38
rs375966381 Chr.6: 47286255-47286255 on Build GRCh38
rs764542617 Chr.6: 47286382-47286382 on Build GRCh38
rs772959706 Chr.6: 47286435-47286435 on Build GRCh38
rs200152044 Chr.6: 47286451-47286451 on Build GRCh38
rs754441930 Chr.6: 47286419-47286419 on Build GRCh38
rs376748511 Chr.6: 47286398-47286398 on Build GRCh38
rs142206681 Chr.6: 47286433-47286433 on Build GRCh38
rs1052062404 Chr.6: 47286444-47286444 on Build GRCh38
rs769500818 Chr.6: 47286434-47286434 on Build GRCh38
rs747757642 Chr.6: 47286269-47286269 on Build GRCh38
rs369353250 Chr.6: 47286256-47286256 on Build GRCh38
rs749130136 Chr.6: 47286436-47286436 on Build GRCh38
rs774305126 Chr.6: 47286443-47286443 on Build GRCh38
rs143890976 Chr.6: 47286257-47286257 on Build GRCh38

View Full Product Details