Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 83592416-83592641 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNAP91
Entrez Gene ID
9892
Gene Name
synaptosome associated protein 91
Gene Aliases
AP180, CALM
UniGene
Hs.368046
Gene Chromosome Location
Chr.6: 83552885-83709408 on Build GRCh38
Transcripts
NM_001242792.1 NM_001242793.1 NM_001242794.1 NM_001256717.1 NM_001256718.1 NM_014841.2 XM_005248770.4 XM_006715615.1 XM_011536265.1 XM_011536266.1 XM_011536269.1 XM_011536271.1 XM_011536273.2 XM_011536275.2 XM_011536276.2 XM_011536277.1 XM_017011554.1 XM_017011555.1 XM_017011556.1 XM_017011557.1 XM_017011558.1 XM_017011559.1 XM_017011560.1 XM_017011561.1 XM_017011562.1 XM_017011563.1 XM_017011564.1 XM_017011565.1 XM_017011566.1 XM_017011567.1 XM_017011568.1 XM_017011569.1 XM_017011570.1 XM_017011571.1 XM_017011572.1 XM_017011573.1 XM_017011574.1 XM_017011575.1 XM_017011576.1 XM_017011577.1 XM_017011578.1 XM_017011579.1 XM_017011580.1 XM_017011581.1 XM_017011582.1 XM_017011583.1 XM_017011584.1 XM_017011585.1 XM_017011586.1 XM_017011587.1 XM_017011588.1 XM_017011589.1 XM_017011590.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1446174 c.1926T>G p.F642L Chr.6: 83592459-83592459 on Build GRCh38
395349 c.1930G>T p.D644Y Chr.6: 83592455-83592455 on Build GRCh38
4974785 c.1849G>C p.D617H Chr.6: 83592536-83592536 on Build GRCh38
4569665 c.1924T>G p.F642V Chr.6: 83592461-83592461 on Build GRCh38
384487 c.1871C>A p.P624H Chr.6: 83592514-83592514 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769490273 Chr.6: 83592575-83592575 on Build GRCh38
rs747631007 Chr.6: 83592567-83592567 on Build GRCh38
rs376960586 Chr.6: 83592533-83592533 on Build GRCh38
rs902920889 Chr.6: 83592451-83592451 on Build GRCh38
rs779854303 Chr.6: 83592469-83592469 on Build GRCh38
rs751522665 Chr.6: 83592561-83592561 on Build GRCh38
rs373298932 Chr.6: 83592552-83592552 on Build GRCh38
rs768171500 Chr.6: 83592481-83592481 on Build GRCh38
rs376419762 Chr.6: 83592566-83592566 on Build GRCh38
rs374662830 Chr.6: 83592492-83592492 on Build GRCh38
rs527889745 Chr.6: 83592553-83592553 on Build GRCh38
rs769408827 Chr.6: 83592495-83592495 on Build GRCh38
rs369764947 Chr.6: 83592475-83592475 on Build GRCh38
rs547672251 Chr.6: 83592565-83592565 on Build GRCh38
rs1028561907 Chr.6: 83592538-83592538 on Build GRCh38
rs779843791 Chr.6: 83592557-83592557 on Build GRCh38
rs961764773 Chr.6: 83592599-83592599 on Build GRCh38
rs746898754 Chr.6: 83592474-83592474 on Build GRCh38
rs201065263 Chr.6: 83592508-83592508 on Build GRCh38
rs760060053 Chr.6: 83592512-83592512 on Build GRCh38
rs370310713 Chr.6: 83592537-83592537 on Build GRCh38
rs777283362 Chr.6: 83592586-83592586 on Build GRCh38
rs773671092 Chr.6: 83592496-83592496 on Build GRCh38
rs771532118 Chr.6: 83592499-83592499 on Build GRCh38
rs560752250 Chr.6: 83592541-83592541 on Build GRCh38
rs763210208 Chr.6: 83592498-83592498 on Build GRCh38
rs767727844 Chr.6: 83592521-83592521 on Build GRCh38
rs750179855 Chr.6: 83592544-83592545 on Build GRCh38
rs776072694 Chr.6: 83592488-83592488 on Build GRCh38

View Full Product Details