Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38064931-38065194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PICK1
Entrez Gene ID
9463
Gene Name
protein interacting with PRKCA 1
Gene Aliases
PICK, PRKCABP
UniGene
Hs.180871
Gene Chromosome Location
Chr.22: 38056311-38075704 on Build GRCh38
Transcripts
NM_001039583.1 NM_001039584.1 NM_012407.3 XM_006724379.2 XM_006724380.2 XM_011530517.2 XM_017029090.1 XM_017029091.1 XM_017029092.1 XM_017029093.1 XM_017029094.1 XM_017029095.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1191662 c.173C>A p.A58E Chr.22: 38065021-38065021 on Build GRCh38
6468158 c.274G>C p.E92Q Chr.22: 38065122-38065122 on Build GRCh38
4582398 c.276G>A p.E92E Chr.22: 38065124-38065124 on Build GRCh38
3842672 c.205G>A p.E69K Chr.22: 38065053-38065053 on Build GRCh38
3554324 c.212C>T p.T71I Chr.22: 38065060-38065060 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779941301 Chr.22: 38065148-38065148 on Build GRCh38
rs764110971 Chr.22: 38065108-38065108 on Build GRCh38
rs750223558 Chr.22: 38065037-38065037 on Build GRCh38
rs117707976 Chr.22: 38065031-38065031 on Build GRCh38
rs376461202 Chr.22: 38065061-38065061 on Build GRCh38
rs767477262 Chr.22: 38065139-38065139 on Build GRCh38
rs528844657 Chr.22: 38065019-38065019 on Build GRCh38
rs1034840098 Chr.22: 38065143-38065143 on Build GRCh38
rs374134468 Chr.22: 38065161-38065161 on Build GRCh38
rs148164306 Chr.22: 38064977-38064977 on Build GRCh38
rs868326767 Chr.22: 38065093-38065093 on Build GRCh38
rs371875067 Chr.22: 38064973-38064973 on Build GRCh38
rs185413229 Chr.22: 38065070-38065070 on Build GRCh38
rs771440049 Chr.22: 38065069-38065069 on Build GRCh38
rs748882885 Chr.22: 38065049-38065049 on Build GRCh38
rs903788869 Chr.22: 38065138-38065138 on Build GRCh38
rs751472759 Chr.22: 38065032-38065032 on Build GRCh38
rs770073408 Chr.22: 38065078-38065078 on Build GRCh38
rs775702594 Chr.22: 38065094-38065094 on Build GRCh38
rs753571527 Chr.22: 38065156-38065156 on Build GRCh38
rs759886998 Chr.22: 38065077-38065077 on Build GRCh38
rs759801137 Chr.22: 38064979-38064979 on Build GRCh38
rs767034288 Chr.22: 38065163-38065163 on Build GRCh38
rs373308734 Chr.22: 38064986-38064986 on Build GRCh38
rs768613937 Chr.22: 38065125-38065125 on Build GRCh38
rs755889746 Chr.22: 38065045-38065045 on Build GRCh38
rs779849358 Chr.22: 38065046-38065046 on Build GRCh38
rs765429765 Chr.22: 38064981-38064981 on Build GRCh38
rs908750329 Chr.22: 38065105-38065105 on Build GRCh38
rs757176545 Chr.22: 38065034-38065034 on Build GRCh38
rs751625775 Chr.22: 38065109-38065109 on Build GRCh38
rs373401475 Chr.22: 38065024-38065024 on Build GRCh38
rs767008199 Chr.22: 38065113-38065113 on Build GRCh38
rs201199072 Chr.22: 38065146-38065146 on Build GRCh38
rs1043621768 Chr.22: 38065126-38065126 on Build GRCh38
rs780886922 Chr.22: 38065036-38065036 on Build GRCh38
rs924714914 Chr.22: 38064970-38064970 on Build GRCh38
rs144498214 Chr.22: 38065062-38065062 on Build GRCh38

View Full Product Details