Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 89325498-89325723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SECISBP2
Entrez Gene ID
79048
Gene Name
SECIS binding protein 2
Gene Aliases
SBP2
UniGene
Hs.59804
Gene Chromosome Location
Chr.9: 89318473-89367117 on Build GRCh38
Transcripts
NM_001282688.1 NM_001282689.1 NM_001282690.1 NM_024077.4 XM_005252196.2 XM_006717282.2 XM_011519000.2 XM_011519001.1 XM_011519002.1 XM_011519003.1 XM_017015122.1 XM_017015123.1 XM_017015124.1 XM_017015125.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1110970 c.383G>A p.R128Q Chr.9: 89325627-89325627 on Build GRCh38
1110968 c.278C>G p.S93C Chr.9: 89325522-89325522 on Build GRCh38
6013524 c.401C>T p.P134L Chr.9: 89325645-89325645 on Build GRCh38
1110969 c.382C>T p.R128* Chr.9: 89325626-89325626 on Build GRCh38
7147364 c.390G>C p.E130D Chr.9: 89325634-89325634 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369918420 Chr.9: 89325536-89325536 on Build GRCh38
rs772199609 Chr.9: 89325532-89325532 on Build GRCh38
rs772557373 Chr.9: 89325637-89325637 on Build GRCh38
rs373079221 Chr.9: 89325563-89325563 on Build GRCh38
rs757133015 Chr.9: 89325627-89325627 on Build GRCh38
rs746037464 Chr.9: 89325529-89325529 on Build GRCh38
rs747284759 Chr.9: 89325651-89325651 on Build GRCh38
rs762905999 Chr.9: 89325556-89325556 on Build GRCh38
rs773115168 Chr.9: 89325549-89325549 on Build GRCh38
rs1055993083 Chr.9: 89325665-89325665 on Build GRCh38
rs879108630 Chr.9: 89325594-89325594 on Build GRCh38
rs747514658 Chr.9: 89325589-89325590 on Build GRCh38
rs368643206 Chr.9: 89325640-89325640 on Build GRCh38
rs71510294 Chr.9: 89325673-89325673 on Build GRCh38
rs755662391 Chr.9: 89325601-89325601 on Build GRCh38
rs751321557 Chr.9: 89325568-89325568 on Build GRCh38
rs1036691762 Chr.9: 89325564-89325564 on Build GRCh38
rs754858358 Chr.9: 89325573-89325573 on Build GRCh38
rs949774318 Chr.9: 89325691-89325691 on Build GRCh38
rs376067869 Chr.9: 89325603-89325603 on Build GRCh38
rs767070826 Chr.9: 89325589-89325589 on Build GRCh38
rs757696851 Chr.9: 89325594-89325596 on Build GRCh38
rs752432903 Chr.9: 89325592-89325592 on Build GRCh38
rs922546563 Chr.9: 89325531-89325531 on Build GRCh38
rs766210796 Chr.9: 89325660-89325660 on Build GRCh38
rs760743400 Chr.9: 89325541-89325541 on Build GRCh38
rs757844295 Chr.9: 89325622-89325622 on Build GRCh38
rs776693254 Chr.9: 89325655-89325655 on Build GRCh38
rs768678703 Chr.9: 89325654-89325654 on Build GRCh38
rs62638730 Chr.9: 89325666-89325666 on Build GRCh38
rs779280246 Chr.9: 89325626-89325626 on Build GRCh38
rs777447200 Chr.9: 89325602-89325602 on Build GRCh38
rs372635973 Chr.9: 89325543-89325543 on Build GRCh38

View Full Product Details