Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 53089735-53089994 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC1A7
Entrez Gene ID
6512
Gene Name
solute carrier family 1 member 7
Gene Aliases
AAAT, EAAT5
UniGene
Hs.104637
Gene Chromosome Location
Chr.1: 53087179-53142632 on Build GRCh38
Transcripts
NM_001287595.1 NM_001287597.1 NM_006671.5 NR_109858.1 XM_011542002.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5496024 c.1324G>T p.D442Y Chr.1: 53089837-53089837 on Build GRCh38
1265746 c.1327G>A p.D443N Chr.1: 53089834-53089834 on Build GRCh38
910758 c.1227-1G>T p.? Chr.1: 53089935-53089935 on Build GRCh38
5420776 c.1272G>T p.Q424H Chr.1: 53089889-53089889 on Build GRCh38
4726643 c.1360C>A p.L454M Chr.1: 53089801-53089801 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551059696 Chr.1: 53089838-53089838 on Build GRCh38
rs772893550 Chr.1: 53089917-53089917 on Build GRCh38
rs145443647 Chr.1: 53089852-53089852 on Build GRCh38
rs775929530 Chr.1: 53089944-53089944 on Build GRCh38
rs1003584353 Chr.1: 53089956-53089956 on Build GRCh38
rs530578463 Chr.1: 53089879-53089879 on Build GRCh38
rs772032993 Chr.1: 53089849-53089849 on Build GRCh38
rs986827840 Chr.1: 53089859-53089859 on Build GRCh38
rs763466867 Chr.1: 53089952-53089952 on Build GRCh38
rs376870782 Chr.1: 53089823-53089823 on Build GRCh38
rs750036026 Chr.1: 53089886-53089886 on Build GRCh38
rs774489382 Chr.1: 53089837-53089837 on Build GRCh38
rs143355856 Chr.1: 53089916-53089916 on Build GRCh38
rs767187667 Chr.1: 53089888-53089888 on Build GRCh38
rs766656173 Chr.1: 53089886-53089915 on Build GRCh38
rs141913685 Chr.1: 53089885-53089885 on Build GRCh38
rs950501645 Chr.1: 53089790-53089790 on Build GRCh38
rs768329543 Chr.1: 53089905-53089905 on Build GRCh38
rs747734846 Chr.1: 53089942-53089942 on Build GRCh38
rs1046729073 Chr.1: 53089972-53089972 on Build GRCh38
rs747186673 Chr.1: 53089925-53089925 on Build GRCh38
rs369327397 Chr.1: 53089865-53089865 on Build GRCh38
rs202187580 Chr.1: 53089793-53089793 on Build GRCh38
rs373326915 Chr.1: 53089871-53089871 on Build GRCh38
rs764376577 Chr.1: 53089957-53089957 on Build GRCh38
rs36059472 Chr.1: 53089888-53089888 on Build GRCh38
rs540949444 Chr.1: 53089776-53089776 on Build GRCh38
rs776965478 Chr.1: 53089935-53089935 on Build GRCh38
rs143461330 Chr.1: 53089853-53089853 on Build GRCh38
rs567526262 Chr.1: 53089975-53089975 on Build GRCh38
rs768710932 Chr.1: 53089831-53089831 on Build GRCh38
rs779823364 Chr.1: 53089889-53089889 on Build GRCh38
rs145807376 Chr.1: 53089930-53089939 on Build GRCh38
rs780795904 Chr.1: 53089813-53089813 on Build GRCh38
rs779178370 Chr.1: 53089784-53089784 on Build GRCh38
rs751083102 Chr.1: 53089770-53089770 on Build GRCh38
rs748735189 Chr.1: 53089902-53089902 on Build GRCh38
rs760775686 Chr.1: 53089757-53089757 on Build GRCh38
rs376848498 Chr.1: 53089907-53089907 on Build GRCh38
rs769301056 Chr.1: 53089815-53089815 on Build GRCh38
rs370913970 Chr.1: 53089911-53089911 on Build GRCh38
rs751868043 Chr.1: 53089969-53089969 on Build GRCh38
rs749184336 Chr.1: 53089825-53089825 on Build GRCh38
rs200606407 Chr.1: 53089864-53089864 on Build GRCh38
rs758815960 Chr.1: 53089806-53089806 on Build GRCh38
rs774525042 Chr.1: 53089891-53089891 on Build GRCh38
rs771049177 Chr.1: 53089933-53089933 on Build GRCh38
rs1045990523 Chr.1: 53089951-53089951 on Build GRCh38
rs778135532 Chr.1: 53089882-53089882 on Build GRCh38
rs374989145 Chr.1: 53089814-53089814 on Build GRCh38
rs759584677 Chr.1: 53089939-53089939 on Build GRCh38
rs183624599 Chr.1: 53089782-53089782 on Build GRCh38

View Full Product Details