Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 44905938-44906175 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YWHAB
Entrez Gene ID
7529
Gene Name
tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein beta
Gene Aliases
GW128, HEL-S-1, HS1, KCIP-1, YWHAA
UniGene
Hs.643544
Gene Chromosome Location
Chr.20: 44885599-44908532 on Build GRCh38
Transcripts
NM_003404.4 NM_139323.3 XM_017028039.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3725292 c.622C>T p.L208L Chr.20: 44906034-44906034 on Build GRCh38
1729288 c.649A>T p.T217S Chr.20: 44906061-44906061 on Build GRCh38
6224405 c.620C>T p.T207M Chr.20: 44906032-44906032 on Build GRCh38
1270648 c.684+1_684+2insT p.? Chr.20: 44906097-44906098 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765587390 Chr.20: 44905984-44905984 on Build GRCh38
rs368087121 Chr.20: 44906132-44906132 on Build GRCh38
rs771621179 Chr.20: 44906110-44906110 on Build GRCh38
rs376067295 Chr.20: 44905976-44905976 on Build GRCh38
rs761703648 Chr.20: 44906124-44906124 on Build GRCh38
rs542237358 Chr.20: 44906135-44906135 on Build GRCh38
rs776953403 Chr.20: 44906048-44906048 on Build GRCh38
rs754976856 Chr.20: 44906144-44906144 on Build GRCh38
rs753129464 Chr.20: 44906115-44906115 on Build GRCh38
rs1041213830 Chr.20: 44906113-44906113 on Build GRCh38
rs374058745 Chr.20: 44906032-44906032 on Build GRCh38
rs370845746 Chr.20: 44906117-44906117 on Build GRCh38
rs777490924 Chr.20: 44905978-44905978 on Build GRCh38
rs763907268 Chr.20: 44905972-44905972 on Build GRCh38
rs758417751 Chr.20: 44906018-44906018 on Build GRCh38
rs755545276 Chr.20: 44906051-44906051 on Build GRCh38
rs781631165 Chr.20: 44906062-44906062 on Build GRCh38
rs553553500 Chr.20: 44906103-44906103 on Build GRCh38
rs200157783 Chr.20: 44905979-44905979 on Build GRCh38
rs376330601 Chr.20: 44905969-44905969 on Build GRCh38
rs901378097 Chr.20: 44906133-44906133 on Build GRCh38
rs573702386 Chr.20: 44906116-44906116 on Build GRCh38
rs745592677 Chr.20: 44906107-44906107 on Build GRCh38
rs374163232 Chr.20: 44906141-44906141 on Build GRCh38
rs776670359 Chr.20: 44906121-44906121 on Build GRCh38
rs750604097 Chr.20: 44906001-44906001 on Build GRCh38
rs367989746 Chr.20: 44906034-44906034 on Build GRCh38
rs140330183 Chr.20: 44906042-44906042 on Build GRCh38
rs748546824 Chr.20: 44906064-44906064 on Build GRCh38
rs756407085 Chr.20: 44906099-44906099 on Build GRCh38

View Full Product Details