Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 135120880-135121153 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RAB3GAP1
Entrez Gene ID
22930
Gene Name
RAB3 GTPase activating protein catalytic subunit 1
Gene Aliases
P130, RAB3GAP, RAB3GAP130, WARBM1
UniGene
Hs.306327
Gene Chromosome Location
Chr.2: 135052265-135176667 on Build GRCh38
Transcripts
NM_001172435.1 NM_012233.2 XM_011510823.2 XM_011510825.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5018765 c.726delT p.W244fs*8 Chr.2: 135120896-135120896 on Build GRCh38
5018766 c.594delT p.W200fs*8 Chr.2: 135120896-135120896 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs587776651 Chr.2: 135120919-135120919 on Build GRCh38
rs1032548769 Chr.2: 135120999-135120999 on Build GRCh38
rs758784625 Chr.2: 135120953-135120953 on Build GRCh38
rs775490485 Chr.2: 135120906-135120906 on Build GRCh38
rs541536954 Chr.2: 135120909-135120909 on Build GRCh38
rs542224856 Chr.2: 135120955-135120955 on Build GRCh38
rs1057389006 Chr.2: 135121009-135121009 on Build GRCh38
rs761440730 Chr.2: 135120935-135120935 on Build GRCh38
rs954544020 Chr.2: 135120951-135120951 on Build GRCh38
rs913325012 Chr.2: 135120971-135120971 on Build GRCh38
rs1047634275 Chr.2: 135121098-135121099 on Build GRCh38
rs564567418 Chr.2: 135121114-135121114 on Build GRCh38
rs201480336 Chr.2: 135120936-135120936 on Build GRCh38
rs994345069 Chr.2: 135121096-135121096 on Build GRCh38
rs937597505 Chr.2: 135121001-135121001 on Build GRCh38
rs372760255 Chr.2: 135120907-135120907 on Build GRCh38
rs111925495 Chr.2: 135120996-135120996 on Build GRCh38
rs563409598 Chr.2: 135120960-135120960 on Build GRCh38
rs763677321 Chr.2: 135120908-135120908 on Build GRCh38
rs916349492 Chr.2: 135121119-135121119 on Build GRCh38
rs369923063 Chr.2: 135120970-135120970 on Build GRCh38
rs1043073596 Chr.2: 135120980-135120980 on Build GRCh38
rs947421214 Chr.2: 135120975-135120975 on Build GRCh38
rs368668498 Chr.2: 135120932-135120932 on Build GRCh38
rs752534783 Chr.2: 135120969-135120969 on Build GRCh38
rs187925063 Chr.2: 135121051-135121051 on Build GRCh38
rs779029679 Chr.2: 135120948-135120951 on Build GRCh38
rs998211340 Chr.2: 135120927-135120927 on Build GRCh38
rs771764402 Chr.2: 135120903-135120903 on Build GRCh38
rs990947142 Chr.2: 135121099-135121099 on Build GRCh38

View Full Product Details