Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 43077102-43077345 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RET
Entrez Gene ID
5979
Gene Name
ret proto-oncogene
Gene Aliases
CDHF12, CDHR16, HSCR1, MEN2A, MEN2B, MTC1, PTC, RET-ELE1, RET51
UniGene
Hs.350321
Gene Chromosome Location
Chr.10: 43077069-43130351 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6363523 c.58C>T p.P20S Chr.10: 43077316-43077316 on Build GRCh38
6363522 c.58C>T p.P20S Chr.10: 43077316-43077316 on Build GRCh38
6517801 c.16T>A p.S6T Chr.10: 43077274-43077274 on Build GRCh38
7341797 c.44_46delTGC p.L19delL Chr.10: 43077302-43077304 on Build GRCh38
7341796 c.44_46delTGC p.L19delL Chr.10: 43077302-43077304 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778434166 Chr.10: 43077302-43077304 on Build GRCh38
rs1001790437 Chr.10: 43077164-43077164 on Build GRCh38
rs587780812 Chr.10: 43077289-43077289 on Build GRCh38
rs1008836714 Chr.10: 43077308-43077309 on Build GRCh38
rs1035564388 Chr.10: 43077188-43077188 on Build GRCh38
rs944333660 Chr.10: 43077321-43077321 on Build GRCh38
rs985601017 Chr.10: 43077256-43077256 on Build GRCh38
rs989618487 Chr.10: 43077192-43077192 on Build GRCh38
rs1020734515 Chr.10: 43077205-43077205 on Build GRCh38
rs905665558 Chr.10: 43077152-43077152 on Build GRCh38
rs751005619 Chr.10: 43077222-43077222 on Build GRCh38
rs876657980 Chr.10: 43077257-43077257 on Build GRCh38
rs1011350432 Chr.10: 43077127-43077127 on Build GRCh38
rs794728684 Chr.10: 43077259-43077259 on Build GRCh38
rs886046985 Chr.10: 43077127-43077127 on Build GRCh38
rs1013584871 Chr.10: 43077203-43077203 on Build GRCh38
rs765384640 Chr.10: 43077208-43077208 on Build GRCh38
rs959953942 Chr.10: 43077195-43077195 on Build GRCh38
rs954199293 Chr.10: 43077236-43077236 on Build GRCh38
rs1050242868 Chr.10: 43077288-43077288 on Build GRCh38
rs876660157 Chr.10: 43077303-43077303 on Build GRCh38
rs3128725 Chr.10: 43077191-43077191 on Build GRCh38
rs1015641781 Chr.10: 43077206-43077206 on Build GRCh38
rs915755946 Chr.10: 43077120-43077120 on Build GRCh38
rs878855065 Chr.10: 43077323-43077324 on Build GRCh38
rs768132465 Chr.10: 43077301-43077302 on Build GRCh38
rs974158136 Chr.10: 43077212-43077212 on Build GRCh38
rs1043270606 Chr.10: 43077133-43077133 on Build GRCh38

View Full Product Details