Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 78050121-78050386 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TNRC6C
Entrez Gene ID
57690
Gene Name
trinucleotide repeat containing 6C
Gene Aliases
-
UniGene
Hs.406810
Gene Chromosome Location
Chr.17: 77974984-78108835 on Build GRCh38
Transcripts
NM_001142640.1 NM_018996.3 XM_006721996.3 XM_006721997.3 XM_006722000.3 XM_011525088.2 XM_017024906.1 XM_017024907.1 XM_017024908.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5799940 c.1291C>G p.H431D Chr.17: 78050353-78050353 on Build GRCh38
5343035 c.1138G>T p.E380* Chr.17: 78050200-78050200 on Build GRCh38
5810589 c.1268A>G p.D423G Chr.17: 78050330-78050330 on Build GRCh38
4630348 c.1206T>A p.G402G Chr.17: 78050268-78050268 on Build GRCh38
6805725 c.1259G>A p.R420Q Chr.17: 78050321-78050321 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749461238 Chr.17: 78050235-78050235 on Build GRCh38
rs1034648374 Chr.17: 78050182-78050182 on Build GRCh38
rs376718734 Chr.17: 78050356-78050356 on Build GRCh38
rs73999610 Chr.17: 78050184-78050184 on Build GRCh38
rs756171360 Chr.17: 78050212-78050212 on Build GRCh38
rs202206566 Chr.17: 78050185-78050185 on Build GRCh38
rs1021143036 Chr.17: 78050244-78050244 on Build GRCh38
rs867124430 Chr.17: 78050353-78050353 on Build GRCh38
rs139060581 Chr.17: 78050191-78050191 on Build GRCh38
rs778670893 Chr.17: 78050276-78050276 on Build GRCh38
rs781742093 Chr.17: 78050207-78050207 on Build GRCh38
rs753174079 Chr.17: 78050211-78050211 on Build GRCh38
rs745716049 Chr.17: 78050277-78050277 on Build GRCh38
rs1006128659 Chr.17: 78050241-78050241 on Build GRCh38
rs373843245 Chr.17: 78050347-78050347 on Build GRCh38
rs777905479 Chr.17: 78050337-78050337 on Build GRCh38
rs549081839 Chr.17: 78050305-78050305 on Build GRCh38
rs766732484 Chr.17: 78050311-78050312 on Build GRCh38
rs1002161956 Chr.17: 78050162-78050162 on Build GRCh38
rs371164154 Chr.17: 78050145-78050145 on Build GRCh38
rs897891847 Chr.17: 78050329-78050329 on Build GRCh38
rs755518368 Chr.17: 78050197-78050197 on Build GRCh38
rs746122613 Chr.17: 78050351-78050351 on Build GRCh38
rs760368686 Chr.17: 78050175-78050207 on Build GRCh38
rs753191295 Chr.17: 78050326-78050326 on Build GRCh38
rs777730188 Chr.17: 78050213-78050213 on Build GRCh38
rs979330024 Chr.17: 78050270-78050270 on Build GRCh38
rs560729273 Chr.17: 78050282-78050282 on Build GRCh38
rs760540682 Chr.17: 78050290-78050290 on Build GRCh38
rs556098632 Chr.17: 78050309-78050309 on Build GRCh38
rs932194812 Chr.17: 78050288-78050288 on Build GRCh38
rs774844463 Chr.17: 78050171-78050171 on Build GRCh38
rs537887612 Chr.17: 78050321-78050321 on Build GRCh38
rs763533162 Chr.17: 78050309-78050310 on Build GRCh38
rs759937474 Chr.17: 78050314-78050314 on Build GRCh38
rs1027519598 Chr.17: 78050147-78050147 on Build GRCh38
rs944726329 Chr.17: 78050316-78050316 on Build GRCh38
rs767504746 Chr.17: 78050167-78050167 on Build GRCh38
rs766842690 Chr.17: 78050194-78050194 on Build GRCh38
rs760275877 Chr.17: 78050178-78050178 on Build GRCh38
rs757452785 Chr.17: 78050346-78050346 on Build GRCh38
rs569253210 Chr.17: 78050310-78050310 on Build GRCh38
rs752028839 Chr.17: 78050195-78050195 on Build GRCh38
rs376745906 Chr.17: 78050161-78050161 on Build GRCh38
rs753976934 Chr.17: 78050341-78050341 on Build GRCh38
rs753278448 Chr.17: 78050306-78050307 on Build GRCh38
rs770251104 Chr.17: 78050226-78050226 on Build GRCh38
rs142685898 Chr.17: 78050286-78050286 on Build GRCh38
rs372596080 Chr.17: 78050258-78050258 on Build GRCh38
rs768142013 Chr.17: 78050296-78050296 on Build GRCh38
rs763723143 Chr.17: 78050303-78050304 on Build GRCh38
rs189050450 Chr.17: 78050327-78050327 on Build GRCh38

View Full Product Details