Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 171014272-171014533 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC2A2
Entrez Gene ID
6514
Gene Name
solute carrier family 2 member 2
Gene Aliases
GLUT2
UniGene
Hs.167584
Gene Chromosome Location
Chr.3: 170996341-171026979 on Build GRCh38
Transcripts
NM_000340.1 NM_001278658.1 NM_001278659.1 XM_011513087.2 XM_011513089.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5681313 c.328A>T p.T110S Chr.3: 171014512-171014512 on Build GRCh38
6484879 c.367G>T p.G123* Chr.3: 171014473-171014473 on Build GRCh38
4453857 c.359A>T p.D120V Chr.3: 171014481-171014481 on Build GRCh38
1041157 c.339C>A p.F113L Chr.3: 171014501-171014501 on Build GRCh38
6490227 c.360C>A p.D120E Chr.3: 171014480-171014480 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs144368965 Chr.3: 171014310-171014310 on Build GRCh38
rs766531758 Chr.3: 171014419-171014419 on Build GRCh38
rs377238940 Chr.3: 171014509-171014509 on Build GRCh38
rs779800298 Chr.3: 171014483-171014483 on Build GRCh38
rs768407637 Chr.3: 171014493-171014493 on Build GRCh38
rs895768356 Chr.3: 171014337-171014337 on Build GRCh38
rs189047794 Chr.3: 171014392-171014392 on Build GRCh38
rs574083163 Chr.3: 171014407-171014407 on Build GRCh38
rs763313888 Chr.3: 171014418-171014418 on Build GRCh38
rs761364319 Chr.3: 171014460-171014460 on Build GRCh38
rs5400 Chr.3: 171014511-171014511 on Build GRCh38
rs772002572 Chr.3: 171014478-171014478 on Build GRCh38
rs150539315 Chr.3: 171014349-171014349 on Build GRCh38
rs752870085 Chr.3: 171014450-171014450 on Build GRCh38
rs542904712 Chr.3: 171014370-171014370 on Build GRCh38
rs541443764 Chr.3: 171014465-171014465 on Build GRCh38
rs185252274 Chr.3: 171014463-171014463 on Build GRCh38
rs778974360 Chr.3: 171014474-171014474 on Build GRCh38
rs745588913 Chr.3: 171014477-171014477 on Build GRCh38
rs746632604 Chr.3: 171014484-171014484 on Build GRCh38
rs753980727 Chr.3: 171014489-171014489 on Build GRCh38
rs781101254 Chr.3: 171014446-171014446 on Build GRCh38
rs1040242292 Chr.3: 171014425-171014425 on Build GRCh38
rs56261896 Chr.3: 171014354-171014354 on Build GRCh38
rs901669788 Chr.3: 171014426-171014426 on Build GRCh38
rs1036892474 Chr.3: 171014333-171014333 on Build GRCh38
rs751803113 Chr.3: 171014437-171014437 on Build GRCh38
rs939919352 Chr.3: 171014312-171014312 on Build GRCh38

View Full Product Details