Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 132210101-132210372 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
P4HA2
Entrez Gene ID
8974
Gene Name
prolyl 4-hydroxylase subunit alpha 2
Gene Aliases
-
UniGene
Hs.519568
Gene Chromosome Location
Chr.5: 132190147-132227863 on Build GRCh38
Transcripts
NM_001017973.1 NM_001017974.1 NM_001142598.1 NM_001142599.1 NM_004199.2 XM_005272116.4 XM_005272117.4 XM_005272118.4 XM_005272119.4 XM_005272120.4 XM_006714728.3 XM_006714729.3 XM_006714730.3 XM_011543705.2 XM_017010011.1 XM_017010012.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6101524 c.709G>T p.D237Y Chr.5: 132210284-132210284 on Build GRCh38
3137211 c.674G>A p.R225H Chr.5: 132210319-132210319 on Build GRCh38
6736584 c.673C>T p.R225C Chr.5: 132210320-132210320 on Build GRCh38
6736585 c.673C>T p.R225C Chr.5: 132210320-132210320 on Build GRCh38
3608434 c.630G>T p.L210L Chr.5: 132210363-132210363 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746888081 Chr.5: 132210326-132210326 on Build GRCh38
rs200355999 Chr.5: 132210301-132210301 on Build GRCh38
rs979150749 Chr.5: 132210292-132210292 on Build GRCh38
rs751843221 Chr.5: 132210257-132210257 on Build GRCh38
rs144299745 Chr.5: 132210320-132210320 on Build GRCh38
rs267600343 Chr.5: 132210289-132210289 on Build GRCh38
rs960422119 Chr.5: 132210175-132210175 on Build GRCh38
rs534113524 Chr.5: 132210283-132210283 on Build GRCh38
rs1050505355 Chr.5: 132210173-132210173 on Build GRCh38
rs771957451 Chr.5: 132210319-132210319 on Build GRCh38
rs749593937 Chr.5: 132210309-132210309 on Build GRCh38
rs773151852 Chr.5: 132210264-132210264 on Build GRCh38
rs201056908 Chr.5: 132210327-132210327 on Build GRCh38
rs540078172 Chr.5: 132210331-132210331 on Build GRCh38
rs149593706 Chr.5: 132210302-132210302 on Build GRCh38
rs577387855 Chr.5: 132210311-132210311 on Build GRCh38
rs146783114 Chr.5: 132210179-132210179 on Build GRCh38
rs369753157 Chr.5: 132210345-132210345 on Build GRCh38
rs148740080 Chr.5: 132210339-132210339 on Build GRCh38
rs994478594 Chr.5: 132210317-132210317 on Build GRCh38
rs770629032 Chr.5: 132210337-132210337 on Build GRCh38
rs981033357 Chr.5: 132210347-132210347 on Build GRCh38
rs201223748 Chr.5: 132210298-132210298 on Build GRCh38
rs764575486 Chr.5: 132210237-132210237 on Build GRCh38
rs747868073 Chr.5: 132210316-132210316 on Build GRCh38
rs997642907 Chr.5: 132210172-132210172 on Build GRCh38
rs886600106 Chr.5: 132210167-132210167 on Build GRCh38
rs202121444 Chr.5: 132210299-132210299 on Build GRCh38
rs1012299681 Chr.5: 132210200-132210200 on Build GRCh38
rs142426918 Chr.5: 132210348-132210348 on Build GRCh38
rs139647613 Chr.5: 132210191-132210191 on Build GRCh38
rs565805794 Chr.5: 132210201-132210201 on Build GRCh38
rs767257294 Chr.5: 132210177-132210177 on Build GRCh38

View Full Product Details