Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 18563745-18564016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NT5C1B
Entrez Gene ID
93034
Gene Name
5'-nucleotidase, cytosolic IB
Gene Aliases
AIRP, CN-IB, CN1B
UniGene
Hs.744358
Gene Chromosome Location
Chr.2: 18562871-18589580 on Build GRCh38
Transcripts
NM_001002006.2 NM_001199086.1 NM_001199087.1 NM_001199088.1 NM_033253.3 NM_001199103.1 NM_001199104.1

Gene Symbol
NT5C1B-RDH14
Entrez Gene ID
100526794
Gene Name
NT5C1B-RDH14 readthrough
Gene Aliases
-
UniGene
Hs.744358
Gene Chromosome Location
Chr.2: 18554723-18589580 on Build GRCh38
Transcripts
NM_001002006.2 NM_001199086.1 NM_001199087.1 NM_001199088.1 NM_033253.3 NM_001199103.1 NM_001199104.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7426443 c.1802C>A p.A601D Chr.2: 18563827-18563827 on Build GRCh38
5778622 c.1769T>C p.I590T Chr.2: 18563860-18563860 on Build GRCh38
1013821 c.1681G>A p.E561K Chr.2: 18563948-18563948 on Build GRCh38
291053 c.1622C>T p.A541V Chr.2: 18564007-18564007 on Build GRCh38
6730465 c.1733G>A p.R578Q Chr.2: 18563896-18563896 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs183686876 Chr.2: 18563769-18563769 on Build GRCh38
rs111976608 Chr.2: 18563837-18563837 on Build GRCh38
rs780956283 Chr.2: 18563891-18563891 on Build GRCh38
rs777926079 Chr.2: 18563990-18563990 on Build GRCh38
rs927218090 Chr.2: 18563901-18563901 on Build GRCh38
rs551143562 Chr.2: 18563797-18563797 on Build GRCh38
rs760296227 Chr.2: 18563845-18563845 on Build GRCh38
rs147855687 Chr.2: 18563968-18563968 on Build GRCh38
rs79501117 Chr.2: 18563821-18563821 on Build GRCh38
rs144165826 Chr.2: 18563947-18563947 on Build GRCh38
rs201665017 Chr.2: 18563952-18563952 on Build GRCh38
rs753661424 Chr.2: 18563859-18563859 on Build GRCh38
rs749534769 Chr.2: 18563921-18563921 on Build GRCh38
rs779363806 Chr.2: 18563942-18563942 on Build GRCh38
rs758443500 Chr.2: 18563874-18563874 on Build GRCh38
rs200342649 Chr.2: 18563962-18563962 on Build GRCh38
rs752807785 Chr.2: 18563983-18563983 on Build GRCh38
rs374183150 Chr.2: 18563909-18563909 on Build GRCh38
rs771468715 Chr.2: 18563783-18563783 on Build GRCh38
rs775001075 Chr.2: 18563825-18563825 on Build GRCh38
rs140939592 Chr.2: 18563948-18563948 on Build GRCh38
rs146822895 Chr.2: 18563953-18563953 on Build GRCh38
rs750272718 Chr.2: 18563969-18563969 on Build GRCh38
rs528292689 Chr.2: 18563832-18563832 on Build GRCh38
rs761539349 Chr.2: 18563964-18563964 on Build GRCh38
rs763692331 Chr.2: 18563850-18563850 on Build GRCh38
rs777928885 Chr.2: 18563899-18563899 on Build GRCh38
rs772615000 Chr.2: 18563949-18563949 on Build GRCh38
rs142999822 Chr.2: 18563877-18563877 on Build GRCh38
rs770165137 Chr.2: 18563768-18563768 on Build GRCh38
rs762950541 Chr.2: 18563971-18563971 on Build GRCh38
rs774658746 Chr.2: 18563786-18563786 on Build GRCh38
rs760038236 Chr.2: 18563818-18563818 on Build GRCh38
rs937248379 Chr.2: 18563920-18563920 on Build GRCh38
rs746345419 Chr.2: 18563982-18563983 on Build GRCh38
rs1036003032 Chr.2: 18563806-18563806 on Build GRCh38
rs769673623 Chr.2: 18563994-18563994 on Build GRCh38
rs965387111 Chr.2: 18563993-18563993 on Build GRCh38
rs199969629 Chr.2: 18563773-18563773 on Build GRCh38
rs147939092 Chr.2: 18563860-18563860 on Build GRCh38
rs577374217 Chr.2: 18563989-18563989 on Build GRCh38
rs949756131 Chr.2: 18563829-18563829 on Build GRCh38
rs575659223 Chr.2: 18563972-18563972 on Build GRCh38
rs536648892 Chr.2: 18563897-18563897 on Build GRCh38
rs750261728 Chr.2: 18563863-18563863 on Build GRCh38
rs771130661 Chr.2: 18563936-18563936 on Build GRCh38
rs202025887 Chr.2: 18563889-18563889 on Build GRCh38

View Full Product Details