Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 75220746-75221005 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VDAC2
Entrez Gene ID
7417
Gene Name
voltage dependent anion channel 2
Gene Aliases
POR
UniGene
Hs.355927
Gene Chromosome Location
Chr.10: 75210154-75231448 on Build GRCh38
Transcripts
NM_001184783.2 NM_001184823.1 NM_001324087.1 NM_001324088.1 NM_001324089.1 NM_001324090.1 NM_003375.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6367899 c.481C>A p.L161I Chr.10: 75220867-75220867 on Build GRCh38
1965424 c.405C>T p.N135N Chr.10: 75220791-75220791 on Build GRCh38
920565 c.379_381delTCT p.S128delS Chr.10: 75220765-75220767 on Build GRCh38
7397259 c.530C>T p.A177V Chr.10: 75220871-75220871 on Build GRCh38
920566 c.557C>A p.T186N Chr.10: 75220943-75220943 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201204474 Chr.10: 75220837-75220837 on Build GRCh38
rs766467323 Chr.10: 75220914-75220914 on Build GRCh38
rs200958763 Chr.10: 75220903-75220903 on Build GRCh38
rs755678074 Chr.10: 75220808-75220808 on Build GRCh38
rs532500582 Chr.10: 75220916-75220916 on Build GRCh38
rs748945693 Chr.10: 75220828-75220828 on Build GRCh38
rs60564032 Chr.10: 75220948-75220949 on Build GRCh38
rs764408721 Chr.10: 75220964-75220964 on Build GRCh38
rs900316416 Chr.10: 75220906-75220906 on Build GRCh38
rs929326272 Chr.10: 75220783-75220783 on Build GRCh38
rs746853385 Chr.10: 75220851-75220851 on Build GRCh38
rs34676908 Chr.10: 75220935-75220935 on Build GRCh38
rs753067185 Chr.10: 75220953-75220953 on Build GRCh38
rs761081608 Chr.10: 75220963-75220963 on Build GRCh38
rs562602317 Chr.10: 75220779-75220779 on Build GRCh38
rs772012986 Chr.10: 75220842-75220842 on Build GRCh38
rs768385444 Chr.10: 75220868-75220868 on Build GRCh38
rs547260094 Chr.10: 75220831-75220831 on Build GRCh38
rs767932841 Chr.10: 75220782-75220782 on Build GRCh38
rs569341637 Chr.10: 75220945-75220945 on Build GRCh38
rs199505540 Chr.10: 75220791-75220791 on Build GRCh38
rs773452150 Chr.10: 75220875-75220875 on Build GRCh38
rs775201116 Chr.10: 75220846-75220846 on Build GRCh38
rs777507958 Chr.10: 75220823-75220823 on Build GRCh38
rs879310958 Chr.10: 75220977-75220977 on Build GRCh38
rs753526405 Chr.10: 75220968-75220968 on Build GRCh38
rs778464483 Chr.10: 75220834-75220834 on Build GRCh38
rs142991938 Chr.10: 75220901-75220901 on Build GRCh38

View Full Product Details