Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 153026387-153026648 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RGS17
Entrez Gene ID
26575
Gene Name
regulator of G-protein signaling 17
Gene Aliases
RGS-17, RGSZ2, hRGS17
UniGene
Hs.166313
Gene Chromosome Location
Chr.6: 153010891-153131284 on Build GRCh38
Transcripts
NM_012419.4 XM_017010733.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4586364 c.205G>A p.E69K Chr.6: 153026458-153026458 on Build GRCh38
3859790 c.171C>T p.H57H Chr.6: 153026492-153026492 on Build GRCh38
3859791 c.147G>T p.G49G Chr.6: 153026516-153026516 on Build GRCh38
7234762 c.184G>C p.E62Q Chr.6: 153026479-153026479 on Build GRCh38
1287613 c.162A>G p.R54R Chr.6: 153026501-153026501 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34438717 Chr.6: 153026508-153026508 on Build GRCh38
rs763484564 Chr.6: 153026507-153026507 on Build GRCh38
rs756123514 Chr.6: 153026439-153026439 on Build GRCh38
rs769843767 Chr.6: 153026608-153026608 on Build GRCh38
rs998727853 Chr.6: 153026605-153026605 on Build GRCh38
rs200709570 Chr.6: 153026467-153026467 on Build GRCh38
rs908313056 Chr.6: 153026488-153026488 on Build GRCh38
rs982949114 Chr.6: 153026473-153026473 on Build GRCh38
rs756038289 Chr.6: 153026518-153026519 on Build GRCh38
rs765863505 Chr.6: 153026556-153026557 on Build GRCh38
rs1007716721 Chr.6: 153026596-153026596 on Build GRCh38
rs762558678 Chr.6: 153026499-153026499 on Build GRCh38
rs753686780 Chr.6: 153026563-153026563 on Build GRCh38
rs201455256 Chr.6: 153026489-153026489 on Build GRCh38
rs778845473 Chr.6: 153026575-153026575 on Build GRCh38
rs772484147 Chr.6: 153026485-153026485 on Build GRCh38
rs7755306 Chr.6: 153026436-153026436 on Build GRCh38
rs754866210 Chr.6: 153026545-153026545 on Build GRCh38
rs754970943 Chr.6: 153026463-153026463 on Build GRCh38
rs964994351 Chr.6: 153026410-153026410 on Build GRCh38
rs767403109 Chr.6: 153026413-153026413 on Build GRCh38
rs920765788 Chr.6: 153026418-153026418 on Build GRCh38
rs761548542 Chr.6: 153026512-153026512 on Build GRCh38
rs61759904 Chr.6: 153026520-153026520 on Build GRCh38
rs767208968 Chr.6: 153026514-153026514 on Build GRCh38
rs776956296 Chr.6: 153026494-153026494 on Build GRCh38
rs898107527 Chr.6: 153026549-153026549 on Build GRCh38
rs757161630 Chr.6: 153026409-153026409 on Build GRCh38
rs755922370 Chr.6: 153026554-153026554 on Build GRCh38
rs7768898 Chr.6: 153026617-153026617 on Build GRCh38
rs188950239 Chr.6: 153026416-153026416 on Build GRCh38
rs779235103 Chr.6: 153026466-153026466 on Build GRCh38
rs889299704 Chr.6: 153026579-153026579 on Build GRCh38
rs915569051 Chr.6: 153026518-153026518 on Build GRCh38
rs766369431 Chr.6: 153026559-153026559 on Build GRCh38
rs199591913 Chr.6: 153026458-153026458 on Build GRCh38
rs750254403 Chr.6: 153026537-153026537 on Build GRCh38
rs370883735 Chr.6: 153026570-153026570 on Build GRCh38
rs771443022 Chr.6: 153026493-153026493 on Build GRCh38
rs950891679 Chr.6: 153026461-153026461 on Build GRCh38
rs200509433 Chr.6: 153026495-153026495 on Build GRCh38
rs915747562 Chr.6: 153026443-153026443 on Build GRCh38
rs865917676 Chr.6: 153026527-153026527 on Build GRCh38
rs193116742 Chr.6: 153026491-153026491 on Build GRCh38

View Full Product Details