Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119070185-119070458 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS11
Entrez Gene ID
55823
Gene Name
VPS11, CORVET/HOPS core subunit
Gene Aliases
END1, HLD12, PEP5, RNF108, hVPS11
UniGene
Hs.234282
Gene Chromosome Location
Chr.11: 119067752-119081978 on Build GRCh38
Transcripts
NM_001290185.1 NM_021729.5 XM_011542898.1 XM_011542899.2

SNP Detail

SNP ID Chromosome Location
rs902732564 Chr.11: 119070400-119070400 on Build GRCh38
rs782788829 Chr.11: 119070345-119070345 on Build GRCh38
rs782429510 Chr.11: 119070392-119070392 on Build GRCh38
rs782630479 Chr.11: 119070422-119070422 on Build GRCh38
rs782470900 Chr.11: 119070432-119070432 on Build GRCh38
rs983964814 Chr.11: 119070401-119070401 on Build GRCh38
rs782067451 Chr.11: 119070335-119070335 on Build GRCh38
rs782384435 Chr.11: 119070277-119070277 on Build GRCh38
rs782185125 Chr.11: 119070433-119070433 on Build GRCh38
rs376001982 Chr.11: 119070211-119070211 on Build GRCh38
rs782223607 Chr.11: 119070294-119070294 on Build GRCh38
rs946988820 Chr.11: 119070361-119070361 on Build GRCh38
rs370582110 Chr.11: 119070219-119070219 on Build GRCh38
rs375062653 Chr.11: 119070350-119070350 on Build GRCh38
rs782009208 Chr.11: 119070278-119070278 on Build GRCh38
rs921909177 Chr.11: 119070354-119070354 on Build GRCh38
rs191133267 Chr.11: 119070257-119070257 on Build GRCh38
rs377164897 Chr.11: 119070284-119070284 on Build GRCh38
rs986999947 Chr.11: 119070318-119070318 on Build GRCh38
rs781881442 Chr.11: 119070403-119070403 on Build GRCh38
rs372421136 Chr.11: 119070271-119070271 on Build GRCh38
rs782617829 Chr.11: 119070241-119070241 on Build GRCh38
rs368583640 Chr.11: 119070321-119070321 on Build GRCh38
rs782558144 Chr.11: 119070420-119070420 on Build GRCh38
rs782670953 Chr.11: 119070255-119070255 on Build GRCh38
rs950788333 Chr.11: 119070359-119070359 on Build GRCh38
rs782730868 Chr.11: 119070370-119070370 on Build GRCh38
rs201606629 Chr.11: 119070347-119070347 on Build GRCh38
rs782314006 Chr.11: 119070251-119070251 on Build GRCh38
rs570151697 Chr.11: 119070349-119070349 on Build GRCh38
rs782710324 Chr.11: 119070394-119070394 on Build GRCh38
rs1784302 Chr.11: 119070247-119070247 on Build GRCh38
rs781921640 Chr.11: 119070325-119070325 on Build GRCh38
rs951254157 Chr.11: 119070316-119070316 on Build GRCh38
rs782262968 Chr.11: 119070427-119070427 on Build GRCh38
rs369389837 Chr.11: 119070391-119070391 on Build GRCh38
rs1043973254 Chr.11: 119070362-119070362 on Build GRCh38

View Full Product Details