Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 168016926-168017197 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STK39
Entrez Gene ID
27347
Gene Name
serine/threonine kinase 39
Gene Aliases
DCHT, PASK, SPAK
UniGene
Hs.276271
Gene Chromosome Location
Chr.2: 167954020-168247595 on Build GRCh38
Transcripts
NM_013233.2 XM_005246465.2 XM_017003813.1 XM_017003815.1 XM_017003817.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4526624 c.1406G>A p.R469Q Chr.2: 168017066-168017066 on Build GRCh38
1729931 c.1410T>G p.F470L Chr.2: 168017062-168017062 on Build GRCh38
4465417 c.1382C>T p.S461F Chr.2: 168017090-168017090 on Build GRCh38
3670298 c.1405C>T p.R469* Chr.2: 168017067-168017067 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs202003302 Chr.2: 168017059-168017059 on Build GRCh38
rs200294746 Chr.2: 168017061-168017061 on Build GRCh38
rs575197522 Chr.2: 168017130-168017130 on Build GRCh38
rs752765964 Chr.2: 168017065-168017065 on Build GRCh38
rs746528388 Chr.2: 168017126-168017126 on Build GRCh38
rs756880314 Chr.2: 168017169-168017169 on Build GRCh38
rs976658069 Chr.2: 168017004-168017004 on Build GRCh38
rs754871608 Chr.2: 168017021-168017022 on Build GRCh38
rs141341591 Chr.2: 168017105-168017105 on Build GRCh38
rs201061121 Chr.2: 168017132-168017132 on Build GRCh38
rs779379835 Chr.2: 168017108-168017108 on Build GRCh38
rs746652291 Chr.2: 168017019-168017019 on Build GRCh38
rs750967080 Chr.2: 168017116-168017116 on Build GRCh38
rs780879251 Chr.2: 168017138-168017138 on Build GRCh38
rs754201506 Chr.2: 168017093-168017093 on Build GRCh38
rs895477045 Chr.2: 168017117-168017117 on Build GRCh38
rs949796558 Chr.2: 168017148-168017148 on Build GRCh38
rs532650326 Chr.2: 168017147-168017147 on Build GRCh38
rs187368587 Chr.2: 168017010-168017010 on Build GRCh38
rs779636117 Chr.2: 168017014-168017014 on Build GRCh38
rs966447448 Chr.2: 168016999-168016999 on Build GRCh38
rs753362711 Chr.2: 168017067-168017067 on Build GRCh38
rs973267534 Chr.2: 168017080-168017080 on Build GRCh38
rs776359375 Chr.2: 168017038-168017038 on Build GRCh38
rs376931187 Chr.2: 168016991-168016991 on Build GRCh38
rs567132642 Chr.2: 168016977-168016977 on Build GRCh38
rs201574179 Chr.2: 168017029-168017029 on Build GRCh38
rs199758345 Chr.2: 168017124-168017124 on Build GRCh38
rs778908388 Chr.2: 168017005-168017006 on Build GRCh38
rs769776880 Chr.2: 168017049-168017049 on Build GRCh38
rs764344658 Chr.2: 168017084-168017084 on Build GRCh38
rs760864948 Chr.2: 168017075-168017075 on Build GRCh38
rs773159718 Chr.2: 168017052-168017052 on Build GRCh38
rs372693786 Chr.2: 168016976-168016976 on Build GRCh38
rs867233454 Chr.2: 168017128-168017128 on Build GRCh38
rs989321066 Chr.2: 168016960-168016960 on Build GRCh38
rs184094873 Chr.2: 168016975-168016975 on Build GRCh38
rs746327774 Chr.2: 168017002-168017002 on Build GRCh38
rs747710392 Chr.2: 168017141-168017141 on Build GRCh38
rs761639622 Chr.2: 168017046-168017046 on Build GRCh38
rs746974824 Chr.2: 168017018-168017020 on Build GRCh38
rs776745758 Chr.2: 168016987-168016987 on Build GRCh38
rs200275983 Chr.2: 168017125-168017125 on Build GRCh38

View Full Product Details