Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18123874-18124131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAST3
Entrez Gene ID
23031
Gene Name
microtubule associated serine/threonine kinase 3
Gene Aliases
-
UniGene
Hs.466184
Gene Chromosome Location
Chr.19: 18097764-18151692 on Build GRCh38
Transcripts
NM_015016.1 XM_005259823.3 XM_005259824.2 XM_005259825.2 XM_005259826.3 XM_005259827.2 XM_006722698.2 XM_006722699.2 XM_006722700.3 XM_011527823.1 XM_017026506.1 XM_017026507.1 XM_017026508.1 XM_017026509.1 XM_017026510.1 XM_017026511.1 XM_017026512.1 XM_017026513.1 XM_017026514.1 XM_017026515.1 XM_017026516.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4279691 c.569G>A p.R190H Chr.19: 18123961-18123961 on Build GRCh38
6348501 c.679G>T p.A227S Chr.19: 18124071-18124071 on Build GRCh38
4279692 c.597G>A p.A199A Chr.19: 18123989-18123989 on Build GRCh38
4075575 c.614C>T p.A205V Chr.19: 18124006-18124006 on Build GRCh38
5626738 c.732G>C p.E244D Chr.19: 18124124-18124124 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764844560 Chr.19: 18124092-18124092 on Build GRCh38
rs777024234 Chr.19: 18123923-18123923 on Build GRCh38
rs751970201 Chr.19: 18123999-18123999 on Build GRCh38
rs770435172 Chr.19: 18124082-18124082 on Build GRCh38
rs35945810 Chr.19: 18124000-18124000 on Build GRCh38
rs374782434 Chr.19: 18123902-18123902 on Build GRCh38
rs758937239 Chr.19: 18123917-18123917 on Build GRCh38
rs370057215 Chr.19: 18123971-18123971 on Build GRCh38
rs753506687 Chr.19: 18123989-18123989 on Build GRCh38
rs770128384 Chr.19: 18124019-18124019 on Build GRCh38
rs377366405 Chr.19: 18124102-18124102 on Build GRCh38
rs902583854 Chr.19: 18123969-18123969 on Build GRCh38
rs772638403 Chr.19: 18123980-18123980 on Build GRCh38
rs753039272 Chr.19: 18124097-18124097 on Build GRCh38
rs369492608 Chr.19: 18124073-18124073 on Build GRCh38
rs200608721 Chr.19: 18124047-18124047 on Build GRCh38
rs773227016 Chr.19: 18124078-18124078 on Build GRCh38
rs530169140 Chr.19: 18124049-18124049 on Build GRCh38
rs778308319 Chr.19: 18123996-18123996 on Build GRCh38
rs763402548 Chr.19: 18124101-18124101 on Build GRCh38
rs189142718 Chr.19: 18124109-18124109 on Build GRCh38
rs774221250 Chr.19: 18123961-18123961 on Build GRCh38
rs765659545 Chr.19: 18123993-18123993 on Build GRCh38
rs768584320 Chr.19: 18123960-18123960 on Build GRCh38
rs556227078 Chr.19: 18123921-18123921 on Build GRCh38
rs746038425 Chr.19: 18123931-18123931 on Build GRCh38
rs760263860 Chr.19: 18123986-18123986 on Build GRCh38
rs929360580 Chr.19: 18123934-18123934 on Build GRCh38
rs935511189 Chr.19: 18123978-18123978 on Build GRCh38
rs781362233 Chr.19: 18124007-18124007 on Build GRCh38
rs755362555 Chr.19: 18123901-18123901 on Build GRCh38
rs56022120 Chr.19: 18123979-18123979 on Build GRCh38
rs760475536 Chr.19: 18124080-18124080 on Build GRCh38
rs372528449 Chr.19: 18124083-18124083 on Build GRCh38
rs369578021 Chr.19: 18123995-18123995 on Build GRCh38
rs375539745 Chr.19: 18123992-18123992 on Build GRCh38
rs367784879 Chr.19: 18123918-18123918 on Build GRCh38
rs965022644 Chr.19: 18124095-18124095 on Build GRCh38
rs771094128 Chr.19: 18123922-18123922 on Build GRCh38
rs201536465 Chr.19: 18124040-18124040 on Build GRCh38
rs542708516 Chr.19: 18123904-18123904 on Build GRCh38
rs765831120 Chr.19: 18123987-18123987 on Build GRCh38

View Full Product Details