Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 40779319-40779580 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KRT27
Entrez Gene ID
342574
Gene Name
keratin 27
Gene Aliases
K25IRS3, KRT25C
UniGene
Hs.59363
Gene Chromosome Location
Chr.17: 40776808-40782534 on Build GRCh38
Transcripts
NM_181537.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4696803 c.971C>T p.T324M Chr.17: 40779503-40779503 on Build GRCh38
4999935 c.914T>C p.I305T Chr.17: 40779560-40779560 on Build GRCh38
1640683 c.972+7G>A p.? Chr.17: 40779495-40779495 on Build GRCh38
6697946 c.959C>T p.S320F Chr.17: 40779515-40779515 on Build GRCh38
1285828 c.952C>A p.L318I Chr.17: 40779522-40779522 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs958384287 Chr.17: 40779453-40779453 on Build GRCh38
rs116871734 Chr.17: 40779498-40779498 on Build GRCh38
rs567124584 Chr.17: 40779448-40779448 on Build GRCh38
rs1048781719 Chr.17: 40779342-40779342 on Build GRCh38
rs116274728 Chr.17: 40779528-40779528 on Build GRCh38
rs556081117 Chr.17: 40779499-40779499 on Build GRCh38
rs75024250 Chr.17: 40779543-40779543 on Build GRCh38
rs769572227 Chr.17: 40779460-40779460 on Build GRCh38
rs1015212801 Chr.17: 40779384-40779384 on Build GRCh38
rs1033992757 Chr.17: 40779452-40779453 on Build GRCh38
rs764407338 Chr.17: 40779525-40779525 on Build GRCh38
rs766505066 Chr.17: 40779503-40779503 on Build GRCh38
rs758727717 Chr.17: 40779521-40779521 on Build GRCh38
rs981684 Chr.17: 40779560-40779560 on Build GRCh38
rs757848490 Chr.17: 40779539-40779539 on Build GRCh38
rs745456516 Chr.17: 40779459-40779459 on Build GRCh38
rs775312685 Chr.17: 40779470-40779470 on Build GRCh38
rs757184489 Chr.17: 40779452-40779452 on Build GRCh38
rs768618277 Chr.17: 40779491-40779491 on Build GRCh38
rs753929201 Chr.17: 40779504-40779504 on Build GRCh38
rs968156656 Chr.17: 40779533-40779533 on Build GRCh38
rs558722873 Chr.17: 40779511-40779511 on Build GRCh38
rs536361547 Chr.17: 40779352-40779352 on Build GRCh38
rs752062444 Chr.17: 40779530-40779530 on Build GRCh38
rs756461195 Chr.17: 40779519-40779521 on Build GRCh38
rs1003066252 Chr.17: 40779430-40779430 on Build GRCh38
rs114010637 Chr.17: 40779514-40779514 on Build GRCh38
rs959655153 Chr.17: 40779508-40779508 on Build GRCh38
rs555100425 Chr.17: 40779386-40779386 on Build GRCh38
rs915313313 Chr.17: 40779527-40779527 on Build GRCh38
rs762681877 Chr.17: 40779490-40779490 on Build GRCh38
rs887557319 Chr.17: 40779391-40779391 on Build GRCh38
rs368451287 Chr.17: 40779483-40779483 on Build GRCh38
rs375409398 Chr.17: 40779510-40779510 on Build GRCh38
rs752695919 Chr.17: 40779512-40779513 on Build GRCh38
rs961612623 Chr.17: 40779487-40779487 on Build GRCh38
rs990993482 Chr.17: 40779453-40779453 on Build GRCh38
rs745401113 Chr.17: 40779548-40779548 on Build GRCh38

View Full Product Details