Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10110306-10110531 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPAN
Entrez Gene ID
56342
Gene Name
peter pan homolog (Drosophila)
Gene Aliases
BXDC3, SSF, SSF-1, SSF1, SSF2
UniGene
Hs.14468
Gene Chromosome Location
Chr.19: 10106223-10111299 on Build GRCh38
Transcripts
NM_020230.5 NM_001040664.2 NM_001198690.1

Gene Symbol
PPAN-P2RY11
Entrez Gene ID
692312
Gene Name
PPAN-P2RY11 readthrough
Gene Aliases
-
UniGene
Hs.14468
Gene Chromosome Location
Chr.19: 10106223-10115389 on Build GRCh38
Transcripts
NM_020230.5 NM_001040664.2 NM_001198690.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
417782 c.885G>A p.V295V Chr.19: 10110386-10110386 on Build GRCh38
5079950 c.901+9G>A p.? Chr.19: 10110411-10110411 on Build GRCh38
4716794 c.867C>T p.G289G Chr.19: 10110368-10110368 on Build GRCh38
1262796 c.879C>A p.G293G Chr.19: 10110380-10110380 on Build GRCh38
3932858 c.838A>G p.T280A Chr.19: 10110339-10110339 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757962826 Chr.19: 10110396-10110396 on Build GRCh38
rs771091822 Chr.19: 10110418-10110418 on Build GRCh38
rs376891727 Chr.19: 10110371-10110371 on Build GRCh38
rs368442537 Chr.19: 10110419-10110419 on Build GRCh38
rs374656690 Chr.19: 10110417-10110417 on Build GRCh38
rs773649374 Chr.19: 10110488-10110488 on Build GRCh38
rs936201623 Chr.19: 10110376-10110376 on Build GRCh38
rs752665586 Chr.19: 10110512-10110512 on Build GRCh38
rs200765108 Chr.19: 10110468-10110468 on Build GRCh38
rs74668408 Chr.19: 10110410-10110410 on Build GRCh38
rs746852941 Chr.19: 10110347-10110347 on Build GRCh38
rs201477429 Chr.19: 10110457-10110457 on Build GRCh38
rs941617465 Chr.19: 10110489-10110489 on Build GRCh38
rs768050754 Chr.19: 10110366-10110366 on Build GRCh38
rs756854948 Chr.19: 10110336-10110336 on Build GRCh38
rs369547483 Chr.19: 10110469-10110469 on Build GRCh38
rs138578033 Chr.19: 10110342-10110342 on Build GRCh38
rs768741739 Chr.19: 10110425-10110425 on Build GRCh38
rs761158899 Chr.19: 10110368-10110368 on Build GRCh38
rs2305792 Chr.19: 10110475-10110475 on Build GRCh38
rs776043912 Chr.19: 10110423-10110423 on Build GRCh38
rs762708825 Chr.19: 10110494-10110494 on Build GRCh38
rs756437869 Chr.19: 10110329-10110329 on Build GRCh38
rs368424664 Chr.19: 10110454-10110454 on Build GRCh38
rs754894405 Chr.19: 10110444-10110444 on Build GRCh38
rs143785583 Chr.19: 10110339-10110339 on Build GRCh38
rs760050931 Chr.19: 10110326-10110326 on Build GRCh38
rs759574387 Chr.19: 10110355-10110355 on Build GRCh38
rs1024150387 Chr.19: 10110364-10110364 on Build GRCh38
rs909077048 Chr.19: 10110373-10110373 on Build GRCh38
rs749996645 Chr.19: 10110384-10110384 on Build GRCh38
rs777175945 Chr.19: 10110471-10110471 on Build GRCh38
rs758052599 Chr.19: 10110437-10110437 on Build GRCh38
rs1015575652 Chr.19: 10110337-10110337 on Build GRCh38
rs937920697 Chr.19: 10110377-10110377 on Build GRCh38
rs767401932 Chr.19: 10110499-10110499 on Build GRCh38
rs563999387 Chr.19: 10110426-10110426 on Build GRCh38
rs769072397 Chr.19: 10110470-10110470 on Build GRCh38
rs202217610 Chr.19: 10110416-10110416 on Build GRCh38
rs775952135 Chr.19: 10110367-10110367 on Build GRCh38
rs764932589 Chr.19: 10110383-10110383 on Build GRCh38
rs751023514 Chr.19: 10110443-10110443 on Build GRCh38
rs372740190 Chr.19: 10110456-10110456 on Build GRCh38
rs557947734 Chr.19: 10110327-10110327 on Build GRCh38
rs966033811 Chr.19: 10110338-10110338 on Build GRCh38
rs765996535 Chr.19: 10110438-10110438 on Build GRCh38
rs149310564 Chr.19: 10110344-10110344 on Build GRCh38
rs371789394 Chr.19: 10110495-10110495 on Build GRCh38
rs759259417 Chr.19: 10110411-10110411 on Build GRCh38
rs772125522 Chr.19: 10110420-10110420 on Build GRCh38
rs370405248 Chr.19: 10110372-10110372 on Build GRCh38
rs555647674 Chr.19: 10110369-10110369 on Build GRCh38
rs753419269 Chr.19: 10110334-10110334 on Build GRCh38
rs751063733 Chr.19: 10110496-10110496 on Build GRCh38
rs147502879 Chr.19: 10110425-10110426 on Build GRCh38
rs147270714 Chr.19: 10110354-10110354 on Build GRCh38
rs777530475 Chr.19: 10110331-10110331 on Build GRCh38
rs545914485 Chr.19: 10110424-10110424 on Build GRCh38
rs746943100 Chr.19: 10110411-10110411 on Build GRCh38
rs759057734 Chr.19: 10110497-10110497 on Build GRCh38
rs780202798 Chr.19: 10110401-10110401 on Build GRCh38
rs372347412 Chr.19: 10110434-10110434 on Build GRCh38
rs755879756 Chr.19: 10110455-10110455 on Build GRCh38
rs781145032 Chr.19: 10110453-10110453 on Build GRCh38
rs762144987 Chr.19: 10110472-10110472 on Build GRCh38
rs771127342 Chr.19: 10110352-10110352 on Build GRCh38
rs762194912 Chr.19: 10110430-10110430 on Build GRCh38
rs182090136 Chr.19: 10110435-10110435 on Build GRCh38
rs768326598 Chr.19: 10110348-10110348 on Build GRCh38

View Full Product Details