Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 21333894-21334147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL9
Entrez Gene ID
55958
Gene Name
kelch like family member 9
Gene Aliases
-
UniGene
Hs.701778
Gene Chromosome Location
Chr.9: 21331019-21335430 on Build GRCh38
Transcripts
NM_018847.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6696913 c.872C>T p.T291I Chr.9: 21333988-21333988 on Build GRCh38
753766 c.865G>A p.D289N Chr.9: 21333995-21333995 on Build GRCh38
5888050 c.748C>T p.L250F Chr.9: 21334112-21334112 on Build GRCh38
3906599 c.965A>G p.E322G Chr.9: 21333895-21333895 on Build GRCh38
1636137 c.727C>T p.P243S Chr.9: 21334133-21334133 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761702498 Chr.9: 21334037-21334037 on Build GRCh38
rs750655571 Chr.9: 21333990-21333993 on Build GRCh38
rs752818282 Chr.9: 21334018-21334018 on Build GRCh38
rs780333498 Chr.9: 21334080-21334080 on Build GRCh38
rs757416726 Chr.9: 21333966-21333966 on Build GRCh38
rs201886256 Chr.9: 21333956-21333956 on Build GRCh38
rs764487124 Chr.9: 21334062-21334062 on Build GRCh38
rs149732449 Chr.9: 21334075-21334075 on Build GRCh38
rs778634335 Chr.9: 21334092-21334092 on Build GRCh38
rs781372474 Chr.9: 21333968-21333968 on Build GRCh38
rs771596341 Chr.9: 21334039-21334039 on Build GRCh38
rs779777080 Chr.9: 21334008-21334008 on Build GRCh38
rs752135096 Chr.9: 21334066-21334066 on Build GRCh38
rs770025342 Chr.9: 21333999-21333999 on Build GRCh38
rs553467751 Chr.9: 21334072-21334072 on Build GRCh38
rs772666827 Chr.9: 21334115-21334115 on Build GRCh38
rs1011272745 Chr.9: 21333975-21333975 on Build GRCh38
rs766052132 Chr.9: 21334046-21334046 on Build GRCh38
rs781460600 Chr.9: 21334071-21334071 on Build GRCh38
rs377006324 Chr.9: 21333926-21333926 on Build GRCh38
rs753063600 Chr.9: 21334047-21334047 on Build GRCh38
rs369605312 Chr.9: 21333927-21333927 on Build GRCh38
rs768466113 Chr.9: 21334091-21334091 on Build GRCh38
rs767797835 Chr.9: 21333933-21333933 on Build GRCh38
rs774272026 Chr.9: 21334024-21334024 on Build GRCh38
rs915008622 Chr.9: 21333991-21333991 on Build GRCh38
rs763390095 Chr.9: 21334061-21334061 on Build GRCh38
rs757815699 Chr.9: 21334068-21334068 on Build GRCh38
rs772759819 Chr.9: 21334041-21334041 on Build GRCh38
rs989613432 Chr.9: 21333974-21333974 on Build GRCh38
rs755761257 Chr.9: 21334016-21334016 on Build GRCh38
rs749400388 Chr.9: 21334089-21334089 on Build GRCh38
rs748135862 Chr.9: 21334105-21334105 on Build GRCh38
rs779534972 Chr.9: 21334022-21334022 on Build GRCh38
rs746125213 Chr.9: 21333973-21333973 on Build GRCh38
rs200705897 Chr.9: 21333979-21333979 on Build GRCh38
rs907732776 Chr.9: 21334026-21334026 on Build GRCh38
rs533792568 Chr.9: 21333965-21333965 on Build GRCh38
rs200991754 Chr.9: 21333962-21333962 on Build GRCh38
rs766950905 Chr.9: 21333925-21333925 on Build GRCh38
rs768378191 Chr.9: 21334020-21334020 on Build GRCh38
rs771968486 Chr.9: 21334114-21334114 on Build GRCh38

View Full Product Details