Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 153671310-153671571 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PNCK
Entrez Gene ID
139728
Gene Name
pregnancy up-regulated nonubiquitous CaM kinase
Gene Aliases
BSTK3, CaMK1b
UniGene
Hs.436667
Gene Chromosome Location
Chr.X: 153669723-153687568 on Build GRCh38
Transcripts
NM_001039582.3 NM_001135740.1 XM_005274651.2 XM_006724809.3 XM_011531107.2 XM_011531108.2 XM_011531110.2 XM_011531111.2 XM_011531112.2 XM_017029277.1 XM_017029278.1 XM_017029279.1 XM_017029280.1

SNP Detail

SNP ID Chromosome Location
rs782663920 Chr.X: 153671431-153671431 on Build GRCh38
rs782442885 Chr.X: 153671400-153671400 on Build GRCh38
rs376399747 Chr.X: 153671413-153671413 on Build GRCh38
rs782689278 Chr.X: 153671513-153671513 on Build GRCh38
rs782365402 Chr.X: 153671452-153671452 on Build GRCh38
rs781797331 Chr.X: 153671501-153671501 on Build GRCh38
rs782791462 Chr.X: 153671379-153671379 on Build GRCh38
rs782327258 Chr.X: 153671353-153671353 on Build GRCh38
rs782177334 Chr.X: 153671428-153671428 on Build GRCh38
rs782201125 Chr.X: 153671352-153671352 on Build GRCh38
rs373717346 Chr.X: 153671432-153671432 on Build GRCh38
rs782702567 Chr.X: 153671418-153671418 on Build GRCh38
rs373526914 Chr.X: 153671384-153671384 on Build GRCh38
rs376511007 Chr.X: 153671455-153671455 on Build GRCh38
rs782096324 Chr.X: 153671492-153671492 on Build GRCh38
rs782365846 Chr.X: 153671343-153671343 on Build GRCh38
rs150107065 Chr.X: 153671377-153671377 on Build GRCh38
rs782475350 Chr.X: 153671419-153671419 on Build GRCh38
rs984931602 Chr.X: 153671457-153671457 on Build GRCh38
rs782251902 Chr.X: 153671440-153671440 on Build GRCh38
rs368618313 Chr.X: 153671535-153671535 on Build GRCh38
rs781884120 Chr.X: 153671517-153671517 on Build GRCh38
rs782518787 Chr.X: 153671532-153671532 on Build GRCh38
rs373918058 Chr.X: 153671430-153671430 on Build GRCh38
rs782189775 Chr.X: 153671331-153671331 on Build GRCh38
rs781963016 Chr.X: 153671344-153671344 on Build GRCh38
rs781895157 Chr.X: 153671402-153671402 on Build GRCh38
rs147403907 Chr.X: 153671412-153671412 on Build GRCh38
rs201274962 Chr.X: 153671500-153671500 on Build GRCh38
rs781920254 Chr.X: 153671368-153671368 on Build GRCh38
rs933755630 Chr.X: 153671533-153671533 on Build GRCh38
rs149305071 Chr.X: 153671401-153671401 on Build GRCh38
rs370636003 Chr.X: 153671464-153671464 on Build GRCh38
rs370262918 Chr.X: 153671424-153671424 on Build GRCh38
rs141163387 Chr.X: 153671393-153671393 on Build GRCh38
rs781955442 Chr.X: 153671453-153671453 on Build GRCh38
rs782430839 Chr.X: 153671506-153671506 on Build GRCh38
rs182261010 Chr.X: 153671383-153671383 on Build GRCh38
rs781854027 Chr.X: 153671487-153671487 on Build GRCh38
rs369961126 Chr.X: 153671335-153671335 on Build GRCh38
rs564994030 Chr.X: 153671539-153671539 on Build GRCh38
rs377279617 Chr.X: 153671332-153671332 on Build GRCh38
rs782526274 Chr.X: 153671403-153671403 on Build GRCh38
rs370760495 Chr.X: 153671540-153671540 on Build GRCh38
rs782713797 Chr.X: 153671496-153671496 on Build GRCh38
rs782786170 Chr.X: 153671481-153671481 on Build GRCh38

View Full Product Details