Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41264419-41264644 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HNRNPUL1
Entrez Gene ID
11100
Gene Name
heterogeneous nuclear ribonucleoprotein U like 1
Gene Aliases
E1B-AP5, E1BAP5, HNRPUL1
UniGene
Hs.155218
Gene Chromosome Location
Chr.19: 41262476-41307783 on Build GRCh38
Transcripts
NM_001321208.1 NM_007040.5 NM_144732.4 XM_005258459.3 XM_005258463.3 XM_005258464.3 XM_011526392.2 XM_011526393.2 XM_011526395.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6035142 c.75C>G p.L25L Chr.19: 41264578-41264578 on Build GRCh38
6035141 c.69A>C p.R23R Chr.19: 41264572-41264572 on Build GRCh38
5176374 c.71G>A p.G24D Chr.19: 41264574-41264574 on Build GRCh38
6035143 c.75C>G p.L25L Chr.19: 41264578-41264578 on Build GRCh38
6502425 c.5A>T p.D2V Chr.19: 41264508-41264508 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769110431 Chr.19: 41264462-41264462 on Build GRCh38
rs765505112 Chr.19: 41264491-41264491 on Build GRCh38
rs770370851 Chr.19: 41264585-41264585 on Build GRCh38
rs757286884 Chr.19: 41264545-41264545 on Build GRCh38
rs955371124 Chr.19: 41264475-41264475 on Build GRCh38
rs759076900 Chr.19: 41264499-41264499 on Build GRCh38
rs1011146280 Chr.19: 41264525-41264525 on Build GRCh38
rs764068137 Chr.19: 41264534-41264534 on Build GRCh38
rs922181810 Chr.19: 41264619-41264619 on Build GRCh38
rs994265630 Chr.19: 41264439-41264439 on Build GRCh38
rs975007400 Chr.19: 41264610-41264610 on Build GRCh38
rs770462584 Chr.19: 41264585-41264593 on Build GRCh38
rs777009112 Chr.19: 41264483-41264483 on Build GRCh38
rs979671413 Chr.19: 41264508-41264508 on Build GRCh38
rs563816385 Chr.19: 41264512-41264512 on Build GRCh38
rs369468843 Chr.19: 41264561-41264561 on Build GRCh38
rs952296567 Chr.19: 41264627-41264627 on Build GRCh38
rs55820664 Chr.19: 41264466-41264466 on Build GRCh38
rs1018269180 Chr.19: 41264494-41264494 on Build GRCh38
rs772322070 Chr.19: 41264456-41264456 on Build GRCh38
rs781389529 Chr.19: 41264581-41264581 on Build GRCh38
rs767323304 Chr.19: 41264511-41264511 on Build GRCh38
rs768734124 Chr.19: 41264576-41264576 on Build GRCh38
rs548431563 Chr.19: 41264463-41264463 on Build GRCh38
rs1022092301 Chr.19: 41264530-41264530 on Build GRCh38
rs755694742 Chr.19: 41264533-41264533 on Build GRCh38
rs964326367 Chr.19: 41264498-41264498 on Build GRCh38
rs780284901 Chr.19: 41264565-41264565 on Build GRCh38
rs753709122 Chr.19: 41264537-41264537 on Build GRCh38
rs750141693 Chr.19: 41264552-41264552 on Build GRCh38
rs752202621 Chr.19: 41264517-41264517 on Build GRCh38
rs909631795 Chr.19: 41264446-41264446 on Build GRCh38
rs771866921 Chr.19: 41264504-41264504 on Build GRCh38
rs200257089 Chr.19: 41264455-41264455 on Build GRCh38
rs926289365 Chr.19: 41264559-41264559 on Build GRCh38
rs771787711 Chr.19: 41264620-41264620 on Build GRCh38
rs773422950 Chr.19: 41264601-41264601 on Build GRCh38
rs939749345 Chr.19: 41264448-41264448 on Build GRCh38
rs750281511 Chr.19: 41264557-41264557 on Build GRCh38
rs897674510 Chr.19: 41264589-41264589 on Build GRCh38
rs775299547 Chr.19: 41264628-41264628 on Build GRCh38
rs1044733901 Chr.19: 41264544-41264544 on Build GRCh38
rs11556497 Chr.19: 41264516-41264516 on Build GRCh38
rs1056852 Chr.19: 41264582-41264582 on Build GRCh38
rs778542918 Chr.19: 41264546-41264546 on Build GRCh38
rs748595975 Chr.19: 41264583-41264583 on Build GRCh38
rs775525038 Chr.19: 41264457-41264457 on Build GRCh38
rs763428674 Chr.19: 41264609-41264609 on Build GRCh38
rs773689517 Chr.19: 41264493-41264493 on Build GRCh38
rs552267140 Chr.19: 41264572-41264572 on Build GRCh38
rs1003489667 Chr.19: 41264480-41264480 on Build GRCh38
rs1051589833 Chr.19: 41264617-41264617 on Build GRCh38
rs747115231 Chr.19: 41264461-41264461 on Build GRCh38

View Full Product Details