Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 159815320-159815545 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LY75
Entrez Gene ID
4065
Gene Name
lymphocyte antigen 75
Gene Aliases
CD205, CLEC13B, DEC-205, GP200-MR6, LY-75
UniGene
Hs.153563
Gene Chromosome Location
Chr.2: 159803355-159904756 on Build GRCh38
Transcripts
NM_002349.3 NM_001198759.1 NM_001198760.1

Gene Symbol
LY75-CD302
Entrez Gene ID
100526664
Gene Name
LY75-CD302 readthrough
Gene Aliases
-
UniGene
Hs.153563
Gene Chromosome Location
Chr.2: 159768628-159904756 on Build GRCh38
Transcripts
NM_002349.3 NM_001198759.1 NM_001198760.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5547227 c.4440delA p.G1481fs*95 Chr.2: 159815514-159815514 on Build GRCh38
5966056 c.4437G>T p.W1479C Chr.2: 159815517-159815517 on Build GRCh38
4494963 c.4432C>A p.P1478T Chr.2: 159815522-159815522 on Build GRCh38
4494964 c.4432C>T p.P1478S Chr.2: 159815522-159815522 on Build GRCh38
3568287 c.4451C>T p.S1484F Chr.2: 159815503-159815503 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370444148 Chr.2: 159815475-159815475 on Build GRCh38
rs774765706 Chr.2: 159815487-159815487 on Build GRCh38
rs780972086 Chr.2: 159815451-159815451 on Build GRCh38
rs183714454 Chr.2: 159815384-159815384 on Build GRCh38
rs975359417 Chr.2: 159815411-159815411 on Build GRCh38
rs765327132 Chr.2: 159815401-159815401 on Build GRCh38
rs370628297 Chr.2: 159815362-159815362 on Build GRCh38
rs751343272 Chr.2: 159815438-159815438 on Build GRCh38
rs772252692 Chr.2: 159815483-159815483 on Build GRCh38
rs752205195 Chr.2: 159815457-159815457 on Build GRCh38
rs779195428 Chr.2: 159815477-159815477 on Build GRCh38
rs777196981 Chr.2: 159815468-159815468 on Build GRCh38
rs767191150 Chr.2: 159815456-159815456 on Build GRCh38
rs762921805 Chr.2: 159815410-159815410 on Build GRCh38
rs921167335 Chr.2: 159815441-159815441 on Build GRCh38
rs1024828214 Chr.2: 159815376-159815376 on Build GRCh38
rs755632337 Chr.2: 159815464-159815464 on Build GRCh38
rs773299840 Chr.2: 159815404-159815404 on Build GRCh38
rs951467276 Chr.2: 159815377-159815377 on Build GRCh38
rs201028227 Chr.2: 159815393-159815393 on Build GRCh38
rs768864607 Chr.2: 159815507-159815507 on Build GRCh38
rs376112166 Chr.2: 159815388-159815388 on Build GRCh38
rs780312797 Chr.2: 159815485-159815485 on Build GRCh38
rs771289659 Chr.2: 159815363-159815363 on Build GRCh38
rs747721260 Chr.2: 159815499-159815501 on Build GRCh38
rs762118914 Chr.2: 159815499-159815499 on Build GRCh38
rs142810241 Chr.2: 159815436-159815436 on Build GRCh38
rs754624203 Chr.2: 159815445-159815445 on Build GRCh38
rs746213981 Chr.2: 159815481-159815481 on Build GRCh38
rs746100639 Chr.2: 159815370-159815370 on Build GRCh38

View Full Product Details