Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 118020809-118021082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RFC5
Entrez Gene ID
5985
Gene Name
replication factor C subunit 5
Gene Aliases
RFC36
UniGene
Hs.731908
Gene Chromosome Location
Chr.12: 118016701-118033568 on Build GRCh38
Transcripts
NM_001130112.2 NM_001206801.1 NM_007370.5 NM_181578.3 XM_011538643.2 XM_011538645.2 XM_017019779.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1264038 c.317T>A p.L106Q Chr.12: 118020955-118020955 on Build GRCh38
6841652 c.287G>A p.R96Q Chr.12: 118020925-118020925 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs200927192 Chr.12: 118020866-118020866 on Build GRCh38
rs752462565 Chr.12: 118021036-118021036 on Build GRCh38
rs774345818 Chr.12: 118020891-118020891 on Build GRCh38
rs761715858 Chr.12: 118020988-118020988 on Build GRCh38
rs776521264 Chr.12: 118020882-118020883 on Build GRCh38
rs767060845 Chr.12: 118021035-118021035 on Build GRCh38
rs754598795 Chr.12: 118021030-118021030 on Build GRCh38
rs747672890 Chr.12: 118020908-118020908 on Build GRCh38
rs760886198 Chr.12: 118020950-118020950 on Build GRCh38
rs891024926 Chr.12: 118020971-118020971 on Build GRCh38
rs199586748 Chr.12: 118020939-118020939 on Build GRCh38
rs1019822274 Chr.12: 118020959-118020959 on Build GRCh38
rs368790413 Chr.12: 118020988-118020988 on Build GRCh38
rs139634196 Chr.12: 118020872-118020872 on Build GRCh38
rs919221210 Chr.12: 118020911-118020911 on Build GRCh38
rs779558443 Chr.12: 118020924-118020924 on Build GRCh38
rs1030724937 Chr.12: 118021043-118021043 on Build GRCh38
rs534826245 Chr.12: 118020958-118020958 on Build GRCh38
rs144220409 Chr.12: 118021009-118021009 on Build GRCh38
rs746417604 Chr.12: 118020927-118020927 on Build GRCh38
rs527363092 Chr.12: 118021040-118021040 on Build GRCh38
rs746587494 Chr.12: 118021018-118021018 on Build GRCh38
rs369972918 Chr.12: 118020942-118020942 on Build GRCh38
rs762896014 Chr.12: 118021017-118021017 on Build GRCh38
rs754616055 Chr.12: 118020878-118020878 on Build GRCh38
rs771401544 Chr.12: 118021008-118021008 on Build GRCh38
rs34660755 Chr.12: 118020953-118020953 on Build GRCh38
rs780684800 Chr.12: 118020902-118020902 on Build GRCh38
rs375772110 Chr.12: 118020854-118020854 on Build GRCh38
rs758162638 Chr.12: 118020859-118020859 on Build GRCh38
rs972152212 Chr.12: 118020886-118020886 on Build GRCh38
rs768221441 Chr.12: 118021024-118021024 on Build GRCh38
rs933326302 Chr.12: 118020943-118020943 on Build GRCh38
rs759025948 Chr.12: 118020949-118020949 on Build GRCh38

View Full Product Details