Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 81933157-81933418 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PYCR1
Entrez Gene ID
5831
Gene Name
pyrroline-5-carboxylate reductase 1
Gene Aliases
ARCL2B, ARCL3B, P5C, P5CR, PIG45, PP222, PRO3, PYCR
UniGene
Hs.163451
Gene Chromosome Location
Chr.17: 81932384-81937328 on Build GRCh38
Transcripts
NM_001282279.1 NM_001282280.1 NM_001282281.1 NM_006907.3 NM_153824.2 XM_005256381.2 XM_011523583.2 XM_011523584.2 XM_011523585.2 XM_017024909.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7139969 c.867+3G>A p.? Chr.17: 81933304-81933304 on Build GRCh38
7239911 c.844A>T p.I282F Chr.17: 81933330-81933330 on Build GRCh38
4130834 c.903A>G p.S301S Chr.17: 81933271-81933271 on Build GRCh38
7139968 c.870G>A p.V290V Chr.17: 81933304-81933304 on Build GRCh38
4580255 c.884C>A p.P295H Chr.17: 81933290-81933290 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749430852 Chr.17: 81933313-81933313 on Build GRCh38
rs768639229 Chr.17: 81933320-81933320 on Build GRCh38
rs750254015 Chr.17: 81933389-81933389 on Build GRCh38
rs1031999999 Chr.17: 81933215-81933215 on Build GRCh38
rs768959337 Chr.17: 81933263-81933263 on Build GRCh38
rs61747618 Chr.17: 81933271-81933271 on Build GRCh38
rs143982437 Chr.17: 81933243-81933243 on Build GRCh38
rs761233583 Chr.17: 81933353-81933353 on Build GRCh38
rs140898502 Chr.17: 81933274-81933274 on Build GRCh38
rs946246241 Chr.17: 81933395-81933395 on Build GRCh38
rs752023923 Chr.17: 81933191-81933191 on Build GRCh38
rs34575645 Chr.17: 81933308-81933308 on Build GRCh38
rs770632870 Chr.17: 81933345-81933345 on Build GRCh38
rs748825283 Chr.17: 81933321-81933323 on Build GRCh38
rs754496020 Chr.17: 81933211-81933211 on Build GRCh38
rs757965450 Chr.17: 81933284-81933284 on Build GRCh38
rs1024706359 Chr.17: 81933270-81933270 on Build GRCh38
rs556118853 Chr.17: 81933252-81933252 on Build GRCh38
rs948856695 Chr.17: 81933372-81933372 on Build GRCh38
rs777400028 Chr.17: 81933223-81933223 on Build GRCh38
rs767043460 Chr.17: 81933354-81933354 on Build GRCh38
rs1031354107 Chr.17: 81933295-81933295 on Build GRCh38
rs373840170 Chr.17: 81933272-81933272 on Build GRCh38
rs773976380 Chr.17: 81933367-81933367 on Build GRCh38
rs772932462 Chr.17: 81933241-81933242 on Build GRCh38
rs752280717 Chr.17: 81933212-81933212 on Build GRCh38
rs1012345765 Chr.17: 81933396-81933396 on Build GRCh38
rs563377698 Chr.17: 81933357-81933357 on Build GRCh38
rs35589179 Chr.17: 81933280-81933280 on Build GRCh38
rs763519359 Chr.17: 81933382-81933382 on Build GRCh38
rs779989105 Chr.17: 81933394-81933394 on Build GRCh38
rs773780326 Chr.17: 81933349-81933349 on Build GRCh38
rs143377300 Chr.17: 81933275-81933275 on Build GRCh38
rs752293774 Chr.17: 81933282-81933282 on Build GRCh38
rs150368748 Chr.17: 81933334-81933334 on Build GRCh38
rs756020961 Chr.17: 81933390-81933390 on Build GRCh38
rs762191841 Chr.17: 81933192-81933192 on Build GRCh38
rs377033274 Chr.17: 81933299-81933299 on Build GRCh38
rs770658432 Chr.17: 81933226-81933226 on Build GRCh38
rs756964865 Chr.17: 81933384-81933384 on Build GRCh38
rs140400626 Chr.17: 81933307-81933307 on Build GRCh38
rs576089259 Chr.17: 81933210-81933210 on Build GRCh38
rs542207317 Chr.17: 81933198-81933198 on Build GRCh38
rs779964744 Chr.17: 81933310-81933310 on Build GRCh38
rs746871856 Chr.17: 81933224-81933224 on Build GRCh38
rs781071840 Chr.17: 81933227-81933227 on Build GRCh38
rs767292029 Chr.17: 81933388-81933388 on Build GRCh38
rs763754137 Chr.17: 81933297-81933297 on Build GRCh38
rs111893988 Chr.17: 81933202-81933202 on Build GRCh38
rs990588215 Chr.17: 81933242-81933242 on Build GRCh38
rs545475719 Chr.17: 81933300-81933300 on Build GRCh38
rs772506039 Chr.17: 81933336-81933336 on Build GRCh38
rs185044245 Chr.17: 81933279-81933279 on Build GRCh38
rs751169681 Chr.17: 81933383-81933383 on Build GRCh38
rs749464021 Chr.17: 81933261-81933261 on Build GRCh38
rs901236548 Chr.17: 81933374-81933374 on Build GRCh38
rs745812325 Chr.17: 81933238-81933238 on Build GRCh38
rs371347855 Chr.17: 81933333-81933333 on Build GRCh38
rs748322418 Chr.17: 81933335-81933335 on Build GRCh38
rs745470155 Chr.17: 81933258-81933258 on Build GRCh38
rs201685515 Chr.17: 81933298-81933298 on Build GRCh38
rs758139342 Chr.17: 81933213-81933213 on Build GRCh38

View Full Product Details