Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15661069-15661334 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RSC1A1
Entrez Gene ID
6248
Gene Name
regulatory solute carrier protein, family 1, member 1
Gene Aliases
RS1
UniGene
Hs.239459
Gene Chromosome Location
Chr.1: 15659713-15662030 on Build GRCh38
Transcripts
NM_006511.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
424293 c.1455C>T p.H485H Chr.1: 15661323-15661323 on Build GRCh38
1747865 c.1381C>A p.P461T Chr.1: 15661249-15661249 on Build GRCh38
3984377 c.1285T>C p.L429L Chr.1: 15661153-15661153 on Build GRCh38
3705157 c.1205A>G p.N402S Chr.1: 15661073-15661073 on Build GRCh38
6347309 c.1232G>T p.C411F Chr.1: 15661100-15661100 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376446510 Chr.1: 15661215-15661215 on Build GRCh38
rs906296542 Chr.1: 15661278-15661278 on Build GRCh38
rs533336082 Chr.1: 15661167-15661167 on Build GRCh38
rs759773926 Chr.1: 15661234-15661234 on Build GRCh38
rs777614181 Chr.1: 15661151-15661151 on Build GRCh38
rs765668691 Chr.1: 15661237-15661237 on Build GRCh38
rs758434749 Chr.1: 15661156-15661156 on Build GRCh38
rs138618442 Chr.1: 15661229-15661229 on Build GRCh38
rs200264420 Chr.1: 15661253-15661253 on Build GRCh38
rs142980050 Chr.1: 15661214-15661214 on Build GRCh38
rs542753721 Chr.1: 15661114-15661114 on Build GRCh38
rs114838139 Chr.1: 15661272-15661272 on Build GRCh38
rs776373074 Chr.1: 15661303-15661304 on Build GRCh38
rs200079396 Chr.1: 15661158-15661158 on Build GRCh38
rs777034689 Chr.1: 15661233-15661233 on Build GRCh38
rs779055774 Chr.1: 15661141-15661141 on Build GRCh38
rs766837954 Chr.1: 15661231-15661231 on Build GRCh38
rs199821679 Chr.1: 15661200-15661200 on Build GRCh38
rs762598808 Chr.1: 15661108-15661108 on Build GRCh38
rs761264316 Chr.1: 15661113-15661113 on Build GRCh38
rs912779770 Chr.1: 15661210-15661211 on Build GRCh38
rs757186434 Chr.1: 15661260-15661260 on Build GRCh38
rs372326643 Chr.1: 15661112-15661112 on Build GRCh38
rs116499237 Chr.1: 15661126-15661126 on Build GRCh38
rs770562519 Chr.1: 15661273-15661275 on Build GRCh38
rs768210759 Chr.1: 15661216-15661216 on Build GRCh38
rs145098407 Chr.1: 15661219-15661219 on Build GRCh38
rs372649223 Chr.1: 15661129-15661129 on Build GRCh38
rs1034421889 Chr.1: 15661110-15661112 on Build GRCh38
rs769464609 Chr.1: 15661186-15661186 on Build GRCh38
rs763716199 Chr.1: 15661110-15661110 on Build GRCh38
rs947751900 Chr.1: 15661251-15661251 on Build GRCh38
rs974212789 Chr.1: 15661131-15661131 on Build GRCh38
rs144378011 Chr.1: 15661157-15661157 on Build GRCh38
rs200911647 Chr.1: 15661264-15661264 on Build GRCh38
rs754225902 Chr.1: 15661123-15661123 on Build GRCh38
rs752942165 Chr.1: 15661155-15661155 on Build GRCh38
rs768478542 Chr.1: 15661282-15661282 on Build GRCh38
rs1018361976 Chr.1: 15661173-15661173 on Build GRCh38
rs116029163 Chr.1: 15661175-15661175 on Build GRCh38
rs35549389 Chr.1: 15661107-15661107 on Build GRCh38
rs745839782 Chr.1: 15661271-15661271 on Build GRCh38
rs755821385 Chr.1: 15661275-15661275 on Build GRCh38
rs758632334 Chr.1: 15661246-15661246 on Build GRCh38
rs137912259 Chr.1: 15661240-15661240 on Build GRCh38
rs190576412 Chr.1: 15661130-15661130 on Build GRCh38
rs149782354 Chr.1: 15661160-15661160 on Build GRCh38
rs769399349 Chr.1: 15661100-15661100 on Build GRCh38
rs138904592 Chr.1: 15661280-15661280 on Build GRCh38

View Full Product Details