Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39786003-39786238 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LEUTX
Entrez Gene ID
342900
Gene Name
leucine twenty homeobox
Gene Aliases
-
UniGene
Hs.451638
Gene Chromosome Location
Chr.19: 39776594-39786135 on Build GRCh38
Transcripts
NM_001143832.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4645345 c.453T>A p.N151K Chr.19: 39786081-39786081 on Build GRCh38
474721 c.381C>T p.S127S Chr.19: 39786009-39786009 on Build GRCh38
7374938 c.439A>C p.T147P Chr.19: 39786067-39786067 on Build GRCh38
223989 c.400G>A p.E134K Chr.19: 39786028-39786028 on Build GRCh38
996417 c.401A>C p.E134A Chr.19: 39786029-39786029 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772138251 Chr.19: 39786110-39786110 on Build GRCh38
rs949323487 Chr.19: 39786165-39786165 on Build GRCh38
rs200128724 Chr.19: 39786145-39786145 on Build GRCh38
rs779949872 Chr.19: 39786211-39786211 on Build GRCh38
rs777493010 Chr.19: 39786056-39786056 on Build GRCh38
rs540263352 Chr.19: 39786084-39786084 on Build GRCh38
rs147692654 Chr.19: 39786101-39786101 on Build GRCh38
rs917940016 Chr.19: 39786159-39786159 on Build GRCh38
rs61995735 Chr.19: 39786067-39786067 on Build GRCh38
rs770903563 Chr.19: 39786086-39786086 on Build GRCh38
rs1039342704 Chr.19: 39786172-39786172 on Build GRCh38
rs774382036 Chr.19: 39786093-39786093 on Build GRCh38
rs1038281537 Chr.19: 39786078-39786079 on Build GRCh38
rs905277512 Chr.19: 39786192-39786192 on Build GRCh38
rs754316195 Chr.19: 39786185-39786185 on Build GRCh38
rs149172013 Chr.19: 39786213-39786213 on Build GRCh38
rs926899678 Chr.19: 39786051-39786051 on Build GRCh38
rs760881319 Chr.19: 39786120-39786120 on Build GRCh38
rs570106042 Chr.19: 39786195-39786195 on Build GRCh38
rs377434841 Chr.19: 39786117-39786117 on Build GRCh38
rs980758380 Chr.19: 39786028-39786028 on Build GRCh38
rs930205289 Chr.19: 39786124-39786124 on Build GRCh38
rs983001717 Chr.19: 39786157-39786157 on Build GRCh38
rs745927165 Chr.19: 39786185-39786186 on Build GRCh38
rs571718226 Chr.19: 39786114-39786114 on Build GRCh38
rs919404875 Chr.19: 39786080-39786080 on Build GRCh38
rs1054016006 Chr.19: 39786208-39786208 on Build GRCh38
rs1000704423 Chr.19: 39786212-39786212 on Build GRCh38

View Full Product Details