Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 119617962-119618235 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLA1A
Entrez Gene ID
51365
Gene Name
phospholipase A1 member A
Gene Aliases
PS-PLA1, PSPLA1
UniGene
Hs.437451
Gene Chromosome Location
Chr.3: 119597848-119629811 on Build GRCh38
Transcripts
NM_001206960.1 NM_001206961.1 NM_001293225.1 NM_015900.3 NR_120610.1 XM_017006572.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4559719 c.816G>A p.E272E Chr.3: 119618080-119618080 on Build GRCh38
3914636 c.858G>A p.K286K Chr.3: 119618122-119618122 on Build GRCh38
3585763 c.799T>C p.Y267H Chr.3: 119618063-119618063 on Build GRCh38
6163080 c.786G>T p.R262S Chr.3: 119618050-119618050 on Build GRCh38
5694793 c.917G>T p.R306M Chr.3: 119618181-119618181 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755813713 Chr.3: 119618096-119618096 on Build GRCh38
rs994295789 Chr.3: 119618111-119618111 on Build GRCh38
rs767853819 Chr.3: 119618072-119618072 on Build GRCh38
rs2692622 Chr.3: 119618115-119618115 on Build GRCh38
rs746874424 Chr.3: 119618130-119618130 on Build GRCh38
rs778947808 Chr.3: 119618013-119618013 on Build GRCh38
rs748862154 Chr.3: 119617994-119617994 on Build GRCh38
rs567965587 Chr.3: 119618201-119618201 on Build GRCh38
rs774297705 Chr.3: 119618143-119618143 on Build GRCh38
rs758356076 Chr.3: 119618203-119618203 on Build GRCh38
rs775545038 Chr.3: 119618058-119618058 on Build GRCh38
rs113578288 Chr.3: 119618151-119618151 on Build GRCh38
rs778305760 Chr.3: 119618108-119618108 on Build GRCh38
rs777614341 Chr.3: 119618129-119618129 on Build GRCh38
rs761157942 Chr.3: 119618183-119618183 on Build GRCh38
rs149622019 Chr.3: 119618059-119618059 on Build GRCh38
rs939338424 Chr.3: 119618012-119618012 on Build GRCh38
rs754599725 Chr.3: 119618002-119618003 on Build GRCh38
rs750050449 Chr.3: 119617989-119617989 on Build GRCh38
rs373062235 Chr.3: 119618182-119618182 on Build GRCh38
rs111929583 Chr.3: 119618106-119618106 on Build GRCh38
rs747870078 Chr.3: 119618024-119618024 on Build GRCh38
rs771863062 Chr.3: 119618029-119618029 on Build GRCh38
rs766105812 Chr.3: 119618185-119618185 on Build GRCh38
rs753489795 Chr.3: 119618186-119618186 on Build GRCh38
rs764748501 Chr.3: 119618192-119618192 on Build GRCh38
rs143661120 Chr.3: 119617992-119617992 on Build GRCh38
rs115114323 Chr.3: 119618071-119618071 on Build GRCh38
rs773704485 Chr.3: 119618152-119618152 on Build GRCh38
rs1004620324 Chr.3: 119618019-119618019 on Build GRCh38
rs895394764 Chr.3: 119618036-119618036 on Build GRCh38
rs766146755 Chr.3: 119618134-119618134 on Build GRCh38
rs368870376 Chr.3: 119618138-119618138 on Build GRCh38
rs767966523 Chr.3: 119618147-119618147 on Build GRCh38
rs1055344286 Chr.3: 119618034-119618034 on Build GRCh38
rs779916653 Chr.3: 119618102-119618102 on Build GRCh38
rs762323519 Chr.3: 119618146-119618146 on Build GRCh38
rs750766764 Chr.3: 119618083-119618083 on Build GRCh38
rs549859286 Chr.3: 119618191-119618191 on Build GRCh38
rs758286098 Chr.3: 119618125-119618125 on Build GRCh38
rs764488329 Chr.3: 119618070-119618070 on Build GRCh38
rs138449882 Chr.3: 119618049-119618049 on Build GRCh38
rs768705242 Chr.3: 119618009-119618009 on Build GRCh38
rs753363283 Chr.3: 119618105-119618105 on Build GRCh38
rs749536205 Chr.3: 119618139-119618139 on Build GRCh38
rs202063140 Chr.3: 119618040-119618040 on Build GRCh38

View Full Product Details