Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 34319874-34320147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0355
Entrez Gene ID
9710
Gene Name
KIAA0355
Gene Aliases
-
UniGene
Hs.330073
Gene Chromosome Location
Chr.19: 34254551-34355566 on Build GRCh38
Transcripts
NM_014686.3 XM_005259444.1 XM_017027525.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3532291 c.622A>C p.S208R Chr.19: 34320033-34320033 on Build GRCh38
5181018 c.526C>A p.Q176K Chr.19: 34319937-34319937 on Build GRCh38
6259846 c.606G>T p.V202V Chr.19: 34320017-34320017 on Build GRCh38
1717118 c.503G>T p.G168V Chr.19: 34319914-34319914 on Build GRCh38
5112390 c.633C>T p.G211G Chr.19: 34320044-34320044 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748040528 Chr.19: 34319897-34319897 on Build GRCh38
rs200482388 Chr.19: 34320035-34320035 on Build GRCh38
rs137997380 Chr.19: 34320024-34320024 on Build GRCh38
rs764106013 Chr.19: 34319934-34319934 on Build GRCh38
rs754154292 Chr.19: 34320048-34320048 on Build GRCh38
rs763144975 Chr.19: 34319933-34319933 on Build GRCh38
rs777213658 Chr.19: 34320092-34320092 on Build GRCh38
rs769630331 Chr.19: 34320105-34320105 on Build GRCh38
rs778495001 Chr.19: 34319984-34319984 on Build GRCh38
rs146969264 Chr.19: 34320020-34320020 on Build GRCh38
rs373171001 Chr.19: 34320044-34320044 on Build GRCh38
rs759221186 Chr.19: 34320055-34320055 on Build GRCh38
rs765004210 Chr.19: 34320061-34320061 on Build GRCh38
rs777743074 Chr.19: 34319994-34319994 on Build GRCh38
rs781001710 Chr.19: 34320103-34320103 on Build GRCh38
rs992582048 Chr.19: 34320123-34320123 on Build GRCh38
rs766706307 Chr.19: 34320047-34320047 on Build GRCh38
rs75615202 Chr.19: 34320023-34320023 on Build GRCh38
rs761141307 Chr.19: 34320045-34320045 on Build GRCh38
rs748623701 Chr.19: 34319984-34319985 on Build GRCh38
rs772182883 Chr.19: 34320038-34320038 on Build GRCh38
rs766722482 Chr.19: 34319974-34319974 on Build GRCh38
rs1009408713 Chr.19: 34320022-34320022 on Build GRCh38
rs752282451 Chr.19: 34319987-34319987 on Build GRCh38
rs143376054 Chr.19: 34319971-34319971 on Build GRCh38
rs779079796 Chr.19: 34320107-34320107 on Build GRCh38
rs768038646 Chr.19: 34319965-34319965 on Build GRCh38
rs758842315 Chr.19: 34320103-34320103 on Build GRCh38
rs748438407 Chr.19: 34320125-34320125 on Build GRCh38
rs141496341 Chr.19: 34320015-34320015 on Build GRCh38
rs758804996 Chr.19: 34320088-34320088 on Build GRCh38
rs369744932 Chr.19: 34319983-34319983 on Build GRCh38
rs148353127 Chr.19: 34319981-34319981 on Build GRCh38
rs775748003 Chr.19: 34319926-34319926 on Build GRCh38
rs762268753 Chr.19: 34319961-34319961 on Build GRCh38
rs375428393 Chr.19: 34320102-34320102 on Build GRCh38
rs749461324 Chr.19: 34320018-34320018 on Build GRCh38
rs770677162 Chr.19: 34319925-34319925 on Build GRCh38
rs757148178 Chr.19: 34320094-34320094 on Build GRCh38
rs758361463 Chr.19: 34319993-34319993 on Build GRCh38
rs960883080 Chr.19: 34320108-34320108 on Build GRCh38
rs772139556 Chr.19: 34319907-34319907 on Build GRCh38
rs199555885 Chr.19: 34320093-34320093 on Build GRCh38
rs375356637 Chr.19: 34319966-34319966 on Build GRCh38
rs76031628 Chr.19: 34320109-34320109 on Build GRCh38
rs370744952 Chr.19: 34319917-34319917 on Build GRCh38
rs757950270 Chr.19: 34320082-34320082 on Build GRCh38
rs746839362 Chr.19: 34319918-34319918 on Build GRCh38
rs746958754 Chr.19: 34319996-34319996 on Build GRCh38
rs981852962 Chr.19: 34320049-34320049 on Build GRCh38
rs774750225 Chr.19: 34320014-34320014 on Build GRCh38
rs752446054 Chr.19: 34320073-34320073 on Build GRCh38
rs149223464 Chr.19: 34319959-34319959 on Build GRCh38

View Full Product Details