Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 55729199-55729460 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RRAGB
Entrez Gene ID
10325
Gene Name
Ras related GTP binding B
Gene Aliases
RAGB, bA465E19.1
UniGene
Hs.50282
Gene Chromosome Location
Chr.X: 55717677-55760209 on Build GRCh38
Transcripts
NM_006064.4 NM_016656.3 XM_005261973.4 XM_017029217.1 XM_017029218.1

SNP Detail

SNP ID Chromosome Location
rs750962497 Chr.X: 55729399-55729399 on Build GRCh38
rs770010761 Chr.X: 55729331-55729331 on Build GRCh38
rs987804252 Chr.X: 55729354-55729354 on Build GRCh38
rs767870303 Chr.X: 55729394-55729394 on Build GRCh38
rs763263878 Chr.X: 55729346-55729346 on Build GRCh38
rs781176789 Chr.X: 55729273-55729273 on Build GRCh38
rs778487275 Chr.X: 55729248-55729248 on Build GRCh38
rs763158984 Chr.X: 55729419-55729419 on Build GRCh38
rs746876257 Chr.X: 55729279-55729279 on Build GRCh38
rs775789778 Chr.X: 55729337-55729337 on Build GRCh38
rs922826792 Chr.X: 55729327-55729327 on Build GRCh38
rs370308035 Chr.X: 55729406-55729406 on Build GRCh38
rs776664010 Chr.X: 55729311-55729311 on Build GRCh38
rs764470852 Chr.X: 55729363-55729363 on Build GRCh38
rs192324793 Chr.X: 55729261-55729261 on Build GRCh38
rs764727475 Chr.X: 55729407-55729407 on Build GRCh38
rs368767476 Chr.X: 55729393-55729393 on Build GRCh38
rs941093679 Chr.X: 55729414-55729414 on Build GRCh38
rs745892188 Chr.X: 55729312-55729312 on Build GRCh38
rs759486880 Chr.X: 55729416-55729416 on Build GRCh38
rs769976699 Chr.X: 55729302-55729302 on Build GRCh38
rs777537702 Chr.X: 55729269-55729269 on Build GRCh38
rs201368308 Chr.X: 55729381-55729381 on Build GRCh38

View Full Product Details