Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 45416520-45416753 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIGT
Entrez Gene ID
51604
Gene Name
phosphatidylinositol glycan anchor biosynthesis class T
Gene Aliases
CGI-06, MCAHS3, NDAP, PNH2
UniGene
Hs.437388
Gene Chromosome Location
Chr.20: 45416067-45426245 on Build GRCh38
Transcripts
NM_001184728.2 NM_001184729.2 NM_001184730.2 NM_015937.5 NR_047691.1 NR_047692.1 NR_047693.1 NR_047694.1 NR_047695.1 XM_005260430.2 XM_005260432.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
478205 c.264A>G p.S88S Chr.20: 45416593-45416593 on Build GRCh38
1412065 c.289C>A p.R97R Chr.20: 45416618-45416618 on Build GRCh38
1027240 c.222G>A p.G74G Chr.20: 45416551-45416551 on Build GRCh38
4098833 c.225G>A p.Q75Q Chr.20: 45416554-45416554 on Build GRCh38
1027238 c.211A>G p.K71E Chr.20: 45416540-45416540 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766618775 Chr.20: 45416555-45416555 on Build GRCh38
rs757606139 Chr.20: 45416696-45416696 on Build GRCh38
rs779102977 Chr.20: 45416699-45416699 on Build GRCh38
rs560732287 Chr.20: 45416603-45416603 on Build GRCh38
rs772946352 Chr.20: 45416636-45416636 on Build GRCh38
rs376863939 Chr.20: 45416685-45416685 on Build GRCh38
rs201701603 Chr.20: 45416635-45416635 on Build GRCh38
rs11556788 Chr.20: 45416645-45416645 on Build GRCh38
rs137946450 Chr.20: 45416550-45416550 on Build GRCh38
rs932941807 Chr.20: 45416575-45416575 on Build GRCh38
rs756632799 Chr.20: 45416579-45416579 on Build GRCh38
rs897714473 Chr.20: 45416659-45416659 on Build GRCh38
rs751648642 Chr.20: 45416556-45416556 on Build GRCh38
rs753279974 Chr.20: 45416577-45416577 on Build GRCh38
rs765035657 Chr.20: 45416546-45416546 on Build GRCh38
rs771684622 Chr.20: 45416593-45416593 on Build GRCh38
rs1026875429 Chr.20: 45416734-45416734 on Build GRCh38
rs542352635 Chr.20: 45416586-45416586 on Build GRCh38
rs534024741 Chr.20: 45416676-45416676 on Build GRCh38
rs751486189 Chr.20: 45416719-45416720 on Build GRCh38
rs759784562 Chr.20: 45416671-45416671 on Build GRCh38
rs762795664 Chr.20: 45416641-45416641 on Build GRCh38
rs751840271 Chr.20: 45416669-45416669 on Build GRCh38
rs993379025 Chr.20: 45416662-45416662 on Build GRCh38
rs1051525043 Chr.20: 45416665-45416665 on Build GRCh38
rs747651292 Chr.20: 45416728-45416728 on Build GRCh38
rs376471871 Chr.20: 45416585-45416585 on Build GRCh38
rs779731196 Chr.20: 45416602-45416602 on Build GRCh38
rs781381066 Chr.20: 45416569-45416569 on Build GRCh38
rs904855157 Chr.20: 45416722-45416722 on Build GRCh38
rs750159732 Chr.20: 45416549-45416549 on Build GRCh38
rs200192855 Chr.20: 45416710-45416710 on Build GRCh38
rs776068420 Chr.20: 45416609-45416609 on Build GRCh38
rs371128851 Chr.20: 45416587-45416587 on Build GRCh38
rs768183630 Chr.20: 45416608-45416608 on Build GRCh38
rs755055454 Chr.20: 45416565-45416565 on Build GRCh38
rs780683262 Chr.20: 45416713-45416713 on Build GRCh38
rs892177137 Chr.20: 45416697-45416697 on Build GRCh38
rs767720044 Chr.20: 45416674-45416674 on Build GRCh38
rs768237032 Chr.20: 45416711-45416711 on Build GRCh38
rs200326105 Chr.20: 45416652-45416652 on Build GRCh38
rs747604533 Chr.20: 45416638-45416638 on Build GRCh38
rs142960172 Chr.20: 45416677-45416677 on Build GRCh38
rs1043268305 Chr.20: 45416576-45416576 on Build GRCh38
rs761751196 Chr.20: 45416623-45416623 on Build GRCh38

View Full Product Details