Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 20537002-20537257 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MED15
Entrez Gene ID
51586
Gene Name
mediator complex subunit 15
Gene Aliases
ARC105, CAG7A, CTG7A, PCQAP, TIG-1, TIG1, TNRC7
UniGene
Hs.517421
Gene Chromosome Location
Chr.22: 20507542-20587632 on Build GRCh38
Transcripts
NM_001003891.2 NM_001293234.1 NM_001293235.1 NM_001293236.1 NM_001293237.1 NM_015889.4 XM_006724263.3 XM_006724264.3 XM_011530214.2 XM_011530218.2 XM_011530219.1 XM_011530220.2 XM_017028818.1 XM_017028820.1 XM_017028821.1 XM_017028822.1 XM_017028823.1 XM_017028824.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7079462 c.81G>A p.M27I Chr.22: 20537129-20537129 on Build GRCh38
6710771 c.102C>T p.H34H Chr.22: 20537150-20537150 on Build GRCh38
6710772 c.102C>T p.H34H Chr.22: 20537150-20537150 on Build GRCh38
3371862 c.104G>T p.S35I Chr.22: 20537152-20537152 on Build GRCh38
7288399 c.95T>C p.V32A Chr.22: 20537143-20537143 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs67604175 Chr.22: 20537212-20537213 on Build GRCh38
rs931323747 Chr.22: 20537060-20537060 on Build GRCh38
rs73386128 Chr.22: 20537195-20537195 on Build GRCh38
rs777700862 Chr.22: 20537117-20537117 on Build GRCh38
rs746869569 Chr.22: 20537120-20537120 on Build GRCh38
rs141012864 Chr.22: 20537201-20537201 on Build GRCh38
rs5995779 Chr.22: 20537079-20537079 on Build GRCh38
rs191935159 Chr.22: 20537078-20537078 on Build GRCh38
rs377582136 Chr.22: 20537068-20537068 on Build GRCh38
rs764384350 Chr.22: 20537080-20537080 on Build GRCh38
rs781683161 Chr.22: 20537212-20537212 on Build GRCh38
rs1012580926 Chr.22: 20537141-20537141 on Build GRCh38
rs3216923 Chr.22: 20537215-20537216 on Build GRCh38
rs769308794 Chr.22: 20537200-20537200 on Build GRCh38
rs769218488 Chr.22: 20537210-20537210 on Build GRCh38
rs75227685 Chr.22: 20537093-20537093 on Build GRCh38
rs776279285 Chr.22: 20537067-20537067 on Build GRCh38
rs367550499 Chr.22: 20537228-20537228 on Build GRCh38
rs746417632 Chr.22: 20537232-20537244 on Build GRCh38
rs758356228 Chr.22: 20537230-20537230 on Build GRCh38
rs60685513 Chr.22: 20537056-20537056 on Build GRCh38
rs201541352 Chr.22: 20537217-20537217 on Build GRCh38
rs1031775446 Chr.22: 20537213-20537213 on Build GRCh38
rs770180506 Chr.22: 20537074-20537074 on Build GRCh38
rs766420404 Chr.22: 20537221-20537221 on Build GRCh38
rs571911918 Chr.22: 20537095-20537095 on Build GRCh38
rs756941517 Chr.22: 20537187-20537187 on Build GRCh38
rs1026270613 Chr.22: 20537031-20537031 on Build GRCh38
rs752608762 Chr.22: 20537100-20537100 on Build GRCh38
rs977373278 Chr.22: 20537083-20537083 on Build GRCh38
rs1022678209 Chr.22: 20537146-20537146 on Build GRCh38
rs975632642 Chr.22: 20537076-20537076 on Build GRCh38
rs751978873 Chr.22: 20537081-20537081 on Build GRCh38
rs762016950 Chr.22: 20537090-20537090 on Build GRCh38
rs745530880 Chr.22: 20537196-20537196 on Build GRCh38
rs201596107 Chr.22: 20537210-20537211 on Build GRCh38
rs374552341 Chr.22: 20537220-20537220 on Build GRCh38
rs749789102 Chr.22: 20537203-20537203 on Build GRCh38
rs921234376 Chr.22: 20537038-20537038 on Build GRCh38
rs148534902 Chr.22: 20537211-20537212 on Build GRCh38
rs952052288 Chr.22: 20537042-20537042 on Build GRCh38
rs539211608 Chr.22: 20537028-20537028 on Build GRCh38
rs896007327 Chr.22: 20537140-20537140 on Build GRCh38
rs1006230444 Chr.22: 20537045-20537045 on Build GRCh38
rs555936126 Chr.22: 20537066-20537066 on Build GRCh38
rs767766150 Chr.22: 20537092-20537092 on Build GRCh38
rs1018121172 Chr.22: 20537065-20537065 on Build GRCh38
rs758284408 Chr.22: 20537102-20537102 on Build GRCh38

View Full Product Details