Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 80654713-80654986 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHFR
Entrez Gene ID
1719
Gene Name
dihydrofolate reductase
Gene Aliases
DHFRP1, DYR
UniGene
Hs.592364
Gene Chromosome Location
Chr.5: 80626226-80654981 on Build GRCh38
Transcripts
NM_000791.3 NM_001290354.1 NM_001290357.1 NR_110936.1 NM_002439.4

Gene Symbol
MSH3
Entrez Gene ID
4437
Gene Name
mutS homolog 3
Gene Aliases
DUP, MRP1
UniGene
Hs.280987
Gene Chromosome Location
Chr.5: 80654648-80876815 on Build GRCh38
Transcripts
NM_000791.3 NM_001290354.1 NM_001290357.1 NR_110936.1 NM_002439.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7143241 c.33C>G p.L11L Chr.5: 80654760-80654760 on Build GRCh38
5445955 c.181-9_181-1delCCAGCTCCC p.? Chr.5: 80654926-80654934 on Build GRCh38
1311352 c.33C>T p.L11L Chr.5: 80654760-80654760 on Build GRCh38
4333492 c.199C>G p.P67A Chr.5: 80654926-80654926 on Build GRCh38
5960260 c.180+2_180+10delCAGCGCCCC p.? Chr.5: 80654909-80654917 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758656822 Chr.5: 80654911-80654911 on Build GRCh38
rs770462221 Chr.5: 80654890-80654898 on Build GRCh38
rs772266495 Chr.5: 80654868-80654868 on Build GRCh38
rs755000466 Chr.5: 80654880-80654881 on Build GRCh38
rs774455792 Chr.5: 80654734-80654734 on Build GRCh38
rs559669988 Chr.5: 80654829-80654829 on Build GRCh38
rs781007137 Chr.5: 80654881-80654907 on Build GRCh38
rs879531814 Chr.5: 80654922-80654931 on Build GRCh38
rs750753473 Chr.5: 80654896-80654917 on Build GRCh38
rs748485429 Chr.5: 80654907-80654907 on Build GRCh38
rs6151597 Chr.5: 80654748-80654748 on Build GRCh38
rs749001291 Chr.5: 80654889-80654893 on Build GRCh38
rs1574197 Chr.5: 80654908-80654908 on Build GRCh38
rs944680882 Chr.5: 80654774-80654774 on Build GRCh38
rs747419370 Chr.5: 80654908-80654909 on Build GRCh38
rs770677713 Chr.5: 80654907-80654908 on Build GRCh38
rs751264470 Chr.5: 80654850-80654850 on Build GRCh38
rs773388641 Chr.5: 80654870-80654870 on Build GRCh38
rs762866873 Chr.5: 80654844-80654844 on Build GRCh38
rs770190473 Chr.5: 80654803-80654803 on Build GRCh38
rs755472095 Chr.5: 80654902-80654902 on Build GRCh38
rs777602777 Chr.5: 80654759-80654759 on Build GRCh38
rs577919981 Chr.5: 80654932-80654932 on Build GRCh38
rs145429648 Chr.5: 80654823-80654823 on Build GRCh38
rs1024354123 Chr.5: 80654866-80654866 on Build GRCh38
rs144776112 Chr.5: 80654896-80654896 on Build GRCh38
rs928378977 Chr.5: 80654855-80654855 on Build GRCh38
rs747024539 Chr.5: 80654871-80654871 on Build GRCh38
rs758960105 Chr.5: 80654890-80654907 on Build GRCh38
rs374904719 Chr.5: 80654743-80654743 on Build GRCh38
rs770922346 Chr.5: 80654916-80654916 on Build GRCh38
rs368081203 Chr.5: 80654841-80654841 on Build GRCh38
rs779180469 Chr.5: 80654863-80654863 on Build GRCh38
rs879145456 Chr.5: 80654802-80654802 on Build GRCh38
rs774275160 Chr.5: 80654877-80654877 on Build GRCh38
rs775307118 Chr.5: 80654872-80654872 on Build GRCh38
rs969715234 Chr.5: 80654762-80654762 on Build GRCh38
rs141080879 Chr.5: 80654856-80654856 on Build GRCh38
rs1049679865 Chr.5: 80654837-80654839 on Build GRCh38
rs774328125 Chr.5: 80654920-80654920 on Build GRCh38
rs144629981 Chr.5: 80654923-80654931 on Build GRCh38
rs762256716 Chr.5: 80654890-80654891 on Build GRCh38
rs761912573 Chr.5: 80654737-80654737 on Build GRCh38
rs766982358 Chr.5: 80654890-80654906 on Build GRCh38
rs780220914 Chr.5: 80654877-80654878 on Build GRCh38
rs751761357 Chr.5: 80654933-80654934 on Build GRCh38
rs2001675 Chr.5: 80654905-80654905 on Build GRCh38
rs766700228 Chr.5: 80654930-80654933 on Build GRCh38
rs3045983 Chr.5: 80654926-80654934 on Build GRCh38
rs1650697 Chr.5: 80654962-80654962 on Build GRCh38
rs60484572 Chr.5: 80654930-80654931 on Build GRCh38
rs563338833 Chr.5: 80654742-80654742 on Build GRCh38
rs201874762 Chr.5: 80654881-80654898 on Build GRCh38
rs1047489 Chr.5: 80654931-80654931 on Build GRCh38
rs767196321 Chr.5: 80654852-80654852 on Build GRCh38
rs751748318 Chr.5: 80654898-80654905 on Build GRCh38
rs759573423 Chr.5: 80654837-80654837 on Build GRCh38
rs758718643 Chr.5: 80654905-80654906 on Build GRCh38
rs564921007 Chr.5: 80654760-80654760 on Build GRCh38
rs773983405 Chr.5: 80654889-80654890 on Build GRCh38
rs758709284 Chr.5: 80654943-80654943 on Build GRCh38
rs755017868 Chr.5: 80654904-80654905 on Build GRCh38
rs765698219 Chr.5: 80654935-80654935 on Build GRCh38
rs557874766 Chr.5: 80654926-80654926 on Build GRCh38
rs146523896 Chr.5: 80654838-80654838 on Build GRCh38
rs780785898 Chr.5: 80654783-80654783 on Build GRCh38
rs564858245 Chr.5: 80654873-80654873 on Build GRCh38
rs773094831 Chr.5: 80654888-80654888 on Build GRCh38
rs70991168 Chr.5: 80654913-80654914 on Build GRCh38
rs765637112 Chr.5: 80654895-80654895 on Build GRCh38
rs780517196 Chr.5: 80654804-80654804 on Build GRCh38
rs753160687 Chr.5: 80654937-80654937 on Build GRCh38
rs756684854 Chr.5: 80654857-80654857 on Build GRCh38
rs769744913 Chr.5: 80654884-80654900 on Build GRCh38
rs530525176 Chr.5: 80654887-80654887 on Build GRCh38
rs763928973 Chr.5: 80654849-80654849 on Build GRCh38
rs767295239 Chr.5: 80654923-80654923 on Build GRCh38
rs201149584 Chr.5: 80654900-80654900 on Build GRCh38
rs879791226 Chr.5: 80654923-80654932 on Build GRCh38
rs201906899 Chr.5: 80654898-80654898 on Build GRCh38
rs910557064 Chr.5: 80654880-80654880 on Build GRCh38
rs777564333 Chr.5: 80654889-80654915 on Build GRCh38
rs148550291 Chr.5: 80654899-80654899 on Build GRCh38
rs780457592 Chr.5: 80654858-80654858 on Build GRCh38
rs748032478 Chr.5: 80654882-80654882 on Build GRCh38
rs745490043 Chr.5: 80654909-80654917 on Build GRCh38
rs767048342 Chr.5: 80654888-80654888 on Build GRCh38
rs761734241 Chr.5: 80654922-80654922 on Build GRCh38
rs774577965 Chr.5: 80654822-80654822 on Build GRCh38
rs757491337 Chr.5: 80654952-80654952 on Build GRCh38
rs759756736 Chr.5: 80654937-80654943 on Build GRCh38
rs758595508 Chr.5: 80654878-80654886 on Build GRCh38
rs972547684 Chr.5: 80654862-80654862 on Build GRCh38
rs749754328 Chr.5: 80654859-80654859 on Build GRCh38
rs748434336 Chr.5: 80654864-80654864 on Build GRCh38
rs761607646 Chr.5: 80654883-80654883 on Build GRCh38
rs746455200 Chr.5: 80654800-80654800 on Build GRCh38
rs751800062 Chr.5: 80654947-80654947 on Build GRCh38
rs773158640 Chr.5: 80654833-80654833 on Build GRCh38
rs372442835 Chr.5: 80654758-80654758 on Build GRCh38
rs369278563 Chr.5: 80654753-80654753 on Build GRCh38
rs2405876 Chr.5: 80654914-80654914 on Build GRCh38
rs2405875 Chr.5: 80654889-80654889 on Build GRCh38
rs768844493 Chr.5: 80654732-80654732 on Build GRCh38
rs976119718 Chr.5: 80654775-80654775 on Build GRCh38
rs535056167 Chr.5: 80654912-80654912 on Build GRCh38
rs765727057 Chr.5: 80654909-80654910 on Build GRCh38
rs773624233 Chr.5: 80654914-80654919 on Build GRCh38
rs756968089 Chr.5: 80654851-80654851 on Build GRCh38
rs2405877 Chr.5: 80654917-80654917 on Build GRCh38
rs768967342 Chr.5: 80654814-80654814 on Build GRCh38
rs749586225 Chr.5: 80654808-80654808 on Build GRCh38
rs778136308 Chr.5: 80654946-80654946 on Build GRCh38
rs758685508 Chr.5: 80654896-80654897 on Build GRCh38
rs768692915 Chr.5: 80654918-80654918 on Build GRCh38
rs755350309 Chr.5: 80654860-80654860 on Build GRCh38
rs756843242 Chr.5: 80654765-80654765 on Build GRCh38
rs772142059 Chr.5: 80654824-80654824 on Build GRCh38

View Full Product Details