Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 119881942-119882215 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTPN4
Entrez Gene ID
5775
Gene Name
protein tyrosine phosphatase, non-receptor type 4
Gene Aliases
MEG, PTPMEG, PTPMEG1
UniGene
Hs.469809
Gene Chromosome Location
Chr.2: 119759631-119984899 on Build GRCh38
Transcripts
NM_002830.3 XM_011511560.2 XM_011511562.2 XM_017004596.1 XM_017004597.1 XM_017004598.1 XM_017004599.1 XM_017004600.1 XM_017004601.1 XM_017004602.1 XM_017004603.1 XM_017004604.1 XM_017004605.1 XM_017004606.1 XM_017004607.1 XM_017004608.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5891702 c.458C>T p.A153V Chr.2: 119882141-119882141 on Build GRCh38
6153574 c.419C>T p.P140L Chr.2: 119882102-119882102 on Build GRCh38
6451501 c.419C>A p.P140H Chr.2: 119882102-119882102 on Build GRCh38
6758949 c.425_427delCTT p.S143delS Chr.2: 119882108-119882110 on Build GRCh38
1305594 c.463C>G p.Q155E Chr.2: 119882146-119882146 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761232747 Chr.2: 119882164-119882165 on Build GRCh38
rs979407553 Chr.2: 119882006-119882006 on Build GRCh38
rs970367604 Chr.2: 119881989-119881993 on Build GRCh38
rs776152148 Chr.2: 119882076-119882076 on Build GRCh38
rs370261737 Chr.2: 119882092-119882092 on Build GRCh38
rs1039693494 Chr.2: 119882181-119882181 on Build GRCh38
rs530674054 Chr.2: 119882003-119882003 on Build GRCh38
rs868582705 Chr.2: 119882102-119882102 on Build GRCh38
rs551388846 Chr.2: 119881975-119881975 on Build GRCh38
rs537581595 Chr.2: 119882068-119882068 on Build GRCh38
rs377716798 Chr.2: 119882082-119882082 on Build GRCh38
rs138928846 Chr.2: 119882110-119882110 on Build GRCh38
rs781205857 Chr.2: 119882146-119882146 on Build GRCh38
rs764578175 Chr.2: 119882189-119882190 on Build GRCh38
rs750037269 Chr.2: 119882087-119882087 on Build GRCh38
rs781638770 Chr.2: 119881983-119881983 on Build GRCh38
rs180770969 Chr.2: 119882173-119882173 on Build GRCh38
rs758252247 Chr.2: 119882091-119882091 on Build GRCh38
rs566644975 Chr.2: 119882150-119882150 on Build GRCh38
rs901159452 Chr.2: 119882184-119882184 on Build GRCh38
rs754993659 Chr.2: 119882141-119882141 on Build GRCh38
rs775190537 Chr.2: 119882076-119882076 on Build GRCh38
rs200467778 Chr.2: 119882055-119882055 on Build GRCh38
rs767106406 Chr.2: 119882068-119882068 on Build GRCh38
rs767963109 Chr.2: 119882072-119882072 on Build GRCh38
rs374217540 Chr.2: 119882171-119882171 on Build GRCh38
rs563213687 Chr.2: 119881981-119881981 on Build GRCh38

View Full Product Details