Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35617247-35617472 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAUS5
Entrez Gene ID
23354
Gene Name
HAUS augmin like complex subunit 5
Gene Aliases
KIAA0841, dgt5
UniGene
Hs.7426
Gene Chromosome Location
Chr.19: 35612744-35625349 on Build GRCh38
Transcripts
NM_015302.1 XM_011526684.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5591225 c.579C>T p.T193T Chr.19: 35617310-35617310 on Build GRCh38
1304445 c.585G>T p.R195R Chr.19: 35617316-35617316 on Build GRCh38
6677743 c.616C>A p.Q206K Chr.19: 35617347-35617347 on Build GRCh38
6898744 c.632G>A p.S211N Chr.19: 35617363-35617363 on Build GRCh38
4620078 c.604C>T p.L202F Chr.19: 35617335-35617335 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769285187 Chr.19: 35617393-35617393 on Build GRCh38
rs370787544 Chr.19: 35617287-35617287 on Build GRCh38
rs201575634 Chr.19: 35617335-35617335 on Build GRCh38
rs3216878 Chr.19: 35617429-35617432 on Build GRCh38
rs777498582 Chr.19: 35617419-35617420 on Build GRCh38
rs376458399 Chr.19: 35617321-35617321 on Build GRCh38
rs758987275 Chr.19: 35617380-35617380 on Build GRCh38
rs779161201 Chr.19: 35617375-35617375 on Build GRCh38
rs1024482828 Chr.19: 35617350-35617350 on Build GRCh38
rs370921589 Chr.19: 35617407-35617407 on Build GRCh38
rs765001943 Chr.19: 35617415-35617415 on Build GRCh38
rs1037716074 Chr.19: 35617442-35617442 on Build GRCh38
rs975436509 Chr.19: 35617414-35617414 on Build GRCh38
rs967825008 Chr.19: 35617365-35617365 on Build GRCh38
rs776186755 Chr.19: 35617387-35617387 on Build GRCh38
rs921159325 Chr.19: 35617449-35617449 on Build GRCh38
rs368652044 Chr.19: 35617397-35617397 on Build GRCh38
rs202231807 Chr.19: 35617297-35617297 on Build GRCh38
rs762385480 Chr.19: 35617405-35617405 on Build GRCh38
rs774941764 Chr.19: 35617296-35617296 on Build GRCh38
rs748079213 Chr.19: 35617269-35617271 on Build GRCh38
rs201350206 Chr.19: 35617388-35617388 on Build GRCh38
rs771306960 Chr.19: 35617384-35617384 on Build GRCh38
rs757073996 Chr.19: 35617333-35617333 on Build GRCh38
rs956355538 Chr.19: 35617309-35617309 on Build GRCh38
rs747317262 Chr.19: 35617283-35617283 on Build GRCh38
rs866040130 Chr.19: 35617318-35617318 on Build GRCh38
rs747437680 Chr.19: 35617382-35617382 on Build GRCh38
rs773755662 Chr.19: 35617314-35617314 on Build GRCh38
rs2301596 Chr.19: 35617369-35617369 on Build GRCh38
rs778355788 Chr.19: 35617381-35617381 on Build GRCh38
rs755048584 Chr.19: 35617370-35617370 on Build GRCh38
rs764087814 Chr.19: 35617411-35617411 on Build GRCh38
rs751363111 Chr.19: 35617315-35617315 on Build GRCh38
rs202201760 Chr.19: 35617288-35617288 on Build GRCh38
rs370452206 Chr.19: 35617355-35617355 on Build GRCh38
rs138930954 Chr.19: 35617424-35617424 on Build GRCh38
rs188968038 Chr.19: 35617378-35617378 on Build GRCh38
rs200534275 Chr.19: 35617282-35617282 on Build GRCh38
rs774255629 Chr.19: 35617418-35617418 on Build GRCh38
rs772081857 Chr.19: 35617322-35617322 on Build GRCh38
rs764773040 Chr.19: 35617292-35617292 on Build GRCh38
rs200781781 Chr.19: 35617281-35617281 on Build GRCh38

View Full Product Details