Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 17994970-17995243 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DRC3
Entrez Gene ID
83450
Gene Name
dynein regulatory complex subunit 3
Gene Aliases
CFAP134, LRRC48
UniGene
Hs.579264
Gene Chromosome Location
Chr.17: 17972766-18016889 on Build GRCh38
Transcripts
NM_001130090.1 NM_001130091.1 NM_001130092.1 NM_031294.3 XM_006721583.3 XM_011524018.2 XM_011524019.2 XM_011524020.2 XM_011524021.2 XM_011524022.2 XM_011524023.2 XM_011524025.2 XM_011524026.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4064429 c.757A>G p.M253V Chr.17: 17995044-17995044 on Build GRCh38
5910934 c.824+5C>T p.? Chr.17: 17995116-17995116 on Build GRCh38
4420000 c.713C>T p.T238I Chr.17: 17995000-17995000 on Build GRCh38
4420001 c.713C>T p.T238I Chr.17: 17995000-17995000 on Build GRCh38
4064428 c.757A>G p.M253V Chr.17: 17995044-17995044 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550474872 Chr.17: 17995124-17995124 on Build GRCh38
rs764932947 Chr.17: 17995094-17995094 on Build GRCh38
rs370425560 Chr.17: 17995004-17995004 on Build GRCh38
rs760734762 Chr.17: 17995074-17995074 on Build GRCh38
rs753600788 Chr.17: 17995082-17995082 on Build GRCh38
rs772135761 Chr.17: 17995147-17995147 on Build GRCh38
rs775375793 Chr.17: 17995072-17995072 on Build GRCh38
rs111309590 Chr.17: 17995065-17995065 on Build GRCh38
rs755670366 Chr.17: 17995046-17995046 on Build GRCh38
rs755842777 Chr.17: 17994997-17994997 on Build GRCh38
rs763629865 Chr.17: 17995089-17995089 on Build GRCh38
rs763786025 Chr.17: 17995079-17995079 on Build GRCh38
rs986062217 Chr.17: 17995148-17995148 on Build GRCh38
rs377575498 Chr.17: 17995003-17995003 on Build GRCh38
rs745329732 Chr.17: 17995023-17995023 on Build GRCh38
rs761543661 Chr.17: 17995092-17995092 on Build GRCh38
rs749630144 Chr.17: 17995050-17995050 on Build GRCh38
rs767633482 Chr.17: 17995066-17995066 on Build GRCh38
rs746128692 Chr.17: 17995134-17995134 on Build GRCh38
rs748725764 Chr.17: 17995039-17995039 on Build GRCh38
rs569342179 Chr.17: 17995179-17995179 on Build GRCh38
rs771310302 Chr.17: 17995131-17995131 on Build GRCh38
rs770046460 Chr.17: 17995045-17995045 on Build GRCh38
rs778711480 Chr.17: 17995015-17995015 on Build GRCh38
rs1051327481 Chr.17: 17995178-17995178 on Build GRCh38
rs756584085 Chr.17: 17995117-17995117 on Build GRCh38
rs775615688 Chr.17: 17995162-17995175 on Build GRCh38
rs771078867 Chr.17: 17995058-17995058 on Build GRCh38
rs549599488 Chr.17: 17995108-17995108 on Build GRCh38
rs533440470 Chr.17: 17995049-17995049 on Build GRCh38
rs750149905 Chr.17: 17995095-17995095 on Build GRCh38
rs763990104 Chr.17: 17995002-17995002 on Build GRCh38
rs73303853 Chr.17: 17995031-17995031 on Build GRCh38
rs774591992 Chr.17: 17995064-17995064 on Build GRCh38
rs753129002 Chr.17: 17995115-17995115 on Build GRCh38
rs779089812 Chr.17: 17995132-17995132 on Build GRCh38
rs749627432 Chr.17: 17995127-17995127 on Build GRCh38
rs118118204 Chr.17: 17995217-17995217 on Build GRCh38
rs752657885 Chr.17: 17994996-17994996 on Build GRCh38
rs772135861 Chr.17: 17995142-17995169 on Build GRCh38
rs779773820 Chr.17: 17995111-17995111 on Build GRCh38
rs547331351 Chr.17: 17995027-17995027 on Build GRCh38

View Full Product Details