Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 117197604-117197835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNK18
Entrez Gene ID
338567
Gene Name
potassium two pore domain channel subfamily K member 18
Gene Aliases
K2p18.1, MGR13, TRESK, TRESK-2, TRESK2, TRIK
UniGene
Hs.449650
Gene Chromosome Location
Chr.10: 117197489-117210299 on Build GRCh38
Transcripts
NM_181840.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5359899 c.138C>T p.D46D Chr.10: 117197626-117197626 on Build GRCh38
7211491 c.141C>T p.G47G Chr.10: 117197629-117197629 on Build GRCh38
4607458 c.198C>A p.C66* Chr.10: 117197686-117197686 on Build GRCh38
6691315 c.184T>C p.L62L Chr.10: 117197672-117197672 on Build GRCh38
1346045 c.204C>A p.I68I Chr.10: 117197692-117197692 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199695275 Chr.10: 117197748-117197748 on Build GRCh38
rs771320769 Chr.10: 117197730-117197730 on Build GRCh38
rs369539052 Chr.10: 117197668-117197668 on Build GRCh38
rs747538877 Chr.10: 117197729-117197729 on Build GRCh38
rs3909165 Chr.10: 117197661-117197661 on Build GRCh38
rs201234152 Chr.10: 117197751-117197751 on Build GRCh38
rs372715018 Chr.10: 117197724-117197724 on Build GRCh38
rs370120362 Chr.10: 117197749-117197749 on Build GRCh38
rs75191586 Chr.10: 117197778-117197778 on Build GRCh38
rs751470844 Chr.10: 117197627-117197627 on Build GRCh38
rs767448125 Chr.10: 117197759-117197759 on Build GRCh38
rs761689670 Chr.10: 117197636-117197636 on Build GRCh38
rs1043428936 Chr.10: 117197787-117197787 on Build GRCh38
rs760711955 Chr.10: 117197762-117197762 on Build GRCh38
rs926493090 Chr.10: 117197739-117197739 on Build GRCh38
rs547522612 Chr.10: 117197760-117197760 on Build GRCh38
rs763203779 Chr.10: 117197758-117197758 on Build GRCh38
rs992232156 Chr.10: 117197767-117197767 on Build GRCh38
rs145257650 Chr.10: 117197693-117197693 on Build GRCh38
rs754699017 Chr.10: 117197689-117197689 on Build GRCh38
rs756073568 Chr.10: 117197653-117197653 on Build GRCh38
rs149711267 Chr.10: 117197626-117197626 on Build GRCh38
rs945116145 Chr.10: 117197786-117197786 on Build GRCh38
rs898814885 Chr.10: 117197625-117197625 on Build GRCh38
rs201294595 Chr.10: 117197709-117197709 on Build GRCh38
rs531334166 Chr.10: 117197706-117197706 on Build GRCh38
rs1033332692 Chr.10: 117197624-117197624 on Build GRCh38
rs767366005 Chr.10: 117197642-117197642 on Build GRCh38
rs201128727 Chr.10: 117197715-117197715 on Build GRCh38
rs753262034 Chr.10: 117197673-117197673 on Build GRCh38

View Full Product Details