Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 42935719-42935954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PSG7
Entrez Gene ID
5676
Gene Name
pregnancy specific beta-1-glycoprotein 7 (gene/pseudogene)
Gene Aliases
PSBG-7, PSG1, PSGGA
UniGene
Hs.709203
Gene Chromosome Location
Chr.19: 42924132-42937178 on Build GRCh38
Transcripts
NM_001206650.1 NM_001290042.1 NM_002783.2

SNP Detail

SNP ID Chromosome Location
rs759784175 Chr.19: 42935881-42935882 on Build GRCh38
rs879074635 Chr.19: 42935874-42935879 on Build GRCh38
rs752278735 Chr.19: 42935878-42935881 on Build GRCh38
rs541089156 Chr.19: 42935911-42935912 on Build GRCh38
rs1020546130 Chr.19: 42935872-42935887 on Build GRCh38
rs562990413 Chr.19: 42935847-42935847 on Build GRCh38
rs12972530 Chr.19: 42935919-42935919 on Build GRCh38
rs201975042 Chr.19: 42935911-42935911 on Build GRCh38
rs544492104 Chr.19: 42935844-42935844 on Build GRCh38
rs879210033 Chr.19: 42935880-42935881 on Build GRCh38
rs67860553 Chr.19: 42935874-42935875 on Build GRCh38
rs774031849 Chr.19: 42935905-42935905 on Build GRCh38
rs147024666 Chr.19: 42935916-42935917 on Build GRCh38
rs957548241 Chr.19: 42935842-42935842 on Build GRCh38
rs376226389 Chr.19: 42935873-42935874 on Build GRCh38
rs559412093 Chr.19: 42935887-42935887 on Build GRCh38
rs62110917 Chr.19: 42935851-42935851 on Build GRCh38
rs781126261 Chr.19: 42935906-42935915 on Build GRCh38
rs777571643 Chr.19: 42935908-42935909 on Build GRCh38
rs879158891 Chr.19: 42935877-42935878 on Build GRCh38
rs757949946 Chr.19: 42935874-42935881 on Build GRCh38
rs552018021 Chr.19: 42935854-42935854 on Build GRCh38
rs60833164 Chr.19: 42935913-42935914 on Build GRCh38
rs756415655 Chr.19: 42935872-42935881 on Build GRCh38
rs867129679 Chr.19: 42935897-42935897 on Build GRCh38
rs867182858 Chr.19: 42935917-42935917 on Build GRCh38
rs878879460 Chr.19: 42935878-42935879 on Build GRCh38
rs12972343 Chr.19: 42935915-42935915 on Build GRCh38
rs761615663 Chr.19: 42935907-42935907 on Build GRCh38
rs981717064 Chr.19: 42935902-42935915 on Build GRCh38
rs766820029 Chr.19: 42935873-42935876 on Build GRCh38
rs57083429 Chr.19: 42935914-42935915 on Build GRCh38
rs967996432 Chr.19: 42935895-42935895 on Build GRCh38
rs72150962 Chr.19: 42935874-42935877 on Build GRCh38

View Full Product Details