Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 198264048-198264319 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NEK7
Entrez Gene ID
140609
Gene Name
NIMA related kinase 7
Gene Aliases
-
UniGene
Hs.24119
Gene Chromosome Location
Chr.1: 198156978-198322420 on Build GRCh38
Transcripts
NM_133494.2 XM_011509209.1 XM_017000344.1 XM_017000345.1 XM_017000346.1 XM_017000347.1 XM_017000348.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6418527 c.333G>T p.L111F Chr.1: 198264196-198264196 on Build GRCh38
4932411 c.318A>G p.E106E Chr.1: 198264181-198264181 on Build GRCh38
1337639 c.288A>C p.K96N Chr.1: 198264151-198264151 on Build GRCh38
1127194 c.336A>G p.E112E Chr.1: 198264199-198264199 on Build GRCh38
901851 c.303C>A p.F101L Chr.1: 198264166-198264166 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748691722 Chr.1: 198264257-198264257 on Build GRCh38
rs757089870 Chr.1: 198264168-198264168 on Build GRCh38
rs772419293 Chr.1: 198264267-198264267 on Build GRCh38
rs747840642 Chr.1: 198264281-198264281 on Build GRCh38
rs148351907 Chr.1: 198264277-198264277 on Build GRCh38
rs774657545 Chr.1: 198264099-198264099 on Build GRCh38
rs532165923 Chr.1: 198264272-198264272 on Build GRCh38
rs368887107 Chr.1: 198264251-198264251 on Build GRCh38
rs1007965058 Chr.1: 198264276-198264276 on Build GRCh38
rs773147678 Chr.1: 198264276-198264277 on Build GRCh38
rs113742135 Chr.1: 198264287-198264287 on Build GRCh38
rs751598241 Chr.1: 198264158-198264158 on Build GRCh38
rs769153349 Chr.1: 198264098-198264098 on Build GRCh38
rs773448263 Chr.1: 198264109-198264109 on Build GRCh38
rs764152220 Chr.1: 198264142-198264142 on Build GRCh38
rs867302189 Chr.1: 198264215-198264215 on Build GRCh38
rs543325421 Chr.1: 198264157-198264157 on Build GRCh38
rs750075203 Chr.1: 198264187-198264187 on Build GRCh38
rs377101584 Chr.1: 198264286-198264286 on Build GRCh38
rs760869047 Chr.1: 198264112-198264112 on Build GRCh38
rs773465955 Chr.1: 198264271-198264271 on Build GRCh38
rs776043157 Chr.1: 198264269-198264272 on Build GRCh38
rs932508017 Chr.1: 198264247-198264247 on Build GRCh38
rs934612074 Chr.1: 198264106-198264106 on Build GRCh38
rs765215568 Chr.1: 198264272-198264273 on Build GRCh38
rs780917196 Chr.1: 198264175-198264175 on Build GRCh38
rs768329832 Chr.1: 198264261-198264265 on Build GRCh38
rs748779876 Chr.1: 198264219-198264219 on Build GRCh38
rs772471999 Chr.1: 198264108-198264108 on Build GRCh38
rs755735209 Chr.1: 198264200-198264200 on Build GRCh38
rs61742184 Chr.1: 198264214-198264214 on Build GRCh38
rs372818218 Chr.1: 198264128-198264128 on Build GRCh38
rs536448832 Chr.1: 198264253-198264253 on Build GRCh38
rs987131962 Chr.1: 198264239-198264239 on Build GRCh38
rs370944444 Chr.1: 198264273-198264273 on Build GRCh38
rs762185049 Chr.1: 198264102-198264102 on Build GRCh38
rs114741246 Chr.1: 198264136-198264136 on Build GRCh38
rs753873003 Chr.1: 198264132-198264132 on Build GRCh38

View Full Product Details