Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26204522-26204791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIST1H4E
Entrez Gene ID
8367
Gene Name
histone cluster 1, H4e
Gene Aliases
H4/j, H4FJ
UniGene
Hs.662174
Gene Chromosome Location
Chr.6: 26204645-26205021 on Build GRCh38
Transcripts
NM_003545.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5578711 c.37A>T p.K13* Chr.6: 26204681-26204681 on Build GRCh38
5623272 c.73G>C p.D25H Chr.6: 26204717-26204717 on Build GRCh38
5442185 c.97C>T p.P33S Chr.6: 26204741-26204741 on Build GRCh38
3829831 c.33G>A p.L11L Chr.6: 26204677-26204677 on Build GRCh38
5712355 c.81C>A p.I27I Chr.6: 26204725-26204725 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755908454 Chr.6: 26204699-26204700 on Build GRCh38
rs758381758 Chr.6: 26204760-26204760 on Build GRCh38
rs758146321 Chr.6: 26204716-26204716 on Build GRCh38
rs760413679 Chr.6: 26204568-26204568 on Build GRCh38
rs923049498 Chr.6: 26204574-26204574 on Build GRCh38
rs572544472 Chr.6: 26204612-26204612 on Build GRCh38
rs1056929799 Chr.6: 26204593-26204593 on Build GRCh38
rs539637729 Chr.6: 26204752-26204752 on Build GRCh38
rs761425139 Chr.6: 26204630-26204637 on Build GRCh38
rs747179580 Chr.6: 26204727-26204727 on Build GRCh38
rs765793762 Chr.6: 26204710-26204710 on Build GRCh38
rs751723647 Chr.6: 26204646-26204646 on Build GRCh38
rs61742995 Chr.6: 26204609-26204609 on Build GRCh38
rs750010829 Chr.6: 26204706-26204706 on Build GRCh38
rs956929051 Chr.6: 26204561-26204561 on Build GRCh38
rs774516460 Chr.6: 26204633-26204633 on Build GRCh38
rs767212732 Chr.6: 26204663-26204663 on Build GRCh38
rs577353282 Chr.6: 26204591-26204591 on Build GRCh38
rs370851019 Chr.6: 26204630-26204630 on Build GRCh38
rs779799415 Chr.6: 26204647-26204647 on Build GRCh38
rs985825510 Chr.6: 26204542-26204542 on Build GRCh38
rs144621549 Chr.6: 26204644-26204644 on Build GRCh38
rs745781167 Chr.6: 26204627-26204627 on Build GRCh38
rs760423117 Chr.6: 26204707-26204707 on Build GRCh38
rs754219809 Chr.6: 26204620-26204620 on Build GRCh38
rs754966292 Chr.6: 26204675-26204675 on Build GRCh38
rs767186911 Chr.6: 26204667-26204668 on Build GRCh38
rs755252848 Chr.6: 26204642-26204642 on Build GRCh38
rs760610500 Chr.6: 26204652-26204652 on Build GRCh38
rs377601396 Chr.6: 26204603-26204603 on Build GRCh38
rs765678494 Chr.6: 26204658-26204658 on Build GRCh38
rs777420822 Chr.6: 26204767-26204767 on Build GRCh38
rs749157094 Chr.6: 26204672-26204672 on Build GRCh38
rs372683763 Chr.6: 26204606-26204606 on Build GRCh38
rs143652738 Chr.6: 26204679-26204679 on Build GRCh38
rs753454645 Chr.6: 26204711-26204711 on Build GRCh38
rs140853277 Chr.6: 26204722-26204722 on Build GRCh38
rs892076716 Chr.6: 26204674-26204674 on Build GRCh38
rs145699171 Chr.6: 26204619-26204619 on Build GRCh38
rs768496913 Chr.6: 26204585-26204585 on Build GRCh38
rs533676183 Chr.6: 26204730-26204730 on Build GRCh38
rs770490166 Chr.6: 26204695-26204695 on Build GRCh38
rs376058980 Chr.6: 26204631-26204631 on Build GRCh38
rs199914113 Chr.6: 26204650-26204650 on Build GRCh38
rs751229941 Chr.6: 26204643-26204643 on Build GRCh38
rs763354325 Chr.6: 26204702-26204702 on Build GRCh38
rs778674883 Chr.6: 26204677-26204677 on Build GRCh38
rs770008463 Chr.6: 26204732-26204732 on Build GRCh38
rs895945395 Chr.6: 26204595-26204595 on Build GRCh38
rs368745174 Chr.6: 26204713-26204713 on Build GRCh38
rs201319649 Chr.6: 26204665-26204665 on Build GRCh38
rs375429854 Chr.6: 26204543-26204543 on Build GRCh38
rs376989143 Chr.6: 26204726-26204726 on Build GRCh38
rs752602906 Chr.6: 26204759-26204759 on Build GRCh38
rs184503150 Chr.6: 26204645-26204645 on Build GRCh38
rs776594935 Chr.6: 26204599-26204599 on Build GRCh38
rs571073582 Chr.6: 26204749-26204749 on Build GRCh38
rs751650802 Chr.6: 26204598-26204598 on Build GRCh38
rs755370533 Chr.6: 26204622-26204622 on Build GRCh38
rs201304438 Chr.6: 26204649-26204649 on Build GRCh38
rs199747912 Chr.6: 26204600-26204600 on Build GRCh38
rs749057585 Chr.6: 26204616-26204616 on Build GRCh38
rs774835775 Chr.6: 26204746-26204746 on Build GRCh38
rs762062147 Chr.6: 26204634-26204634 on Build GRCh38
rs753083420 Chr.6: 26204754-26204754 on Build GRCh38
rs752651799 Chr.6: 26204715-26204715 on Build GRCh38
rs144595768 Chr.6: 26204731-26204731 on Build GRCh38
rs779653681 Chr.6: 26204671-26204671 on Build GRCh38
rs777920279 Chr.6: 26204683-26204683 on Build GRCh38
rs147263244 Chr.6: 26204704-26204704 on Build GRCh38
rs919154679 Chr.6: 26204576-26204576 on Build GRCh38
rs61742993 Chr.6: 26204605-26204605 on Build GRCh38
rs941981311 Chr.6: 26204555-26204555 on Build GRCh38
rs753173805 Chr.6: 26204659-26204659 on Build GRCh38
rs1000899401 Chr.6: 26204728-26204728 on Build GRCh38
rs776145717 Chr.6: 26204696-26204696 on Build GRCh38
rs374139942 Chr.6: 26204611-26204611 on Build GRCh38
rs750253540 Chr.6: 26204675-26204676 on Build GRCh38
rs377665213 Chr.6: 26204596-26204596 on Build GRCh38
rs758993886 Chr.6: 26204755-26204755 on Build GRCh38
rs867918860 Chr.6: 26204588-26204588 on Build GRCh38
rs766724515 Chr.6: 26204640-26204640 on Build GRCh38
rs770887499 Chr.6: 26204687-26204687 on Build GRCh38
rs201324703 Chr.6: 26204641-26204641 on Build GRCh38
rs746685515 Chr.6: 26204624-26204624 on Build GRCh38
rs963122578 Chr.6: 26204733-26204733 on Build GRCh38
rs976235822 Chr.6: 26204572-26204572 on Build GRCh38
rs371154912 Chr.6: 26204610-26204610 on Build GRCh38
rs368144763 Chr.6: 26204638-26204638 on Build GRCh38
rs369563507 Chr.6: 26204571-26204571 on Build GRCh38
rs372764747 Chr.6: 26204714-26204714 on Build GRCh38
rs373816561 Chr.6: 26204601-26204601 on Build GRCh38
rs202183095 Chr.6: 26204689-26204689 on Build GRCh38
rs763587341 Chr.6: 26204737-26204737 on Build GRCh38
rs764213763 Chr.6: 26204597-26204597 on Build GRCh38
rs766105580 Chr.6: 26204727-26204727 on Build GRCh38
rs775282215 Chr.6: 26204558-26204558 on Build GRCh38
rs769679868 Chr.6: 26204629-26204629 on Build GRCh38
rs747289620 Chr.6: 26204686-26204686 on Build GRCh38
rs139616312 Chr.6: 26204656-26204656 on Build GRCh38
rs776825708 Chr.6: 26204688-26204688 on Build GRCh38
rs931635393 Chr.6: 26204575-26204575 on Build GRCh38
rs773883466 Chr.6: 26204609-26204609 on Build GRCh38
rs540442619 Chr.6: 26204618-26204618 on Build GRCh38
rs769118146 Chr.6: 26204740-26204740 on Build GRCh38
rs776092771 Chr.6: 26204734-26204734 on Build GRCh38
rs773423854 Chr.6: 26204651-26204651 on Build GRCh38
rs532184358 Chr.6: 26204623-26204623 on Build GRCh38
rs763732529 Chr.6: 26204603-26204604 on Build GRCh38
rs778951387 Chr.6: 26204578-26204595 on Build GRCh38
rs368355176 Chr.6: 26204626-26204626 on Build GRCh38
rs373542986 Chr.6: 26204594-26204594 on Build GRCh38
rs747867488 Chr.6: 26204678-26204678 on Build GRCh38
rs879202820 Chr.6: 26204608-26204608 on Build GRCh38
rs1050020798 Chr.6: 26204581-26204581 on Build GRCh38
rs768482468 Chr.6: 26204748-26204748 on Build GRCh38

View Full Product Details