Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 77390923-77391150 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SAMD15
Entrez Gene ID
161394
Gene Name
sterile alpha motif domain containing 15
Gene Aliases
C14orf174, FAM15A
UniGene
Hs.421961
Gene Chromosome Location
Chr.14: 77377419-77391244 on Build GRCh38
Transcripts
NM_001010860.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1198079 c.1843C>T p.R615C Chr.14: 77391062-77391062 on Build GRCh38
5547639 c.1797_1817del21 p.R600_L606delRHTQELL Chr.14: 77391016-77391036 on Build GRCh38
4052602 c.1825G>A p.E609K Chr.14: 77391044-77391044 on Build GRCh38
1172231 c.1844G>A p.R615H Chr.14: 77391063-77391063 on Build GRCh38
344322 c.1900C>G p.H634D Chr.14: 77391119-77391119 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753965663 Chr.14: 77390996-77390996 on Build GRCh38
rs939528375 Chr.14: 77391011-77391011 on Build GRCh38
rs778321093 Chr.14: 77391019-77391019 on Build GRCh38
rs1010909448 Chr.14: 77391049-77391049 on Build GRCh38
rs143117906 Chr.14: 77391111-77391111 on Build GRCh38
rs771978459 Chr.14: 77391065-77391065 on Build GRCh38
rs759665289 Chr.14: 77391000-77391000 on Build GRCh38
rs144825571 Chr.14: 77391084-77391084 on Build GRCh38
rs746849584 Chr.14: 77391068-77391068 on Build GRCh38
rs779191520 Chr.14: 77391000-77391000 on Build GRCh38
rs906677734 Chr.14: 77391010-77391010 on Build GRCh38
rs267604065 Chr.14: 77391095-77391095 on Build GRCh38
rs145050832 Chr.14: 77391059-77391059 on Build GRCh38
rs759815065 Chr.14: 77391094-77391094 on Build GRCh38
rs764718175 Chr.14: 77391116-77391116 on Build GRCh38
rs551937879 Chr.14: 77391067-77391067 on Build GRCh38
rs751928457 Chr.14: 77391029-77391029 on Build GRCh38
rs867283016 Chr.14: 77391042-77391042 on Build GRCh38
rs201034938 Chr.14: 77391018-77391018 on Build GRCh38
rs748835296 Chr.14: 77391050-77391050 on Build GRCh38
rs267604064 Chr.14: 77391058-77391058 on Build GRCh38
rs542658833 Chr.14: 77391001-77391001 on Build GRCh38
rs149116567 Chr.14: 77391062-77391062 on Build GRCh38
rs779762258 Chr.14: 77391043-77391043 on Build GRCh38
rs775679700 Chr.14: 77391106-77391106 on Build GRCh38
rs770562347 Chr.14: 77391083-77391083 on Build GRCh38
rs766643382 Chr.14: 77390976-77390976 on Build GRCh38
rs200429320 Chr.14: 77391032-77391032 on Build GRCh38
rs778395006 Chr.14: 77391061-77391061 on Build GRCh38
rs753314157 Chr.14: 77391017-77391017 on Build GRCh38

View Full Product Details