Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66558151-66558416 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CKLF
Entrez Gene ID
51192
Gene Name
chemokine-like factor
Gene Aliases
C32, CKLF1, CKLF2, CKLF3, CKLF4, HSPC224, UCK-1
UniGene
Hs.15159
Gene Chromosome Location
Chr.16: 66552563-66566287 on Build GRCh38
Transcripts
NM_001040138.2 NM_016326.3 NM_016951.3 NM_181640.2 NM_181641.2 NM_001202509.1 NM_001204098.1 NM_001204099.1

Gene Symbol
CKLF-CMTM1
Entrez Gene ID
100529251
Gene Name
CKLF-CMTM1 readthrough
Gene Aliases
-
UniGene
Hs.698621
Gene Chromosome Location
Chr.16: 66552563-66579135 on Build GRCh38
Transcripts
NM_001040138.2 NM_016326.3 NM_016951.3 NM_181640.2 NM_181641.2 NM_001202509.1 NM_001204098.1 NM_001204099.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6629595 c.213G>T p.M71I Chr.16: 66558324-66558324 on Build GRCh38
6566909 c.159C>G p.V53V Chr.16: 66558270-66558270 on Build GRCh38
703936 c.168C>G p.I56M Chr.16: 66558279-66558279 on Build GRCh38
435491 c.156A>T p.E52D Chr.16: 66558267-66558267 on Build GRCh38
6629596 c.206G>A p.R69Q Chr.16: 66558317-66558317 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780766283 Chr.16: 66558349-66558349 on Build GRCh38
rs770812519 Chr.16: 66558226-66558226 on Build GRCh38
rs752670451 Chr.16: 66558300-66558300 on Build GRCh38
rs767635041 Chr.16: 66558277-66558277 on Build GRCh38
rs770326370 Chr.16: 66558378-66558378 on Build GRCh38
rs746507378 Chr.16: 66558317-66558317 on Build GRCh38
rs112815434 Chr.16: 66558273-66558273 on Build GRCh38
rs759581072 Chr.16: 66558281-66558281 on Build GRCh38
rs746217307 Chr.16: 66558359-66558359 on Build GRCh38
rs749047796 Chr.16: 66558204-66558204 on Build GRCh38
rs202172549 Chr.16: 66558255-66558255 on Build GRCh38
rs762035736 Chr.16: 66558274-66558274 on Build GRCh38
rs375374500 Chr.16: 66558245-66558245 on Build GRCh38
rs758362680 Chr.16: 66558311-66558311 on Build GRCh38
rs533965480 Chr.16: 66558316-66558316 on Build GRCh38
rs950194385 Chr.16: 66558201-66558201 on Build GRCh38
rs550053000 Chr.16: 66558250-66558250 on Build GRCh38
rs916200094 Chr.16: 66558198-66558198 on Build GRCh38
rs747939430 Chr.16: 66558349-66558350 on Build GRCh38
rs753659189 Chr.16: 66558279-66558279 on Build GRCh38
rs776989604 Chr.16: 66558241-66558241 on Build GRCh38
rs200692066 Chr.16: 66558320-66558320 on Build GRCh38
rs960012397 Chr.16: 66558367-66558367 on Build GRCh38
rs751850810 Chr.16: 66558176-66558176 on Build GRCh38
rs561301088 Chr.16: 66558186-66558186 on Build GRCh38
rs78927166 Chr.16: 66558338-66558338 on Build GRCh38
rs552134291 Chr.16: 66558329-66558329 on Build GRCh38
rs754840375 Chr.16: 66558208-66558208 on Build GRCh38
rs992414407 Chr.16: 66558368-66558368 on Build GRCh38
rs771618753 Chr.16: 66558225-66558225 on Build GRCh38
rs757454047 Chr.16: 66558183-66558183 on Build GRCh38
rs139678335 Chr.16: 66558259-66558259 on Build GRCh38
rs778885441 Chr.16: 66558215-66558215 on Build GRCh38
rs778799854 Chr.16: 66558296-66558296 on Build GRCh38
rs746630783 Chr.16: 66558228-66558228 on Build GRCh38
rs374608984 Chr.16: 66558292-66558292 on Build GRCh38
rs781098477 Chr.16: 66558192-66558195 on Build GRCh38
rs199961678 Chr.16: 66558247-66558247 on Build GRCh38
rs747643263 Chr.16: 66558217-66558217 on Build GRCh38
rs371342035 Chr.16: 66558234-66558234 on Build GRCh38

View Full Product Details